Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76117532-76118122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP54
Entrez Gene ID
53371
Gene Name
nucleoporin 54
Gene Aliases
-
UniGene
Hs.430435
Gene Chromosome Location
Chr.4: 76114659-76148515 on Build GRCh38
Transcripts
NM_001278603.1 NM_017426.3 NR_103780.1 NR_103781.1 NR_103782.1 XM_006714237.1 XM_011532033.2 XM_011532034.2 XM_017008294.1 XM_017008295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057623 c.1395+1G>A p.? Chr.4: 76117663-76117663 on Build GRCh38
1486102 c.1331_1332delTT p.F444fs*6 Chr.4: 76117727-76117728 on Build GRCh38
3767989 c.1344A>T p.R448S Chr.4: 76117715-76117715 on Build GRCh38
22089 c.1246A>G p.T416A Chr.4: 76118113-76118113 on Build GRCh38
5662273 c.1371A>T p.A457A Chr.4: 76117688-76117688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370887575 Chr.4: 76117696-76117696 on Build GRCh38
rs776805229 Chr.4: 76117824-76117824 on Build GRCh38
rs1026289044 Chr.4: 76117735-76117735 on Build GRCh38
rs868192392 Chr.4: 76117888-76117888 on Build GRCh38
rs1018652457 Chr.4: 76118050-76118050 on Build GRCh38
rs965684354 Chr.4: 76118056-76118056 on Build GRCh38
rs754838344 Chr.4: 76117811-76117811 on Build GRCh38
rs749029158 Chr.4: 76117677-76117677 on Build GRCh38
rs755481769 Chr.4: 76117882-76117885 on Build GRCh38
rs1041854205 Chr.4: 76117950-76117950 on Build GRCh38
rs745895639 Chr.4: 76117826-76117826 on Build GRCh38
rs1032637681 Chr.4: 76117703-76117703 on Build GRCh38
rs1047314822 Chr.4: 76118011-76118011 on Build GRCh38
rs532597994 Chr.4: 76117596-76117596 on Build GRCh38
rs895879873 Chr.4: 76117910-76117910 on Build GRCh38
rs769971976 Chr.4: 76118024-76118024 on Build GRCh38
rs541919016 Chr.4: 76117967-76117967 on Build GRCh38
rs375182485 Chr.4: 76118002-76118002 on Build GRCh38
rs760220228 Chr.4: 76117831-76117831 on Build GRCh38
rs369635113 Chr.4: 76117885-76117885 on Build GRCh38
rs768188121 Chr.4: 76117877-76117877 on Build GRCh38
rs1028717246 Chr.4: 76118094-76118094 on Build GRCh38
rs6532201 Chr.4: 76117821-76117821 on Build GRCh38
rs1056905356 Chr.4: 76117934-76117934 on Build GRCh38
rs898652993 Chr.4: 76117661-76117661 on Build GRCh38
rs898682373 Chr.4: 76117997-76117997 on Build GRCh38
rs1014406005 Chr.4: 76117618-76117620 on Build GRCh38
rs144785979 Chr.4: 76117759-76117759 on Build GRCh38
rs763461131 Chr.4: 76117786-76117786 on Build GRCh38
rs571733387 Chr.4: 76117986-76117986 on Build GRCh38
rs1001529825 Chr.4: 76117583-76117583 on Build GRCh38
rs754691244 Chr.4: 76117697-76117698 on Build GRCh38
rs919094564 Chr.4: 76117833-76117833 on Build GRCh38
rs556319053 Chr.4: 76117891-76117891 on Build GRCh38
rs182063729 Chr.4: 76117797-76117797 on Build GRCh38
rs181930810 Chr.4: 76118069-76118069 on Build GRCh38
rs148816508 Chr.4: 76117604-76117604 on Build GRCh38
rs374613073 Chr.4: 76118098-76118098 on Build GRCh38
rs767057920 Chr.4: 76117795-76117795 on Build GRCh38
rs921784395 Chr.4: 76117846-76117846 on Build GRCh38
rs376613793 Chr.4: 76117700-76117700 on Build GRCh38
rs935347032 Chr.4: 76117927-76117927 on Build GRCh38
rs868720228 Chr.4: 76117577-76117577 on Build GRCh38
rs1048357715 Chr.4: 76117568-76117568 on Build GRCh38
rs369999824 Chr.4: 76117623-76117623 on Build GRCh38
rs745829518 Chr.4: 76117656-76117656 on Build GRCh38
rs780808251 Chr.4: 76117777-76117781 on Build GRCh38
rs756857152 Chr.4: 76117638-76117638 on Build GRCh38
rs538867003 Chr.4: 76117568-76117571 on Build GRCh38
rs545374106 Chr.4: 76117993-76117993 on Build GRCh38
rs751283910 Chr.4: 76117788-76117788 on Build GRCh38
rs370268147 Chr.4: 76117796-76117796 on Build GRCh38
rs919159583 Chr.4: 76117566-76117566 on Build GRCh38
rs765801175 Chr.4: 76117783-76117783 on Build GRCh38
rs7657846 Chr.4: 76117654-76117654 on Build GRCh38
rs749664047 Chr.4: 76117788-76117789 on Build GRCh38
rs1049448626 Chr.4: 76117864-76117864 on Build GRCh38
rs6840007 Chr.4: 76118040-76118040 on Build GRCh38
rs6813732 Chr.4: 76118021-76118021 on Build GRCh38
rs377634247 Chr.4: 76117678-76117678 on Build GRCh38
rs747196062 Chr.4: 76117713-76117713 on Build GRCh38
rs572483387 Chr.4: 76118096-76118096 on Build GRCh38
rs191815270 Chr.4: 76118028-76118028 on Build GRCh38
rs776019180 Chr.4: 76117784-76117784 on Build GRCh38
rs764361490 Chr.4: 76117787-76117787 on Build GRCh38
rs779852833 Chr.4: 76117672-76117672 on Build GRCh38
rs749189335 Chr.4: 76117809-76117809 on Build GRCh38
rs770995638 Chr.4: 76117822-76117822 on Build GRCh38
rs778563273 Chr.4: 76117812-76117812 on Build GRCh38
rs937328811 Chr.4: 76117889-76117889 on Build GRCh38
rs776518357 Chr.4: 76117766-76117766 on Build GRCh38
rs760127132 Chr.4: 76117771-76117771 on Build GRCh38
rs774511348 Chr.4: 76118059-76118059 on Build GRCh38
rs11944780 Chr.4: 76117903-76117903 on Build GRCh38
rs569295613 Chr.4: 76117627-76117627 on Build GRCh38
rs187341694 Chr.4: 76117983-76117983 on Build GRCh38
rs756800683 Chr.4: 76117792-76117792 on Build GRCh38
rs1014766756 Chr.4: 76117914-76117916 on Build GRCh38
rs768414670 Chr.4: 76117691-76117691 on Build GRCh38
rs755977899 Chr.4: 76117660-76117660 on Build GRCh38
rs780012208 Chr.4: 76117803-76117803 on Build GRCh38

View Full Product Details