Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155903542-155904141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDO2
Entrez Gene ID
6999
Gene Name
tryptophan 2,3-dioxygenase
Gene Aliases
TDO, TO, TPH2, TRPO
UniGene
Hs.183671
Gene Chromosome Location
Chr.4: 155903693-155920406 on Build GRCh38
Transcripts
NM_005651.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545876 c.94G>T p.V32L Chr.4: 155904076-155904076 on Build GRCh38
3129772 c.112G>T p.G38* Chr.4: 155904094-155904094 on Build GRCh38
6798968 c.89C>A p.T30N Chr.4: 155904071-155904071 on Build GRCh38
4158883 c.35G>T p.G12V Chr.4: 155903793-155903793 on Build GRCh38
5463326 c.70G>A p.E24K Chr.4: 155904052-155904052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536542437 Chr.4: 155903825-155903825 on Build GRCh38
rs755227439 Chr.4: 155904028-155904028 on Build GRCh38
rs747034548 Chr.4: 155903731-155903731 on Build GRCh38
rs774954938 Chr.4: 155903990-155903990 on Build GRCh38
rs918748992 Chr.4: 155903706-155903706 on Build GRCh38
rs905876897 Chr.4: 155903850-155903850 on Build GRCh38
rs771705026 Chr.4: 155904062-155904062 on Build GRCh38
rs956488327 Chr.4: 155903981-155903981 on Build GRCh38
rs757614756 Chr.4: 155903807-155903807 on Build GRCh38
rs757567915 Chr.4: 155903711-155903711 on Build GRCh38
rs750823258 Chr.4: 155904018-155904018 on Build GRCh38
rs771787132 Chr.4: 155903982-155903982 on Build GRCh38
rs1029533915 Chr.4: 155903888-155903888 on Build GRCh38
rs979088212 Chr.4: 155904036-155904036 on Build GRCh38
rs890943398 Chr.4: 155903900-155903900 on Build GRCh38
rs200196655 Chr.4: 155904051-155904051 on Build GRCh38
rs78544310 Chr.4: 155903758-155903758 on Build GRCh38
rs140590165 Chr.4: 155904087-155904087 on Build GRCh38
rs998007140 Chr.4: 155903859-155903859 on Build GRCh38
rs754230846 Chr.4: 155903708-155903708 on Build GRCh38
rs1045705250 Chr.4: 155903805-155903805 on Build GRCh38
rs762866821 Chr.4: 155903995-155903995 on Build GRCh38
rs979067585 Chr.4: 155903962-155903962 on Build GRCh38
rs910921101 Chr.4: 155903687-155903687 on Build GRCh38
rs766640851 Chr.4: 155904021-155904021 on Build GRCh38
rs553048258 Chr.4: 155903578-155903578 on Build GRCh38
rs182121756 Chr.4: 155903909-155903909 on Build GRCh38
rs201490914 Chr.4: 155903780-155903780 on Build GRCh38
rs376085974 Chr.4: 155903979-155903979 on Build GRCh38
rs780104976 Chr.4: 155903827-155903827 on Build GRCh38
rs765708510 Chr.4: 155904011-155904011 on Build GRCh38
rs765274359 Chr.4: 155903840-155903840 on Build GRCh38
rs769600704 Chr.4: 155904085-155904085 on Build GRCh38
rs766724263 Chr.4: 155904098-155904098 on Build GRCh38
rs760062469 Chr.4: 155903763-155903763 on Build GRCh38
rs987831362 Chr.4: 155904068-155904068 on Build GRCh38
rs746705593 Chr.4: 155904072-155904072 on Build GRCh38
rs557213593 Chr.4: 155903803-155903803 on Build GRCh38
rs144568577 Chr.4: 155904114-155904114 on Build GRCh38
rs777862065 Chr.4: 155903841-155903841 on Build GRCh38
rs773128093 Chr.4: 155903794-155903794 on Build GRCh38
rs779120590 Chr.4: 155903715-155903715 on Build GRCh38
rs779307480 Chr.4: 155903987-155903988 on Build GRCh38
rs898614257 Chr.4: 155903904-155903904 on Build GRCh38
rs373759588 Chr.4: 155903727-155903727 on Build GRCh38
rs751669460 Chr.4: 155904025-155904025 on Build GRCh38
rs767056644 Chr.4: 155903750-155903750 on Build GRCh38
rs1029871554 Chr.4: 155903596-155903596 on Build GRCh38
rs1001503146 Chr.4: 155903872-155903872 on Build GRCh38
rs924943202 Chr.4: 155904039-155904039 on Build GRCh38
rs765653357 Chr.4: 155903814-155903814 on Build GRCh38
rs370551248 Chr.4: 155903742-155903742 on Build GRCh38
rs749112799 Chr.4: 155903739-155903739 on Build GRCh38
rs377146366 Chr.4: 155903732-155903732 on Build GRCh38
rs770978628 Chr.4: 155903844-155903844 on Build GRCh38
rs368286330 Chr.4: 155903833-155903833 on Build GRCh38
rs555186898 Chr.4: 155904109-155904109 on Build GRCh38
rs751944478 Chr.4: 155904101-155904101 on Build GRCh38
rs540234095 Chr.4: 155904001-155904001 on Build GRCh38
rs1016996724 Chr.4: 155903657-155903657 on Build GRCh38
rs935174127 Chr.4: 155903810-155903810 on Build GRCh38
rs143902515 Chr.4: 155903842-155903842 on Build GRCh38
rs774338361 Chr.4: 155903972-155903972 on Build GRCh38
rs1044959533 Chr.4: 155903846-155903846 on Build GRCh38
rs1002039035 Chr.4: 155903645-155903645 on Build GRCh38
rs1030620239 Chr.4: 155903968-155903968 on Build GRCh38
rs577930501 Chr.4: 155903728-155903728 on Build GRCh38
rs776164347 Chr.4: 155904078-155904078 on Build GRCh38
rs888005679 Chr.4: 155903812-155903812 on Build GRCh38
rs373006203 Chr.4: 155903851-155903851 on Build GRCh38
rs146892100 Chr.4: 155903993-155903993 on Build GRCh38
rs535018620 Chr.4: 155903571-155903571 on Build GRCh38
rs542845167 Chr.4: 155903853-155903853 on Build GRCh38
rs754285981 Chr.4: 155903796-155903796 on Build GRCh38
rs773537340 Chr.4: 155904086-155904086 on Build GRCh38
rs202180389 Chr.4: 155904014-155904014 on Build GRCh38
rs1052265671 Chr.4: 155904090-155904090 on Build GRCh38
rs909472691 Chr.4: 155904027-155904027 on Build GRCh38
rs985774023 Chr.4: 155903681-155903681 on Build GRCh38
rs781480973 Chr.4: 155904033-155904033 on Build GRCh38
rs752803657 Chr.4: 155904040-155904040 on Build GRCh38
rs747001628 Chr.4: 155904066-155904066 on Build GRCh38
rs779760124 Chr.4: 155904064-155904064 on Build GRCh38
rs868580528 Chr.4: 155904094-155904094 on Build GRCh38
rs202202273 Chr.4: 155904117-155904117 on Build GRCh38
rs368767491 Chr.4: 155904002-155904002 on Build GRCh38
rs780343663 Chr.4: 155904036-155904037 on Build GRCh38
rs1045388641 Chr.4: 155904095-155904095 on Build GRCh38
rs770314552 Chr.4: 155904081-155904081 on Build GRCh38
rs750769784 Chr.4: 155903816-155903816 on Build GRCh38
rs772240347 Chr.4: 155904028-155904028 on Build GRCh38
rs745696595 Chr.4: 155904052-155904052 on Build GRCh38
rs967902561 Chr.4: 155903939-155903939 on Build GRCh38
rs776424867 Chr.4: 155903599-155903599 on Build GRCh38
rs759321298 Chr.4: 155903744-155903744 on Build GRCh38
rs746058534 Chr.4: 155904008-155904009 on Build GRCh38
rs761278970 Chr.4: 155904080-155904080 on Build GRCh38
rs768279926 Chr.4: 155904074-155904074 on Build GRCh38
rs767818079 Chr.4: 155904113-155904113 on Build GRCh38
rs1018157826 Chr.4: 155903912-155903912 on Build GRCh38
rs758694311 Chr.4: 155903819-155903819 on Build GRCh38
rs970071454 Chr.4: 155903644-155903644 on Build GRCh38
rs764682812 Chr.4: 155903766-155903766 on Build GRCh38
rs918069867 Chr.4: 155903759-155903759 on Build GRCh38

View Full Product Details