Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165340133-165340694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSMO1
Entrez Gene ID
6307
Gene Name
methylsterol monooxygenase 1
Gene Aliases
DESP4, ERG25, MCCPD, SC4MOL
UniGene
Hs.105269
Gene Chromosome Location
Chr.4: 165327666-165343164 on Build GRCh38
Transcripts
NM_001017369.2 NM_006745.4 XM_005263176.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1428397 c.583C>A p.L195I Chr.4: 165340272-165340272 on Build GRCh38
233304 c.625C>T p.H209Y Chr.4: 165340314-165340314 on Build GRCh38
1052948 c.653C>T p.T218I Chr.4: 165340342-165340342 on Build GRCh38
4155031 c.635T>C p.L212P Chr.4: 165340324-165340324 on Build GRCh38
6099442 c.673A>C p.I225L Chr.4: 165340362-165340362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757518565 Chr.4: 165340372-165340372 on Build GRCh38
rs73862307 Chr.4: 165340298-165340298 on Build GRCh38
rs910329914 Chr.4: 165340510-165340510 on Build GRCh38
rs991036124 Chr.4: 165340474-165340474 on Build GRCh38
rs916808969 Chr.4: 165340477-165340477 on Build GRCh38
rs776585384 Chr.4: 165340250-165340250 on Build GRCh38
rs779133528 Chr.4: 165340382-165340382 on Build GRCh38
rs370698299 Chr.4: 165340235-165340235 on Build GRCh38
rs772186774 Chr.4: 165340407-165340407 on Build GRCh38
rs780091592 Chr.4: 165340418-165340418 on Build GRCh38
rs28551032 Chr.4: 165340459-165340459 on Build GRCh38
rs1005145169 Chr.4: 165340266-165340266 on Build GRCh38
rs756643811 Chr.4: 165340343-165340343 on Build GRCh38
rs906763113 Chr.4: 165340672-165340672 on Build GRCh38
rs145987291 Chr.4: 165340362-165340362 on Build GRCh38
rs778187610 Chr.4: 165340205-165340205 on Build GRCh38
rs377232490 Chr.4: 165340221-165340221 on Build GRCh38
rs577412065 Chr.4: 165340433-165340433 on Build GRCh38
rs1008200986 Chr.4: 165340327-165340327 on Build GRCh38
rs748099513 Chr.4: 165340421-165340421 on Build GRCh38
rs774502457 Chr.4: 165340220-165340220 on Build GRCh38
rs746054574 Chr.4: 165340405-165340405 on Build GRCh38
rs73862306 Chr.4: 165340297-165340297 on Build GRCh38
rs561229765 Chr.4: 165340563-165340563 on Build GRCh38
rs977330650 Chr.4: 165340439-165340439 on Build GRCh38
rs745942800 Chr.4: 165340264-165340265 on Build GRCh38
rs200613659 Chr.4: 165340291-165340291 on Build GRCh38
rs897952493 Chr.4: 165340601-165340601 on Build GRCh38
rs941729786 Chr.4: 165340530-165340530 on Build GRCh38
rs113117381 Chr.4: 165340376-165340376 on Build GRCh38
rs142496142 Chr.4: 165340299-165340299 on Build GRCh38
rs764996963 Chr.4: 165340280-165340280 on Build GRCh38
rs560869149 Chr.4: 165340594-165340594 on Build GRCh38
rs772176638 Chr.4: 165340395-165340405 on Build GRCh38
rs779059941 Chr.4: 165340209-165340212 on Build GRCh38
rs765390986 Chr.4: 165340466-165340468 on Build GRCh38
rs985774675 Chr.4: 165340468-165340468 on Build GRCh38
rs889793163 Chr.4: 165340310-165340310 on Build GRCh38
rs949088768 Chr.4: 165340478-165340480 on Build GRCh38
rs1550270 Chr.4: 165340648-165340648 on Build GRCh38
rs7679322 Chr.4: 165340437-165340437 on Build GRCh38
rs778078579 Chr.4: 165340357-165340357 on Build GRCh38
rs763376996 Chr.4: 165340629-165340629 on Build GRCh38
rs200947752 Chr.4: 165340328-165340328 on Build GRCh38
rs929458011 Chr.4: 165340632-165340632 on Build GRCh38
rs749543337 Chr.4: 165340207-165340207 on Build GRCh38
rs7679808 Chr.4: 165340457-165340457 on Build GRCh38
rs759157226 Chr.4: 165340317-165340317 on Build GRCh38
rs542616492 Chr.4: 165340504-165340504 on Build GRCh38
rs112843723 Chr.4: 165340671-165340671 on Build GRCh38
rs761784749 Chr.4: 165340259-165340259 on Build GRCh38
rs753316142 Chr.4: 165340334-165340334 on Build GRCh38
rs771192308 Chr.4: 165340216-165340216 on Build GRCh38
rs931770139 Chr.4: 165340649-165340650 on Build GRCh38

View Full Product Details