Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34686759-34687041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAI14
Entrez Gene ID
26064
Gene Name
retinoic acid induced 14
Gene Aliases
NORPEG, RAI13
UniGene
Hs.431400
Gene Chromosome Location
Chr.5: 34656328-34832612 on Build GRCh38
Transcripts
NM_001145520.1 NM_001145521.1 NM_001145522.1 NM_001145525.1 NM_015577.2 XM_006714469.2 XM_011514017.2 XM_011514018.1 XM_011514019.1 XM_011514020.1 XM_011514021.1 XM_011514022.1 XM_011514023.1 XM_011514024.1 XM_017009335.1 XM_017009336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1190765 c.12delG p.A6fs*67 Chr.5: 34686931-34686931 on Build GRCh38
2686673 c.36+1G>A p.? Chr.5: 34686956-34686956 on Build GRCh38
1634199 c.20A>C p.K7T Chr.5: 34686939-34686939 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371302956 Chr.5: 34686979-34686979 on Build GRCh38
rs748882833 Chr.5: 34686989-34686991 on Build GRCh38
rs112973577 Chr.5: 34686909-34686909 on Build GRCh38
rs978156881 Chr.5: 34686813-34686813 on Build GRCh38
rs754708870 Chr.5: 34687014-34687014 on Build GRCh38
rs763146818 Chr.5: 34687007-34687007 on Build GRCh38
rs372768121 Chr.5: 34686781-34686781 on Build GRCh38
rs773321081 Chr.5: 34686889-34686889 on Build GRCh38
rs539973928 Chr.5: 34686966-34686966 on Build GRCh38
rs769385277 Chr.5: 34686872-34686872 on Build GRCh38
rs773623692 Chr.5: 34687001-34687001 on Build GRCh38
rs770059509 Chr.5: 34686999-34686999 on Build GRCh38
rs781681735 Chr.5: 34686989-34686989 on Build GRCh38
rs759599283 Chr.5: 34686927-34686927 on Build GRCh38
rs528927332 Chr.5: 34686988-34686988 on Build GRCh38
rs531599453 Chr.5: 34687021-34687021 on Build GRCh38
rs564313431 Chr.5: 34686849-34686849 on Build GRCh38
rs577794681 Chr.5: 34686956-34686956 on Build GRCh38
rs200951966 Chr.5: 34686974-34686974 on Build GRCh38
rs745348934 Chr.5: 34686981-34686981 on Build GRCh38
rs1025138354 Chr.5: 34686789-34686789 on Build GRCh38
rs770148151 Chr.5: 34686888-34686888 on Build GRCh38
rs755872354 Chr.5: 34686963-34686963 on Build GRCh38
rs777553193 Chr.5: 34686965-34686965 on Build GRCh38
rs146556857 Chr.5: 34686990-34686990 on Build GRCh38
rs545635040 Chr.5: 34686969-34686969 on Build GRCh38
rs58427583 Chr.5: 34686945-34686945 on Build GRCh38
rs904239909 Chr.5: 34686936-34686936 on Build GRCh38
rs751680194 Chr.5: 34686916-34686916 on Build GRCh38
rs570877286 Chr.5: 34686937-34686937 on Build GRCh38
rs74979843 Chr.5: 34686805-34686805 on Build GRCh38
rs112936408 Chr.5: 34686929-34686929 on Build GRCh38
rs377152269 Chr.5: 34686786-34686786 on Build GRCh38
rs562648092 Chr.5: 34687018-34687018 on Build GRCh38
rs777598670 Chr.5: 34686877-34686878 on Build GRCh38

View Full Product Details