Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10225769-10226112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM173B
Entrez Gene ID
134145
Gene Name
family with sequence similarity 173 member B
Gene Aliases
JS-2
UniGene
Hs.481569
Gene Chromosome Location
Chr.5: 10225507-10249909 on Build GRCh38
Transcripts
NM_001258388.1 NM_001258389.1 NM_199133.3 NR_047668.1 NR_047669.1 NR_047670.1

SNP Detail

SNP ID Chromosome Location
rs994455182 Chr.5: 10226000-10226000 on Build GRCh38
rs140609613 Chr.5: 10225810-10225810 on Build GRCh38
rs115759246 Chr.5: 10225970-10225970 on Build GRCh38
rs1026921268 Chr.5: 10226013-10226013 on Build GRCh38
rs1396469 Chr.5: 10225881-10225881 on Build GRCh38
rs976345506 Chr.5: 10225869-10225869 on Build GRCh38
rs530064382 Chr.5: 10225818-10225818 on Build GRCh38
rs990880738 Chr.5: 10225866-10225866 on Build GRCh38
rs563339681 Chr.5: 10225941-10225941 on Build GRCh38
rs1021415648 Chr.5: 10225811-10225811 on Build GRCh38
rs749002345 Chr.5: 10225909-10225909 on Build GRCh38
rs551078167 Chr.5: 10225876-10225876 on Build GRCh38
rs919957374 Chr.5: 10225823-10225823 on Build GRCh38
rs781132470 Chr.5: 10225819-10225819 on Build GRCh38
rs539617324 Chr.5: 10225864-10225864 on Build GRCh38
rs945623749 Chr.5: 10225988-10225988 on Build GRCh38
rs983134781 Chr.5: 10225883-10225883 on Build GRCh38
rs961919649 Chr.5: 10225998-10226000 on Build GRCh38
rs13175950 Chr.5: 10225828-10225828 on Build GRCh38
rs1049513811 Chr.5: 10225925-10225925 on Build GRCh38
rs1057061296 Chr.5: 10225951-10225951 on Build GRCh38
rs889922241 Chr.5: 10225939-10225939 on Build GRCh38
rs1355096 Chr.5: 10225913-10225913 on Build GRCh38
rs916664602 Chr.5: 10226084-10226084 on Build GRCh38
rs903877904 Chr.5: 10226048-10226048 on Build GRCh38
rs561158524 Chr.5: 10225807-10225807 on Build GRCh38
rs773855630 Chr.5: 10225943-10225943 on Build GRCh38
rs915714571 Chr.5: 10225871-10225871 on Build GRCh38
rs533073187 Chr.5: 10226031-10226031 on Build GRCh38
rs552274268 Chr.5: 10225933-10225933 on Build GRCh38
rs1000882877 Chr.5: 10226068-10226068 on Build GRCh38
rs145723357 Chr.5: 10226026-10226026 on Build GRCh38
rs964533960 Chr.5: 10225840-10225840 on Build GRCh38
rs941726442 Chr.5: 10225900-10225900 on Build GRCh38
rs1396470 Chr.5: 10225805-10225805 on Build GRCh38
rs908880465 Chr.5: 10225884-10225884 on Build GRCh38
rs990828317 Chr.5: 10226060-10226062 on Build GRCh38

View Full Product Details