Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80320969-80321463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPZ1
Entrez Gene ID
84654
Gene Name
spermatogenic leucine zipper 1
Gene Aliases
NYD-TSP1, PPP1R148
UniGene
Hs.519403
Gene Chromosome Location
Chr.5: 80319971-80321842 on Build GRCh38
Transcripts
NM_032567.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
483096 c.789G>A p.E263E Chr.5: 80321004-80321004 on Build GRCh38
1227520 c.1092T>G p.N364K Chr.5: 80321307-80321307 on Build GRCh38
4833890 c.999G>C p.Q333H Chr.5: 80321214-80321214 on Build GRCh38
5951095 c.930G>C p.L310L Chr.5: 80321145-80321145 on Build GRCh38
483097 c.1143A>G p.Q381Q Chr.5: 80321358-80321358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765112127 Chr.5: 80321252-80321252 on Build GRCh38
rs766255372 Chr.5: 80321108-80321108 on Build GRCh38
rs767153726 Chr.5: 80321404-80321404 on Build GRCh38
rs746278866 Chr.5: 80321059-80321059 on Build GRCh38
rs757326569 Chr.5: 80321252-80321253 on Build GRCh38
rs769002129 Chr.5: 80321314-80321314 on Build GRCh38
rs35337118 Chr.5: 80321117-80321117 on Build GRCh38
rs763015998 Chr.5: 80321100-80321100 on Build GRCh38
rs375261534 Chr.5: 80321215-80321215 on Build GRCh38
rs750966909 Chr.5: 80321292-80321292 on Build GRCh38
rs752399875 Chr.5: 80321150-80321150 on Build GRCh38
rs200874440 Chr.5: 80321235-80321235 on Build GRCh38
rs1004436420 Chr.5: 80321052-80321052 on Build GRCh38
rs373336998 Chr.5: 80321180-80321180 on Build GRCh38
rs151102391 Chr.5: 80321255-80321255 on Build GRCh38
rs199936871 Chr.5: 80321190-80321190 on Build GRCh38
rs780236836 Chr.5: 80321209-80321209 on Build GRCh38
rs756525360 Chr.5: 80321296-80321296 on Build GRCh38
rs747741292 Chr.5: 80321004-80321004 on Build GRCh38
rs116481637 Chr.5: 80321204-80321204 on Build GRCh38
rs749651812 Chr.5: 80321309-80321309 on Build GRCh38
rs751254507 Chr.5: 80321264-80321264 on Build GRCh38
rs748311486 Chr.5: 80321332-80321332 on Build GRCh38
rs370058409 Chr.5: 80321212-80321212 on Build GRCh38
rs777082834 Chr.5: 80321017-80321017 on Build GRCh38
rs867071040 Chr.5: 80321378-80321379 on Build GRCh38
rs756890017 Chr.5: 80321268-80321268 on Build GRCh38
rs764237055 Chr.5: 80321171-80321171 on Build GRCh38
rs114915340 Chr.5: 80321116-80321116 on Build GRCh38
rs570862191 Chr.5: 80321232-80321233 on Build GRCh38
rs749947375 Chr.5: 80321406-80321406 on Build GRCh38
rs772125664 Chr.5: 80321218-80321218 on Build GRCh38
rs751019971 Chr.5: 80321352-80321352 on Build GRCh38
rs1015828758 Chr.5: 80321067-80321067 on Build GRCh38
rs571319175 Chr.5: 80321379-80321380 on Build GRCh38
rs763972183 Chr.5: 80321384-80321384 on Build GRCh38
rs761945639 Chr.5: 80321217-80321217 on Build GRCh38
rs759432533 Chr.5: 80321123-80321123 on Build GRCh38
rs375200625 Chr.5: 80321008-80321008 on Build GRCh38
rs777449245 Chr.5: 80321179-80321179 on Build GRCh38
rs772144224 Chr.5: 80321342-80321342 on Build GRCh38
rs141018927 Chr.5: 80321396-80321396 on Build GRCh38
rs780586572 Chr.5: 80321300-80321300 on Build GRCh38
rs747550254 Chr.5: 80321024-80321024 on Build GRCh38
rs6867419 Chr.5: 80321119-80321119 on Build GRCh38
rs754354201 Chr.5: 80321437-80321437 on Build GRCh38
rs764884229 Chr.5: 80321124-80321124 on Build GRCh38
rs755616127 Chr.5: 80321421-80321421 on Build GRCh38
rs770195297 Chr.5: 80321207-80321207 on Build GRCh38
rs374023153 Chr.5: 80321220-80321220 on Build GRCh38
rs778692448 Chr.5: 80320998-80320998 on Build GRCh38
rs774164075 Chr.5: 80321394-80321394 on Build GRCh38
rs769669883 Chr.5: 80321369-80321369 on Build GRCh38
rs142110272 Chr.5: 80321254-80321254 on Build GRCh38
rs34379390 Chr.5: 80321231-80321232 on Build GRCh38
rs536377169 Chr.5: 80321076-80321076 on Build GRCh38
rs777259240 Chr.5: 80321051-80321051 on Build GRCh38
rs745759990 Chr.5: 80321415-80321416 on Build GRCh38
rs762726613 Chr.5: 80321372-80321372 on Build GRCh38
rs987775100 Chr.5: 80321278-80321278 on Build GRCh38
rs775470178 Chr.5: 80321370-80321370 on Build GRCh38
rs111595904 Chr.5: 80321232-80321232 on Build GRCh38
rs775842332 Chr.5: 80321077-80321077 on Build GRCh38
rs780794389 Chr.5: 80321288-80321288 on Build GRCh38
rs759542671 Chr.5: 80321368-80321368 on Build GRCh38
rs369209245 Chr.5: 80321109-80321109 on Build GRCh38
rs771516244 Chr.5: 80321047-80321047 on Build GRCh38
rs766860460 Chr.5: 80321428-80321428 on Build GRCh38
rs867745345 Chr.5: 80321177-80321177 on Build GRCh38

View Full Product Details