Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141421172-141421721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1063294 c.239C>T p.P80L Chr.5: 141421466-141421466 on Build GRCh38
3941153 c.493G>A p.V165M Chr.5: 141421720-141421720 on Build GRCh38
3827197 c.412G>A p.E138K Chr.5: 141421639-141421639 on Build GRCh38
218826 c.388A>T p.N130Y Chr.5: 141421615-141421615 on Build GRCh38
1310749 c.11G>A p.R4Q Chr.5: 141421238-141421238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781622911 Chr.5: 141421617-141421617 on Build GRCh38
rs760512084 Chr.5: 141421554-141421554 on Build GRCh38
rs375885183 Chr.5: 141421412-141421412 on Build GRCh38
rs549572548 Chr.5: 141421360-141421360 on Build GRCh38
rs920128735 Chr.5: 141421238-141421238 on Build GRCh38
rs749916401 Chr.5: 141421363-141421363 on Build GRCh38
rs370020854 Chr.5: 141421318-141421318 on Build GRCh38
rs370284141 Chr.5: 141421654-141421654 on Build GRCh38
rs746753262 Chr.5: 141421221-141421221 on Build GRCh38
rs375319424 Chr.5: 141421580-141421580 on Build GRCh38
rs202220769 Chr.5: 141421348-141421348 on Build GRCh38
rs777151399 Chr.5: 141421551-141421552 on Build GRCh38
rs761522510 Chr.5: 141421357-141421357 on Build GRCh38
rs375937711 Chr.5: 141421524-141421524 on Build GRCh38
rs11167744 Chr.5: 141421537-141421537 on Build GRCh38
rs201429116 Chr.5: 141421313-141421313 on Build GRCh38
rs769354611 Chr.5: 141421442-141421442 on Build GRCh38
rs778666303 Chr.5: 141421619-141421619 on Build GRCh38
rs975619932 Chr.5: 141421535-141421536 on Build GRCh38
rs761399164 Chr.5: 141421237-141421245 on Build GRCh38
rs776952658 Chr.5: 141421229-141421229 on Build GRCh38
rs775131295 Chr.5: 141421638-141421638 on Build GRCh38
rs751572007 Chr.5: 141421699-141421699 on Build GRCh38
rs754141447 Chr.5: 141421489-141421489 on Build GRCh38
rs751341677 Chr.5: 141421296-141421296 on Build GRCh38
rs908483513 Chr.5: 141421422-141421422 on Build GRCh38
rs969154252 Chr.5: 141421218-141421218 on Build GRCh38
rs982627903 Chr.5: 141421421-141421421 on Build GRCh38
rs772564300 Chr.5: 141421433-141421433 on Build GRCh38
rs751563833 Chr.5: 141421382-141421382 on Build GRCh38
rs781110850 Chr.5: 141421309-141421309 on Build GRCh38
rs778839137 Chr.5: 141421419-141421419 on Build GRCh38
rs201076931 Chr.5: 141421233-141421233 on Build GRCh38
rs557991200 Chr.5: 141421532-141421532 on Build GRCh38
rs1029858235 Chr.5: 141421569-141421569 on Build GRCh38
rs762064258 Chr.5: 141421282-141421282 on Build GRCh38
rs758920296 Chr.5: 141421507-141421507 on Build GRCh38
rs757410882 Chr.5: 141421504-141421504 on Build GRCh38
rs560707757 Chr.5: 141421338-141421338 on Build GRCh38
rs368199651 Chr.5: 141421513-141421513 on Build GRCh38
rs367946949 Chr.5: 141421640-141421640 on Build GRCh38
rs538537555 Chr.5: 141421410-141421410 on Build GRCh38
rs907282414 Chr.5: 141421473-141421473 on Build GRCh38
rs759899934 Chr.5: 141421255-141421255 on Build GRCh38
rs35600583 Chr.5: 141421288-141421289 on Build GRCh38
rs770419617 Chr.5: 141421523-141421523 on Build GRCh38
rs748399893 Chr.5: 141421618-141421618 on Build GRCh38
rs759548375 Chr.5: 141421468-141421468 on Build GRCh38
rs749034718 Chr.5: 141421520-141421520 on Build GRCh38
rs779780797 Chr.5: 141421203-141421203 on Build GRCh38
rs765969768 Chr.5: 141421456-141421456 on Build GRCh38
rs552534116 Chr.5: 141421614-141421614 on Build GRCh38
rs377161386 Chr.5: 141421300-141421300 on Build GRCh38
rs780085355 Chr.5: 141421593-141421593 on Build GRCh38
rs865974752 Chr.5: 141421622-141421622 on Build GRCh38
rs763146085 Chr.5: 141421687-141421687 on Build GRCh38
rs755936665 Chr.5: 141421477-141421477 on Build GRCh38
rs975166167 Chr.5: 141421364-141421364 on Build GRCh38
rs772957069 Chr.5: 141421448-141421448 on Build GRCh38
rs531591776 Chr.5: 141421582-141421582 on Build GRCh38
rs1039652739 Chr.5: 141421262-141421262 on Build GRCh38
rs530853994 Chr.5: 141421230-141421230 on Build GRCh38
rs748347420 Chr.5: 141421232-141421232 on Build GRCh38
rs377652360 Chr.5: 141421530-141421530 on Build GRCh38
rs752558543 Chr.5: 141421475-141421475 on Build GRCh38
rs771759412 Chr.5: 141421634-141421634 on Build GRCh38
rs750692137 Chr.5: 141421681-141421681 on Build GRCh38
rs773727821 Chr.5: 141421464-141421464 on Build GRCh38
rs777999539 Chr.5: 141421399-141421399 on Build GRCh38
rs775839500 Chr.5: 141421435-141421435 on Build GRCh38
rs934861223 Chr.5: 141421491-141421491 on Build GRCh38
rs374319762 Chr.5: 141421695-141421695 on Build GRCh38
rs763769838 Chr.5: 141421478-141421478 on Build GRCh38
rs199973694 Chr.5: 141421516-141421516 on Build GRCh38
rs143092131 Chr.5: 141421297-141421297 on Build GRCh38
rs369403750 Chr.5: 141421388-141421388 on Build GRCh38
rs767500900 Chr.5: 141421469-141421469 on Build GRCh38
rs745883683 Chr.5: 141421505-141421505 on Build GRCh38
rs1047768415 Chr.5: 141421246-141421246 on Build GRCh38
rs779984795 Chr.5: 141421294-141421294 on Build GRCh38
rs539072548 Chr.5: 141421546-141421546 on Build GRCh38
rs761468303 Chr.5: 141421659-141421659 on Build GRCh38
rs77665158 Chr.5: 141421595-141421595 on Build GRCh38
rs531911789 Chr.5: 141421284-141421284 on Build GRCh38
rs771422215 Chr.5: 141421444-141421445 on Build GRCh38
rs868219163 Chr.5: 141421618-141421619 on Build GRCh38
rs756414068 Chr.5: 141421396-141421396 on Build GRCh38
rs764010392 Chr.5: 141421557-141421557 on Build GRCh38
rs201673318 Chr.5: 141421626-141421626 on Build GRCh38
rs765475696 Chr.5: 141421660-141421660 on Build GRCh38
rs750973568 Chr.5: 141421697-141421697 on Build GRCh38
rs761059925 Chr.5: 141421322-141421323 on Build GRCh38
rs762490406 Chr.5: 141421455-141421455 on Build GRCh38
rs771398829 Chr.5: 141421251-141421251 on Build GRCh38
rs750525078 Chr.5: 141421586-141421586 on Build GRCh38
rs1053280090 Chr.5: 141421503-141421503 on Build GRCh38
rs780491148 Chr.5: 141421508-141421508 on Build GRCh38
rs960627405 Chr.5: 141421384-141421384 on Build GRCh38
rs761435958 Chr.5: 141421277-141421277 on Build GRCh38
rs774885270 Chr.5: 141421322-141421322 on Build GRCh38
rs773410079 Chr.5: 141421239-141421239 on Build GRCh38
rs373015809 Chr.5: 141421436-141421436 on Build GRCh38
rs369037131 Chr.5: 141421424-141421424 on Build GRCh38
rs199877605 Chr.5: 141421311-141421311 on Build GRCh38
rs903229017 Chr.5: 141421678-141421678 on Build GRCh38
rs758345796 Chr.5: 141421192-141421192 on Build GRCh38
rs80184002 Chr.5: 141421533-141421533 on Build GRCh38
rs368125665 Chr.5: 141421226-141421226 on Build GRCh38
rs1015444758 Chr.5: 141421698-141421698 on Build GRCh38
rs772487391 Chr.5: 141421262-141421263 on Build GRCh38
rs758433133 Chr.5: 141421587-141421587 on Build GRCh38
rs765321947 Chr.5: 141421583-141421583 on Build GRCh38
rs377073702 Chr.5: 141421457-141421457 on Build GRCh38
rs991814876 Chr.5: 141421199-141421199 on Build GRCh38
rs754951142 Chr.5: 141421383-141421383 on Build GRCh38
rs747573417 Chr.5: 141421611-141421611 on Build GRCh38
rs776499706 Chr.5: 141421657-141421657 on Build GRCh38
rs201273667 Chr.5: 141421243-141421243 on Build GRCh38
rs748368476 Chr.5: 141421263-141421265 on Build GRCh38
rs867187212 Chr.5: 141421629-141421630 on Build GRCh38
rs755518757 Chr.5: 141421700-141421700 on Build GRCh38
rs894586889 Chr.5: 141421219-141421219 on Build GRCh38
rs762484968 Chr.5: 141421366-141421366 on Build GRCh38
rs765775416 Chr.5: 141421370-141421370 on Build GRCh38
rs751227393 Chr.5: 141421279-141421279 on Build GRCh38
rs764787116 Chr.5: 141421278-141421278 on Build GRCh38
rs199737254 Chr.5: 141421342-141421342 on Build GRCh38
rs753573473 Chr.5: 141421579-141421579 on Build GRCh38
rs866635411 Chr.5: 141421211-141421211 on Build GRCh38
rs375071225 Chr.5: 141421312-141421312 on Build GRCh38
rs771088122 Chr.5: 141421418-141421418 on Build GRCh38
rs147068995 Chr.5: 141421347-141421347 on Build GRCh38
rs944057868 Chr.5: 141421257-141421257 on Build GRCh38
rs916345810 Chr.5: 141421213-141421213 on Build GRCh38
rs564480755 Chr.5: 141421280-141421280 on Build GRCh38
rs200646513 Chr.5: 141421389-141421389 on Build GRCh38
rs753531462 Chr.5: 141421261-141421261 on Build GRCh38

View Full Product Details