Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181198565-181199163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM7
Entrez Gene ID
81786
Gene Name
tripartite motif containing 7
Gene Aliases
GNIP, RNF90
UniGene
Hs.487412
Gene Chromosome Location
Chr.5: 181193924-181206000 on Build GRCh38
Transcripts
NM_203293.2 NM_203294.1 NM_203295.1 NM_203296.1 NM_203297.1 XM_017009903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795917 c.898C>T p.P300S Chr.5: 181198780-181198780 on Build GRCh38
6378131 c.979A>G p.K327E Chr.5: 181198699-181198699 on Build GRCh38
1595307 c.424A>G p.M142V Chr.5: 181198708-181198708 on Build GRCh38
4843482 c.855C>A p.F285L Chr.5: 181199112-181199112 on Build GRCh38
6808565 c.347C>A p.P116H Chr.5: 181198785-181198785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2770940 Chr.5: 181198672-181198672 on Build GRCh38
rs377597666 Chr.5: 181198775-181198775 on Build GRCh38
rs981189008 Chr.5: 181198980-181198980 on Build GRCh38
rs774225201 Chr.5: 181198702-181198702 on Build GRCh38
rs753105140 Chr.5: 181198763-181198763 on Build GRCh38
rs763797079 Chr.5: 181198739-181198739 on Build GRCh38
rs963093094 Chr.5: 181198944-181198944 on Build GRCh38
rs755343190 Chr.5: 181198673-181198673 on Build GRCh38
rs757481509 Chr.5: 181198839-181198839 on Build GRCh38
rs945843473 Chr.5: 181198975-181198975 on Build GRCh38
rs895368502 Chr.5: 181198784-181198784 on Build GRCh38
rs748263061 Chr.5: 181198676-181198676 on Build GRCh38
rs201350955 Chr.5: 181198708-181198708 on Build GRCh38
rs773086182 Chr.5: 181199078-181199078 on Build GRCh38
rs1016397773 Chr.5: 181198919-181198919 on Build GRCh38
rs200743322 Chr.5: 181198772-181198772 on Build GRCh38
rs145627318 Chr.5: 181198767-181198767 on Build GRCh38
rs759304775 Chr.5: 181198653-181198653 on Build GRCh38
rs143052397 Chr.5: 181199113-181199113 on Build GRCh38
rs745913764 Chr.5: 181199063-181199066 on Build GRCh38
rs534919930 Chr.5: 181198622-181198622 on Build GRCh38
rs937116271 Chr.5: 181198699-181198699 on Build GRCh38
rs79848802 Chr.5: 181199030-181199030 on Build GRCh38
rs764771332 Chr.5: 181198804-181198804 on Build GRCh38
rs752050805 Chr.5: 181198773-181198773 on Build GRCh38
rs768037758 Chr.5: 181198809-181198809 on Build GRCh38
rs762529815 Chr.5: 181199088-181199088 on Build GRCh38
rs1045624557 Chr.5: 181198675-181198675 on Build GRCh38
rs745940866 Chr.5: 181198851-181198851 on Build GRCh38
rs987370187 Chr.5: 181198966-181198966 on Build GRCh38
rs758444100 Chr.5: 181199048-181199048 on Build GRCh38
rs755558773 Chr.5: 181198759-181198759 on Build GRCh38
rs781611089 Chr.5: 181198762-181198762 on Build GRCh38
rs374218449 Chr.5: 181199124-181199124 on Build GRCh38
rs143743449 Chr.5: 181198989-181198989 on Build GRCh38
rs563339666 Chr.5: 181199035-181199035 on Build GRCh38
rs950929937 Chr.5: 181198906-181198906 on Build GRCh38
rs760482731 Chr.5: 181198792-181198792 on Build GRCh38
rs776297811 Chr.5: 181198799-181198799 on Build GRCh38
rs749941645 Chr.5: 181198805-181198805 on Build GRCh38
rs763453852 Chr.5: 181198659-181198659 on Build GRCh38
rs941036977 Chr.5: 181198591-181198591 on Build GRCh38
rs542618244 Chr.5: 181198910-181198910 on Build GRCh38
rs557058588 Chr.5: 181198787-181198787 on Build GRCh38
rs765832844 Chr.5: 181199097-181199097 on Build GRCh38
rs933255968 Chr.5: 181198957-181198957 on Build GRCh38
rs72821730 Chr.5: 181199031-181199031 on Build GRCh38
rs116911833 Chr.5: 181199104-181199104 on Build GRCh38
rs775352339 Chr.5: 181198790-181198790 on Build GRCh38
rs760444617 Chr.5: 181198738-181198738 on Build GRCh38
rs543152078 Chr.5: 181198879-181198879 on Build GRCh38
rs757682266 Chr.5: 181199107-181199107 on Build GRCh38
rs369139201 Chr.5: 181198843-181198843 on Build GRCh38
rs752070032 Chr.5: 181198754-181198754 on Build GRCh38
rs761380942 Chr.5: 181198747-181198747 on Build GRCh38
rs573420542 Chr.5: 181198972-181198972 on Build GRCh38
rs974365505 Chr.5: 181198950-181198950 on Build GRCh38
rs983248835 Chr.5: 181198907-181198907 on Build GRCh38
rs780671746 Chr.5: 181198668-181198668 on Build GRCh38
rs764400523 Chr.5: 181198831-181198831 on Build GRCh38
rs764465044 Chr.5: 181199103-181199103 on Build GRCh38
rs756894305 Chr.5: 181198667-181198667 on Build GRCh38
rs185494747 Chr.5: 181198956-181198956 on Build GRCh38
rs777928839 Chr.5: 181198682-181198682 on Build GRCh38
rs2770941 Chr.5: 181199036-181199036 on Build GRCh38
rs780028157 Chr.5: 181199140-181199140 on Build GRCh38
rs778971294 Chr.5: 181198777-181198777 on Build GRCh38
rs753192840 Chr.5: 181198829-181198829 on Build GRCh38
rs974173596 Chr.5: 181198965-181198965 on Build GRCh38
rs369187975 Chr.5: 181198685-181198685 on Build GRCh38
rs267600589 Chr.5: 181199117-181199117 on Build GRCh38
rs551698732 Chr.5: 181199110-181199110 on Build GRCh38
rs771048476 Chr.5: 181198697-181198697 on Build GRCh38
rs372813001 Chr.5: 181198936-181198936 on Build GRCh38
rs780019430 Chr.5: 181199066-181199066 on Build GRCh38
rs753468703 Chr.5: 181198740-181198740 on Build GRCh38
rs764859133 Chr.5: 181198751-181198751 on Build GRCh38
rs372866184 Chr.5: 181198654-181198654 on Build GRCh38
rs756457128 Chr.5: 181198830-181198830 on Build GRCh38
rs781711703 Chr.5: 181198674-181198674 on Build GRCh38
rs983603348 Chr.5: 181198634-181198634 on Build GRCh38
rs768383845 Chr.5: 181198796-181198796 on Build GRCh38
rs776292938 Chr.5: 181199077-181199077 on Build GRCh38
rs769567949 Chr.5: 181199083-181199083 on Build GRCh38
rs753070208 Chr.5: 181198584-181198584 on Build GRCh38
rs192612658 Chr.5: 181198628-181198628 on Build GRCh38
rs559309397 Chr.5: 181199102-181199102 on Build GRCh38
rs986233594 Chr.5: 181198878-181198878 on Build GRCh38
rs112292069 Chr.5: 181198882-181198882 on Build GRCh38
rs903898238 Chr.5: 181198993-181198993 on Build GRCh38
rs565336920 Chr.5: 181199010-181199010 on Build GRCh38
rs746942365 Chr.5: 181199076-181199076 on Build GRCh38
rs781264105 Chr.5: 181198900-181198900 on Build GRCh38
rs753995860 Chr.5: 181198835-181198835 on Build GRCh38
rs548655912 Chr.5: 181199098-181199098 on Build GRCh38
rs761587423 Chr.5: 181198801-181198801 on Build GRCh38
rs773039975 Chr.5: 181199086-181199086 on Build GRCh38
rs202093181 Chr.5: 181198721-181198721 on Build GRCh38
rs774312709 Chr.5: 181198639-181198639 on Build GRCh38

View Full Product Details