Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27457129-27457717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF184
Entrez Gene ID
7738
Gene Name
zinc finger protein 184
Gene Aliases
kr-ZNF3
UniGene
Hs.158174
Gene Chromosome Location
Chr.6: 27450742-27473118 on Build GRCh38
Transcripts
NM_001318891.1 NM_001318892.1 NM_001318893.1 NM_007149.2 XM_011514861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031252 c.81G>C p.A27A Chr.6: 27457404-27457404 on Build GRCh38
376092 c.192G>T p.L64L Chr.6: 27457293-27457293 on Build GRCh38
4390294 c.186A>T p.T62T Chr.6: 27457299-27457299 on Build GRCh38
5919196 c.179A>T p.N60I Chr.6: 27457306-27457306 on Build GRCh38
1076945 c.164A>T p.D55V Chr.6: 27457321-27457321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766071870 Chr.6: 27457304-27457304 on Build GRCh38
rs771493685 Chr.6: 27457500-27457500 on Build GRCh38
rs532072395 Chr.6: 27457684-27457684 on Build GRCh38
rs751694853 Chr.6: 27457334-27457334 on Build GRCh38
rs778560902 Chr.6: 27457344-27457345 on Build GRCh38
rs556993744 Chr.6: 27457564-27457564 on Build GRCh38
rs561049848 Chr.6: 27457191-27457191 on Build GRCh38
rs776049408 Chr.6: 27457683-27457683 on Build GRCh38
rs775161269 Chr.6: 27457460-27457460 on Build GRCh38
rs267600919 Chr.6: 27457326-27457326 on Build GRCh38
rs929160631 Chr.6: 27457545-27457545 on Build GRCh38
rs369819747 Chr.6: 27457392-27457392 on Build GRCh38
rs114337108 Chr.6: 27457528-27457528 on Build GRCh38
rs894037791 Chr.6: 27457636-27457636 on Build GRCh38
rs781691214 Chr.6: 27457390-27457390 on Build GRCh38
rs373400111 Chr.6: 27457263-27457263 on Build GRCh38
rs954400683 Chr.6: 27457677-27457677 on Build GRCh38
rs61736959 Chr.6: 27457404-27457404 on Build GRCh38
rs538572160 Chr.6: 27457616-27457616 on Build GRCh38
rs570494762 Chr.6: 27457580-27457580 on Build GRCh38
rs760252022 Chr.6: 27457241-27457241 on Build GRCh38
rs920495900 Chr.6: 27457537-27457537 on Build GRCh38
rs981002829 Chr.6: 27457163-27457163 on Build GRCh38
rs183155479 Chr.6: 27457694-27457694 on Build GRCh38
rs1883216 Chr.6: 27457406-27457406 on Build GRCh38
rs538792966 Chr.6: 27457660-27457662 on Build GRCh38
rs768334130 Chr.6: 27457505-27457505 on Build GRCh38
rs754429217 Chr.6: 27457266-27457267 on Build GRCh38
rs776680383 Chr.6: 27457249-27457249 on Build GRCh38
rs917275639 Chr.6: 27457507-27457507 on Build GRCh38
rs777662822 Chr.6: 27457462-27457462 on Build GRCh38
rs545869331 Chr.6: 27457648-27457648 on Build GRCh38
rs905543729 Chr.6: 27457601-27457601 on Build GRCh38
rs754556546 Chr.6: 27457498-27457498 on Build GRCh38
rs114235597 Chr.6: 27457487-27457487 on Build GRCh38
rs181590740 Chr.6: 27457225-27457225 on Build GRCh38
rs749414025 Chr.6: 27457470-27457470 on Build GRCh38
rs201051660 Chr.6: 27457424-27457424 on Build GRCh38
rs757322216 Chr.6: 27457369-27457369 on Build GRCh38
rs1000745604 Chr.6: 27457587-27457587 on Build GRCh38
rs41269265 Chr.6: 27457570-27457570 on Build GRCh38
rs575157542 Chr.6: 27457639-27457639 on Build GRCh38
rs199906744 Chr.6: 27457229-27457229 on Build GRCh38
rs758552695 Chr.6: 27457313-27457313 on Build GRCh38
rs919321905 Chr.6: 27457223-27457223 on Build GRCh38
rs765155050 Chr.6: 27457457-27457457 on Build GRCh38
rs1044085138 Chr.6: 27457582-27457582 on Build GRCh38
rs956658382 Chr.6: 27457371-27457371 on Build GRCh38
rs770544890 Chr.6: 27457433-27457433 on Build GRCh38
rs538050800 Chr.6: 27457664-27457664 on Build GRCh38
rs185358053 Chr.6: 27457239-27457239 on Build GRCh38
rs752497966 Chr.6: 27457302-27457302 on Build GRCh38
rs1047483496 Chr.6: 27457353-27457353 on Build GRCh38
rs1013899935 Chr.6: 27457399-27457399 on Build GRCh38
rs193921063 Chr.6: 27457372-27457372 on Build GRCh38
rs191346721 Chr.6: 27457624-27457624 on Build GRCh38
rs10484398 Chr.6: 27457181-27457181 on Build GRCh38
rs368975277 Chr.6: 27457245-27457245 on Build GRCh38
rs776232234 Chr.6: 27457441-27457441 on Build GRCh38
rs772916087 Chr.6: 27457407-27457407 on Build GRCh38
rs368356547 Chr.6: 27457657-27457657 on Build GRCh38
rs386698292 Chr.6: 27457404-27457406 on Build GRCh38
rs988241755 Chr.6: 27457231-27457231 on Build GRCh38
rs199663360 Chr.6: 27457405-27457405 on Build GRCh38
rs771693183 Chr.6: 27457235-27457235 on Build GRCh38
rs775269468 Chr.6: 27457448-27457449 on Build GRCh38
rs150047687 Chr.6: 27457603-27457604 on Build GRCh38
rs200203668 Chr.6: 27457278-27457278 on Build GRCh38
rs144235614 Chr.6: 27457446-27457446 on Build GRCh38

View Full Product Details