Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111702740-111703260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYN
Entrez Gene ID
2534
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Gene Aliases
SLK, SYN, p59-FYN
UniGene
Hs.390567
Gene Chromosome Location
Chr.6: 111660332-111873452 on Build GRCh38
Transcripts
NM_002037.5 NM_153047.3 NM_153048.3 XM_005266890.3 XM_005266892.3 XM_017010650.1 XM_017010651.1 XM_017010652.1 XM_017010653.1 XM_017010654.1 XM_017010655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104867 c.684A>T p.V228V Chr.6: 111702898-111702898 on Build GRCh38
6983610 c.568C>A p.R190S Chr.6: 111703014-111703014 on Build GRCh38
6940350 c.664G>A p.E222K Chr.6: 111702918-111702918 on Build GRCh38
6104868 c.684A>T p.V228V Chr.6: 111702898-111702898 on Build GRCh38
6938752 c.598G>A p.V200I Chr.6: 111702984-111702984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766191397 Chr.6: 111702985-111702985 on Build GRCh38
rs750940060 Chr.6: 111703043-111703043 on Build GRCh38
rs762837809 Chr.6: 111702982-111702982 on Build GRCh38
rs3752545 Chr.6: 111703059-111703059 on Build GRCh38
rs756342493 Chr.6: 111703013-111703013 on Build GRCh38
rs776054256 Chr.6: 111702935-111702935 on Build GRCh38
rs772664549 Chr.6: 111703060-111703060 on Build GRCh38
rs138389248 Chr.6: 111703133-111703133 on Build GRCh38
rs754083510 Chr.6: 111703072-111703072 on Build GRCh38
rs869289014 Chr.6: 111703088-111703089 on Build GRCh38
rs1039818586 Chr.6: 111703138-111703138 on Build GRCh38
rs779589165 Chr.6: 111702849-111702849 on Build GRCh38
rs770783135 Chr.6: 111703062-111703062 on Build GRCh38
rs774125832 Chr.6: 111703065-111703065 on Build GRCh38
rs943244802 Chr.6: 111702879-111702879 on Build GRCh38
rs943765930 Chr.6: 111703123-111703123 on Build GRCh38
rs765109051 Chr.6: 111702946-111702946 on Build GRCh38
rs367543110 Chr.6: 111703009-111703009 on Build GRCh38
rs764413618 Chr.6: 111703015-111703015 on Build GRCh38
rs199617444 Chr.6: 111703055-111703055 on Build GRCh38
rs967169481 Chr.6: 111702815-111702815 on Build GRCh38
rs753048061 Chr.6: 111702876-111702876 on Build GRCh38
rs928966271 Chr.6: 111703049-111703049 on Build GRCh38
rs903082939 Chr.6: 111703232-111703232 on Build GRCh38
rs752902959 Chr.6: 111703111-111703111 on Build GRCh38
rs1021446340 Chr.6: 111702794-111702794 on Build GRCh38
rs200446110 Chr.6: 111702866-111702866 on Build GRCh38
rs753926758 Chr.6: 111703017-111703017 on Build GRCh38
rs749390587 Chr.6: 111702843-111702843 on Build GRCh38
rs767472360 Chr.6: 111703068-111703068 on Build GRCh38
rs114632025 Chr.6: 111703237-111703237 on Build GRCh38
rs370069902 Chr.6: 111702889-111702889 on Build GRCh38
rs761344763 Chr.6: 111702941-111702941 on Build GRCh38
rs529688712 Chr.6: 111703044-111703044 on Build GRCh38
rs150477486 Chr.6: 111703087-111703087 on Build GRCh38
rs746413328 Chr.6: 111702862-111702862 on Build GRCh38
rs759432050 Chr.6: 111703067-111703067 on Build GRCh38
rs748045277 Chr.6: 111702842-111702848 on Build GRCh38
rs773177629 Chr.6: 111702954-111702954 on Build GRCh38
rs1021477485 Chr.6: 111703203-111703203 on Build GRCh38
rs750297020 Chr.6: 111703050-111703050 on Build GRCh38
rs368062473 Chr.6: 111702870-111702870 on Build GRCh38
rs777982703 Chr.6: 111702839-111702839 on Build GRCh38
rs541596113 Chr.6: 111702776-111702776 on Build GRCh38
rs368225494 Chr.6: 111703165-111703165 on Build GRCh38
rs78214354 Chr.6: 111702943-111702943 on Build GRCh38
rs146187358 Chr.6: 111703030-111703030 on Build GRCh38
rs1054923321 Chr.6: 111703139-111703139 on Build GRCh38
rs1011447623 Chr.6: 111702773-111702773 on Build GRCh38
rs747341065 Chr.6: 111703051-111703051 on Build GRCh38
rs77439992 Chr.6: 111702895-111702895 on Build GRCh38
rs570971797 Chr.6: 111703236-111703236 on Build GRCh38
rs757768764 Chr.6: 111702844-111702844 on Build GRCh38
rs562373735 Chr.6: 111702953-111702953 on Build GRCh38

View Full Product Details