Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 133891733-133892317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF21
Entrez Gene ID
6943
Gene Name
transcription factor 21
Gene Aliases
POD1, bHLHa23
UniGene
Hs.78061
Gene Chromosome Location
Chr.6: 133889121-133895537 on Build GRCh38
Transcripts
NM_003206.3 NM_198392.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153751 c.481G>C p.E161Q Chr.6: 133891743-133891743 on Build GRCh38
450527 c.487G>A p.D163N Chr.6: 133891749-133891749 on Build GRCh38
5035815 c.509C>T p.A170V Chr.6: 133891771-133891771 on Build GRCh38
6567557 c.539G>C p.*180S Chr.6: 133891801-133891801 on Build GRCh38
3174194 c.506C>T p.T169I Chr.6: 133891768-133891768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760685056 Chr.6: 133891761-133891761 on Build GRCh38
rs960531868 Chr.6: 133892081-133892081 on Build GRCh38
rs74322630 Chr.6: 133892102-133892102 on Build GRCh38
rs968183522 Chr.6: 133892196-133892196 on Build GRCh38
rs776004223 Chr.6: 133892252-133892252 on Build GRCh38
rs536221601 Chr.6: 133891933-133891933 on Build GRCh38
rs559140569 Chr.6: 133892144-133892144 on Build GRCh38
rs539101653 Chr.6: 133892065-133892065 on Build GRCh38
rs1050281646 Chr.6: 133892240-133892240 on Build GRCh38
rs1005649382 Chr.6: 133891970-133891970 on Build GRCh38
rs181269946 Chr.6: 133891852-133891852 on Build GRCh38
rs553347368 Chr.6: 133891969-133891969 on Build GRCh38
rs886831767 Chr.6: 133891946-133891946 on Build GRCh38
rs1040641470 Chr.6: 133891909-133891909 on Build GRCh38
rs775406576 Chr.6: 133891842-133891842 on Build GRCh38
rs769420004 Chr.6: 133891837-133891838 on Build GRCh38
rs373864394 Chr.6: 133892261-133892261 on Build GRCh38
rs749885222 Chr.6: 133891853-133891853 on Build GRCh38
rs371517170 Chr.6: 133891793-133891793 on Build GRCh38
rs369916071 Chr.6: 133891841-133891841 on Build GRCh38
rs987959865 Chr.6: 133892121-133892121 on Build GRCh38
rs763917542 Chr.6: 133891769-133891769 on Build GRCh38
rs768700019 Chr.6: 133891759-133891759 on Build GRCh38
rs910536362 Chr.6: 133891878-133891878 on Build GRCh38
rs761671595 Chr.6: 133891847-133891847 on Build GRCh38
rs747594868 Chr.6: 133892246-133892246 on Build GRCh38
rs923092385 Chr.6: 133892079-133892079 on Build GRCh38
rs956655465 Chr.6: 133892001-133892001 on Build GRCh38
rs993639693 Chr.6: 133891921-133891921 on Build GRCh38
rs142892731 Chr.6: 133891792-133891792 on Build GRCh38
rs373325410 Chr.6: 133891826-133891826 on Build GRCh38
rs369704773 Chr.6: 133891839-133891839 on Build GRCh38
rs1038097807 Chr.6: 133892282-133892282 on Build GRCh38
rs929476022 Chr.6: 133892275-133892275 on Build GRCh38
rs201763632 Chr.6: 133891848-133891848 on Build GRCh38
rs933250430 Chr.6: 133892091-133892091 on Build GRCh38
rs1002113672 Chr.6: 133892029-133892029 on Build GRCh38
rs567621197 Chr.6: 133891919-133891919 on Build GRCh38
rs756682579 Chr.6: 133891835-133891835 on Build GRCh38
rs760358719 Chr.6: 133891843-133891843 on Build GRCh38
rs768093248 Chr.6: 133891889-133891889 on Build GRCh38
rs77892079 Chr.6: 133892101-133892101 on Build GRCh38
rs371784661 Chr.6: 133891899-133891899 on Build GRCh38
rs943630437 Chr.6: 133891908-133891908 on Build GRCh38
rs943345921 Chr.6: 133892246-133892246 on Build GRCh38
rs531047577 Chr.6: 133891858-133891858 on Build GRCh38
rs778134064 Chr.6: 133891838-133891838 on Build GRCh38
rs745671255 Chr.6: 133891830-133891848 on Build GRCh38
rs748710162 Chr.6: 133892199-133892199 on Build GRCh38
rs910590219 Chr.6: 133892213-133892213 on Build GRCh38
rs527582894 Chr.6: 133892200-133892200 on Build GRCh38
rs200602364 Chr.6: 133891807-133891807 on Build GRCh38
rs889090922 Chr.6: 133891778-133891778 on Build GRCh38
rs925023973 Chr.6: 133891977-133891977 on Build GRCh38
rs776420567 Chr.6: 133891846-133891846 on Build GRCh38
rs1035858509 Chr.6: 133891872-133891872 on Build GRCh38
rs764158821 Chr.6: 133891844-133891844 on Build GRCh38
rs376667026 Chr.6: 133891832-133891832 on Build GRCh38
rs913991556 Chr.6: 133892143-133892143 on Build GRCh38
rs756756327 Chr.6: 133891771-133891771 on Build GRCh38
rs969060355 Chr.6: 133891930-133891930 on Build GRCh38
rs577902818 Chr.6: 133892153-133892153 on Build GRCh38
rs1006736170 Chr.6: 133891820-133891820 on Build GRCh38
rs781635603 Chr.6: 133891808-133891808 on Build GRCh38
rs201898286 Chr.6: 133891840-133891840 on Build GRCh38
rs969015751 Chr.6: 133892025-133892025 on Build GRCh38
rs781710416 Chr.6: 133891975-133891975 on Build GRCh38
rs57108836 Chr.6: 133892003-133892003 on Build GRCh38
rs550818611 Chr.6: 133891905-133891905 on Build GRCh38
rs993716568 Chr.6: 133891862-133891862 on Build GRCh38

View Full Product Details