Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29923723-29924258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRN1
Entrez Gene ID
9805
Gene Name
secernin 1
Gene Aliases
SES1
UniGene
Hs.520740
Gene Chromosome Location
Chr.7: 29920103-29990289 on Build GRCh38
Transcripts
NM_001145513.1 NM_001145514.1 NM_001145515.1 NM_014766.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1178800 c.1223C>T p.T408M Chr.7: 29923979-29923979 on Build GRCh38
485254 c.1219G>T p.D407Y Chr.7: 29923983-29923983 on Build GRCh38
3637857 c.1210G>A p.D404N Chr.7: 29923992-29923992 on Build GRCh38
6313541 c.1146C>A p.A382A Chr.7: 29924056-29924056 on Build GRCh38
3637859 c.1188G>A p.A396A Chr.7: 29924014-29924014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368251776 Chr.7: 29924197-29924197 on Build GRCh38
rs113598220 Chr.7: 29923791-29923791 on Build GRCh38
rs766914524 Chr.7: 29924066-29924066 on Build GRCh38
rs562333816 Chr.7: 29923763-29923763 on Build GRCh38
rs530184865 Chr.7: 29924073-29924073 on Build GRCh38
rs563084923 Chr.7: 29923978-29923978 on Build GRCh38
rs937984380 Chr.7: 29923894-29923894 on Build GRCh38
rs777886116 Chr.7: 29924105-29924105 on Build GRCh38
rs140280542 Chr.7: 29924014-29924014 on Build GRCh38
rs925308286 Chr.7: 29923888-29923888 on Build GRCh38
rs146686520 Chr.7: 29923984-29923984 on Build GRCh38
rs535954863 Chr.7: 29923790-29923790 on Build GRCh38
rs148215806 Chr.7: 29924127-29924127 on Build GRCh38
rs748113988 Chr.7: 29924005-29924005 on Build GRCh38
rs144432102 Chr.7: 29924071-29924071 on Build GRCh38
rs542364744 Chr.7: 29924080-29924080 on Build GRCh38
rs1044163683 Chr.7: 29923808-29923808 on Build GRCh38
rs1003969452 Chr.7: 29923964-29923964 on Build GRCh38
rs757459053 Chr.7: 29924111-29924111 on Build GRCh38
rs886630585 Chr.7: 29924237-29924237 on Build GRCh38
rs559196897 Chr.7: 29923937-29923937 on Build GRCh38
rs554114449 Chr.7: 29923820-29923820 on Build GRCh38
rs752853654 Chr.7: 29924095-29924095 on Build GRCh38
rs766898589 Chr.7: 29923963-29923966 on Build GRCh38
rs745597388 Chr.7: 29924125-29924125 on Build GRCh38
rs955288043 Chr.7: 29924110-29924110 on Build GRCh38
rs750303399 Chr.7: 29923755-29923755 on Build GRCh38
rs191349402 Chr.7: 29923949-29923949 on Build GRCh38
rs1015230096 Chr.7: 29923911-29923911 on Build GRCh38
rs777136534 Chr.7: 29924146-29924146 on Build GRCh38
rs961041305 Chr.7: 29923931-29923931 on Build GRCh38
rs758651415 Chr.7: 29924006-29924006 on Build GRCh38
rs1055128922 Chr.7: 29924174-29924174 on Build GRCh38
rs751817648 Chr.7: 29924107-29924107 on Build GRCh38
rs928801792 Chr.7: 29924206-29924206 on Build GRCh38
rs773996408 Chr.7: 29924135-29924135 on Build GRCh38
rs777818012 Chr.7: 29924010-29924010 on Build GRCh38
rs775070912 Chr.7: 29924027-29924027 on Build GRCh38
rs532312947 Chr.7: 29924171-29924171 on Build GRCh38
rs761613797 Chr.7: 29924141-29924141 on Build GRCh38
rs763587731 Chr.7: 29923950-29923950 on Build GRCh38
rs565977875 Chr.7: 29924172-29924172 on Build GRCh38
rs770760273 Chr.7: 29924015-29924015 on Build GRCh38
rs768095211 Chr.7: 29924131-29924131 on Build GRCh38
rs769561426 Chr.7: 29924025-29924025 on Build GRCh38
rs770300633 Chr.7: 29924170-29924173 on Build GRCh38
rs759779683 Chr.7: 29924149-29924149 on Build GRCh38
rs764218780 Chr.7: 29924166-29924166 on Build GRCh38
rs193920979 Chr.7: 29923979-29923979 on Build GRCh38
rs765653098 Chr.7: 29923890-29923890 on Build GRCh38
rs749231099 Chr.7: 29924130-29924130 on Build GRCh38
rs767568829 Chr.7: 29924229-29924229 on Build GRCh38
rs955151583 Chr.7: 29923835-29923835 on Build GRCh38
rs918207126 Chr.7: 29924194-29924194 on Build GRCh38
rs571470369 Chr.7: 29924168-29924168 on Build GRCh38
rs1027058084 Chr.7: 29924190-29924190 on Build GRCh38
rs779103581 Chr.7: 29923986-29923986 on Build GRCh38
rs886572748 Chr.7: 29923953-29923953 on Build GRCh38
rs764870475 Chr.7: 29923932-29923932 on Build GRCh38
rs377289232 Chr.7: 29924039-29924039 on Build GRCh38
rs376086790 Chr.7: 29924153-29924153 on Build GRCh38
rs996454440 Chr.7: 29924038-29924038 on Build GRCh38
rs970364485 Chr.7: 29924195-29924195 on Build GRCh38
rs779811504 Chr.7: 29924129-29924129 on Build GRCh38
rs776500891 Chr.7: 29923913-29923913 on Build GRCh38
rs1049570125 Chr.7: 29924158-29924158 on Build GRCh38
rs1028107483 Chr.7: 29923815-29923815 on Build GRCh38
rs763484028 Chr.7: 29924160-29924160 on Build GRCh38
rs763495126 Chr.7: 29923974-29923974 on Build GRCh38
rs754561585 Chr.7: 29923981-29923981 on Build GRCh38
rs945471277 Chr.7: 29923762-29923762 on Build GRCh38
rs200172241 Chr.7: 29924152-29924152 on Build GRCh38
rs1048408291 Chr.7: 29924236-29924236 on Build GRCh38
rs200712843 Chr.7: 29923919-29923919 on Build GRCh38
rs763832784 Chr.7: 29924032-29924032 on Build GRCh38
rs774198843 Chr.7: 29924033-29924033 on Build GRCh38
rs914012446 Chr.7: 29923746-29923746 on Build GRCh38
rs762334666 Chr.7: 29924031-29924031 on Build GRCh38
rs745865313 Chr.7: 29924019-29924019 on Build GRCh38
rs186571213 Chr.7: 29923862-29923862 on Build GRCh38
rs143093540 Chr.7: 29923962-29923962 on Build GRCh38
rs986493577 Chr.7: 29923890-29923893 on Build GRCh38
rs550323703 Chr.7: 29924183-29924183 on Build GRCh38
rs78129573 Chr.7: 29924118-29924118 on Build GRCh38
rs750853246 Chr.7: 29923956-29923956 on Build GRCh38
rs758359979 Chr.7: 29924100-29924100 on Build GRCh38
rs776407954 Chr.7: 29924017-29924017 on Build GRCh38

View Full Product Details