Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155811696-155812145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHH
Entrez Gene ID
6469
Gene Name
sonic hedgehog
Gene Aliases
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
UniGene
Hs.164537
Gene Chromosome Location
Chr.7: 155799984-155812273 on Build GRCh38
Transcripts
NM_000193.3 XM_011516479.2 XM_011516480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3636700 c.159G>A p.E53E Chr.7: 155811964-155811964 on Build GRCh38
3636699 c.223G>A p.E75K Chr.7: 155811900-155811900 on Build GRCh38
745729 c.85G>A p.G29R Chr.7: 155812038-155812038 on Build GRCh38
7321152 c.151G>A p.V51M Chr.7: 155811972-155811972 on Build GRCh38
4748030 c.4_20del17 p.L2fs*56 Chr.7: 155812103-155812119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777969206 Chr.7: 155812069-155812069 on Build GRCh38
rs754993269 Chr.7: 155811887-155811887 on Build GRCh38
rs938066966 Chr.7: 155811732-155811732 on Build GRCh38
rs374679410 Chr.7: 155811952-155811952 on Build GRCh38
rs754272598 Chr.7: 155812033-155812033 on Build GRCh38
rs766818803 Chr.7: 155812029-155812029 on Build GRCh38
rs104894044 Chr.7: 155811825-155811825 on Build GRCh38
rs746900990 Chr.7: 155811803-155811803 on Build GRCh38
rs748809006 Chr.7: 155812098-155812098 on Build GRCh38
rs116412905 Chr.7: 155811806-155811806 on Build GRCh38
rs549733107 Chr.7: 155811906-155811906 on Build GRCh38
rs758966457 Chr.7: 155811810-155811810 on Build GRCh38
rs745751480 Chr.7: 155812084-155812084 on Build GRCh38
rs762641511 Chr.7: 155812000-155812000 on Build GRCh38
rs587778792 Chr.7: 155811823-155811823 on Build GRCh38
rs777486607 Chr.7: 155811917-155811917 on Build GRCh38
rs104894050 Chr.7: 155811860-155811860 on Build GRCh38
rs766928100 Chr.7: 155812111-155812113 on Build GRCh38
rs781079382 Chr.7: 155812082-155812082 on Build GRCh38
rs61729519 Chr.7: 155811988-155811988 on Build GRCh38
rs776420800 Chr.7: 155811940-155811940 on Build GRCh38
rs752787084 Chr.7: 155812061-155812061 on Build GRCh38
rs769976682 Chr.7: 155811919-155811919 on Build GRCh38
rs915392584 Chr.7: 155811768-155811768 on Build GRCh38
rs886038269 Chr.7: 155812074-155812074 on Build GRCh38
rs761113320 Chr.7: 155812024-155812024 on Build GRCh38
rs756628434 Chr.7: 155811861-155811861 on Build GRCh38
rs776142397 Chr.7: 155811809-155811809 on Build GRCh38
rs771827155 Chr.7: 155811796-155811796 on Build GRCh38
rs374343495 Chr.7: 155812063-155812063 on Build GRCh38
rs770780673 Chr.7: 155811925-155811925 on Build GRCh38
rs982760077 Chr.7: 155812014-155812014 on Build GRCh38
rs1011206201 Chr.7: 155811778-155811778 on Build GRCh38
rs200317108 Chr.7: 155811910-155811910 on Build GRCh38
rs1054316936 Chr.7: 155811742-155811742 on Build GRCh38
rs926620009 Chr.7: 155811729-155811729 on Build GRCh38
rs763849281 Chr.7: 155812012-155812012 on Build GRCh38
rs769792527 Chr.7: 155812090-155812090 on Build GRCh38
rs527879586 Chr.7: 155811879-155811879 on Build GRCh38
rs773026927 Chr.7: 155811802-155811802 on Build GRCh38
rs148181557 Chr.7: 155812071-155812071 on Build GRCh38
rs998395224 Chr.7: 155811831-155811831 on Build GRCh38
rs767953314 Chr.7: 155811846-155811846 on Build GRCh38
rs771163472 Chr.7: 155812109-155812109 on Build GRCh38
rs766797846 Chr.7: 155811873-155811873 on Build GRCh38
rs587778789 Chr.7: 155812078-155812085 on Build GRCh38
rs879671540 Chr.7: 155811743-155811744 on Build GRCh38
rs370338764 Chr.7: 155811961-155811961 on Build GRCh38
rs775095767 Chr.7: 155811997-155811997 on Build GRCh38
rs760920236 Chr.7: 155812079-155812079 on Build GRCh38
rs759907145 Chr.7: 155812123-155812123 on Build GRCh38
rs748454262 Chr.7: 155811784-155811784 on Build GRCh38
rs1019351832 Chr.7: 155811850-155811850 on Build GRCh38
rs745864537 Chr.7: 155811946-155811946 on Build GRCh38
rs764858271 Chr.7: 155811813-155811813 on Build GRCh38
rs774131404 Chr.7: 155812103-155812103 on Build GRCh38
rs28936675 Chr.7: 155812032-155812032 on Build GRCh38
rs141162945 Chr.7: 155812096-155812096 on Build GRCh38
rs553996031 Chr.7: 155812117-155812117 on Build GRCh38
rs200944712 Chr.7: 155811781-155811781 on Build GRCh38
rs938130628 Chr.7: 155812100-155812100 on Build GRCh38
rs750835058 Chr.7: 155811852-155811852 on Build GRCh38
rs761601401 Chr.7: 155812104-155812104 on Build GRCh38
rs779093031 Chr.7: 155811909-155811909 on Build GRCh38
rs1008384764 Chr.7: 155811847-155811847 on Build GRCh38
rs112915378 Chr.7: 155811882-155811882 on Build GRCh38
rs768854997 Chr.7: 155811772-155811772 on Build GRCh38
rs587778788 Chr.7: 155812113-155812114 on Build GRCh38
rs773641378 Chr.7: 155812018-155812018 on Build GRCh38
rs765185470 Chr.7: 155812054-155812054 on Build GRCh38
rs758646278 Chr.7: 155811913-155811913 on Build GRCh38
rs755543334 Chr.7: 155812035-155812035 on Build GRCh38
rs762733503 Chr.7: 155811820-155811820 on Build GRCh38
rs778992493 Chr.7: 155811773-155811773 on Build GRCh38
rs143084050 Chr.7: 155811923-155811923 on Build GRCh38

View Full Product Details