Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66831117-66831712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf44-SGK3
Entrez Gene ID
100533105
Gene Name
C8orf44-SGK3 readthrough
Gene Aliases
-
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66667552-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Gene Symbol
SGK3
Entrez Gene ID
23678
Gene Name
serum/glucocorticoid regulated kinase family member 3
Gene Aliases
CISK, SGK2, SGKL
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66712418-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631325 c.472A>G p.K158E Chr.8: 66831258-66831258 on Build GRCh38
7099779 c.305G>A p.G102E Chr.8: 66831292-66831292 on Build GRCh38
4631326 c.271A>G p.K91E Chr.8: 66831258-66831258 on Build GRCh38
76405 c.305G>C p.G102A Chr.8: 66831292-66831292 on Build GRCh38
2721126 c.483C>A p.D161E Chr.8: 66831269-66831269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570184266 Chr.8: 66831335-66831335 on Build GRCh38
rs967998780 Chr.8: 66831399-66831399 on Build GRCh38
rs768331284 Chr.8: 66831302-66831302 on Build GRCh38
rs1050552671 Chr.8: 66831499-66831499 on Build GRCh38
rs530024138 Chr.8: 66831215-66831215 on Build GRCh38
rs899209435 Chr.8: 66831357-66831357 on Build GRCh38
rs1006341609 Chr.8: 66831256-66831256 on Build GRCh38
rs374771800 Chr.8: 66831229-66831229 on Build GRCh38
rs1012919117 Chr.8: 66831631-66831631 on Build GRCh38
rs771785674 Chr.8: 66831277-66831277 on Build GRCh38
rs897975716 Chr.8: 66831583-66831583 on Build GRCh38
rs978975568 Chr.8: 66831490-66831490 on Build GRCh38
rs935031298 Chr.8: 66831175-66831175 on Build GRCh38
rs377492132 Chr.8: 66831353-66831353 on Build GRCh38
rs914955230 Chr.8: 66831174-66831174 on Build GRCh38
rs572654554 Chr.8: 66831444-66831444 on Build GRCh38
rs367787769 Chr.8: 66831187-66831187 on Build GRCh38
rs764197599 Chr.8: 66831359-66831359 on Build GRCh38
rs894587749 Chr.8: 66831612-66831612 on Build GRCh38
rs769637113 Chr.8: 66831336-66831336 on Build GRCh38
rs778123888 Chr.8: 66831234-66831234 on Build GRCh38
rs969362819 Chr.8: 66831391-66831391 on Build GRCh38
rs899272765 Chr.8: 66831632-66831632 on Build GRCh38
rs779918683 Chr.8: 66831285-66831285 on Build GRCh38
rs975837530 Chr.8: 66831465-66831465 on Build GRCh38
rs915031179 Chr.8: 66831464-66831464 on Build GRCh38
rs746253563 Chr.8: 66831354-66831354 on Build GRCh38
rs140982444 Chr.8: 66831527-66831527 on Build GRCh38
rs536547743 Chr.8: 66831191-66831191 on Build GRCh38
rs931657624 Chr.8: 66831479-66831479 on Build GRCh38
rs890666383 Chr.8: 66831539-66831539 on Build GRCh38
rs201904222 Chr.8: 66831237-66831237 on Build GRCh38
rs779288300 Chr.8: 66831271-66831271 on Build GRCh38
rs373982370 Chr.8: 66831342-66831343 on Build GRCh38
rs923040797 Chr.8: 66831468-66831468 on Build GRCh38
rs573286997 Chr.8: 66831675-66831675 on Build GRCh38
rs1050685264 Chr.8: 66831202-66831202 on Build GRCh38
rs781573649 Chr.8: 66831353-66831354 on Build GRCh38
rs752968703 Chr.8: 66831354-66831354 on Build GRCh38
rs923685650 Chr.8: 66831161-66831161 on Build GRCh38
rs756246042 Chr.8: 66831233-66831233 on Build GRCh38
rs751802557 Chr.8: 66831336-66831337 on Build GRCh38
rs1023873303 Chr.8: 66831373-66831373 on Build GRCh38
rs940702304 Chr.8: 66831603-66831603 on Build GRCh38
rs757736419 Chr.8: 66831239-66831239 on Build GRCh38
rs748447591 Chr.8: 66831663-66831663 on Build GRCh38
rs554885052 Chr.8: 66831531-66831531 on Build GRCh38
rs908885800 Chr.8: 66831516-66831516 on Build GRCh38
rs1057205671 Chr.8: 66831604-66831604 on Build GRCh38
rs1040537898 Chr.8: 66831179-66831179 on Build GRCh38
rs150210625 Chr.8: 66831689-66831689 on Build GRCh38
rs1013364156 Chr.8: 66831362-66831362 on Build GRCh38
rs900955763 Chr.8: 66831475-66831475 on Build GRCh38
rs55857008 Chr.8: 66831287-66831287 on Build GRCh38
rs767737490 Chr.8: 66831226-66831226 on Build GRCh38
rs538837016 Chr.8: 66831453-66831453 on Build GRCh38
rs776400322 Chr.8: 66831310-66831310 on Build GRCh38
rs746250144 Chr.8: 66831275-66831275 on Build GRCh38
rs200368158 Chr.8: 66831273-66831273 on Build GRCh38
rs1023693841 Chr.8: 66831672-66831672 on Build GRCh38
rs772999938 Chr.8: 66831338-66831338 on Build GRCh38
rs1002931654 Chr.8: 66831325-66831325 on Build GRCh38
rs760718060 Chr.8: 66831355-66831355 on Build GRCh38
rs979349149 Chr.8: 66831400-66831400 on Build GRCh38
rs1003477872 Chr.8: 66831584-66831584 on Build GRCh38
rs888063247 Chr.8: 66831216-66831216 on Build GRCh38
rs762738111 Chr.8: 66831347-66831347 on Build GRCh38
rs1023550455 Chr.8: 66831367-66831367 on Build GRCh38

View Full Product Details