Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144519799-144520351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC14
Entrez Gene ID
9684
Gene Name
leucine rich repeat containing 14
Gene Aliases
LRRC14A
UniGene
Hs.459391
Gene Chromosome Location
Chr.8: 144517965-144525178 on Build GRCh38
Transcripts
NM_001272036.1 NM_014665.3 XM_005272358.4 XM_005272359.4 XM_005272360.4 XM_017014005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7075317 c.80C>T p.P27L Chr.8: 144519805-144519805 on Build GRCh38
1623789 c.359C>T p.T120M Chr.8: 144520267-144520267 on Build GRCh38
1213797 c.362G>A p.G121D Chr.8: 144520270-144520270 on Build GRCh38
1239359 c.85G>A p.E29K Chr.8: 144519810-144519810 on Build GRCh38
1674342 c.152T>C p.V51A Chr.8: 144519877-144519877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766739715 Chr.8: 144520044-144520044 on Build GRCh38
rs116791876 Chr.8: 144519884-144519884 on Build GRCh38
rs763604937 Chr.8: 144520071-144520071 on Build GRCh38
rs185555518 Chr.8: 144519890-144519890 on Build GRCh38
rs767140744 Chr.8: 144520211-144520211 on Build GRCh38
rs756081440 Chr.8: 144520093-144520093 on Build GRCh38
rs114536271 Chr.8: 144520007-144520007 on Build GRCh38
rs765827521 Chr.8: 144520054-144520054 on Build GRCh38
rs777291199 Chr.8: 144519974-144519974 on Build GRCh38
rs779961185 Chr.8: 144520089-144520089 on Build GRCh38
rs758425438 Chr.8: 144520332-144520332 on Build GRCh38
rs766944855 Chr.8: 144520294-144520294 on Build GRCh38
rs777105669 Chr.8: 144520045-144520045 on Build GRCh38
rs746608334 Chr.8: 144519978-144519978 on Build GRCh38
rs755959705 Chr.8: 144520086-144520086 on Build GRCh38
rs553659773 Chr.8: 144519959-144519959 on Build GRCh38
rs1048758587 Chr.8: 144520064-144520064 on Build GRCh38
rs747099741 Chr.8: 144520097-144520097 on Build GRCh38
rs370798381 Chr.8: 144520196-144520196 on Build GRCh38
rs917037127 Chr.8: 144520030-144520030 on Build GRCh38
rs747694601 Chr.8: 144519873-144519873 on Build GRCh38
rs747101279 Chr.8: 144520252-144520252 on Build GRCh38
rs750141369 Chr.8: 144520212-144520212 on Build GRCh38
rs746032379 Chr.8: 144520265-144520265 on Build GRCh38
rs768490053 Chr.8: 144520091-144520091 on Build GRCh38
rs202101020 Chr.8: 144520246-144520246 on Build GRCh38
rs377265170 Chr.8: 144520076-144520076 on Build GRCh38
rs751051286 Chr.8: 144520073-144520073 on Build GRCh38
rs771258450 Chr.8: 144520103-144520103 on Build GRCh38
rs754789815 Chr.8: 144520323-144520323 on Build GRCh38
rs986103819 Chr.8: 144520178-144520178 on Build GRCh38
rs142979928 Chr.8: 144519867-144519867 on Build GRCh38
rs926790322 Chr.8: 144520315-144520315 on Build GRCh38
rs1022321290 Chr.8: 144520156-144520156 on Build GRCh38
rs756807238 Chr.8: 144520209-144520209 on Build GRCh38
rs573909145 Chr.8: 144520267-144520267 on Build GRCh38
rs772261420 Chr.8: 144520024-144520024 on Build GRCh38
rs745638321 Chr.8: 144520082-144520082 on Build GRCh38
rs953529982 Chr.8: 144520005-144520005 on Build GRCh38
rs1054466558 Chr.8: 144520074-144520074 on Build GRCh38
rs1012695900 Chr.8: 144520119-144520119 on Build GRCh38
rs780861071 Chr.8: 144520077-144520077 on Build GRCh38
rs1049520934 Chr.8: 144520155-144520155 on Build GRCh38
rs757988276 Chr.8: 144519970-144519970 on Build GRCh38
rs777680648 Chr.8: 144520133-144520133 on Build GRCh38
rs780412358 Chr.8: 144519864-144519864 on Build GRCh38
rs754279276 Chr.8: 144519960-144519960 on Build GRCh38
rs555879638 Chr.8: 144520251-144520251 on Build GRCh38
rs375567881 Chr.8: 144520299-144520299 on Build GRCh38
rs373589029 Chr.8: 144520072-144520072 on Build GRCh38
rs750547365 Chr.8: 144519854-144519854 on Build GRCh38
rs780912536 Chr.8: 144519995-144519995 on Build GRCh38
rs1045468423 Chr.8: 144520130-144520130 on Build GRCh38
rs775880960 Chr.8: 144520269-144520269 on Build GRCh38
rs774775277 Chr.8: 144520274-144520274 on Build GRCh38
rs773168879 Chr.8: 144520029-144520029 on Build GRCh38
rs930546846 Chr.8: 144520059-144520059 on Build GRCh38
rs764097760 Chr.8: 144519825-144519825 on Build GRCh38
rs993873474 Chr.8: 144520210-144520210 on Build GRCh38
rs1023781409 Chr.8: 144519861-144519861 on Build GRCh38
rs201052464 Chr.8: 144519821-144519821 on Build GRCh38
rs200791786 Chr.8: 144520201-144520201 on Build GRCh38
rs770166894 Chr.8: 144520268-144520268 on Build GRCh38
rs777052752 Chr.8: 144520104-144520104 on Build GRCh38
rs971923168 Chr.8: 144520191-144520191 on Build GRCh38
rs535632132 Chr.8: 144519842-144519842 on Build GRCh38
rs562811052 Chr.8: 144520308-144520308 on Build GRCh38
rs536560786 Chr.8: 144519946-144519946 on Build GRCh38
rs755544856 Chr.8: 144519868-144519868 on Build GRCh38
rs1005449138 Chr.8: 144520158-144520158 on Build GRCh38
rs759978108 Chr.8: 144520195-144520195 on Build GRCh38
rs773303549 Chr.8: 144520092-144520092 on Build GRCh38
rs189848203 Chr.8: 144520157-144520157 on Build GRCh38
rs893814235 Chr.8: 144519839-144519839 on Build GRCh38
rs138714591 Chr.8: 144520047-144520047 on Build GRCh38
rs911980537 Chr.8: 144520179-144520179 on Build GRCh38
rs771203297 Chr.8: 144520253-144520253 on Build GRCh38
rs769182169 Chr.8: 144520272-144520272 on Build GRCh38
rs776693030 Chr.8: 144519818-144519818 on Build GRCh38
rs760271314 Chr.8: 144520301-144520301 on Build GRCh38
rs901224909 Chr.8: 144520147-144520147 on Build GRCh38
rs750715244 Chr.8: 144519942-144519942 on Build GRCh38
rs770505495 Chr.8: 144519985-144519985 on Build GRCh38
rs1031232529 Chr.8: 144519878-144519878 on Build GRCh38
rs752233460 Chr.8: 144519965-144519965 on Build GRCh38
rs201360371 Chr.8: 144519883-144519883 on Build GRCh38
rs755765195 Chr.8: 144520242-144520242 on Build GRCh38
rs866475438 Chr.8: 144520148-144520148 on Build GRCh38
rs752513740 Chr.8: 144520331-144520331 on Build GRCh38
rs760940602 Chr.8: 144519947-144519947 on Build GRCh38
rs765679620 Chr.8: 144520193-144520193 on Build GRCh38
rs1036156327 Chr.8: 144520027-144520027 on Build GRCh38
rs774370003 Chr.8: 144519918-144519918 on Build GRCh38
rs754913719 Chr.8: 144520247-144520247 on Build GRCh38
rs1017294161 Chr.8: 144520186-144520186 on Build GRCh38
rs141246549 Chr.8: 144520244-144520244 on Build GRCh38
rs745691698 Chr.8: 144519997-144519997 on Build GRCh38
rs369321285 Chr.8: 144519895-144519895 on Build GRCh38
rs199766948 Chr.8: 144520271-144520271 on Build GRCh38
rs769683741 Chr.8: 144520003-144520003 on Build GRCh38
rs944540001 Chr.8: 144520009-144520009 on Build GRCh38
rs760892410 Chr.8: 144520041-144520041 on Build GRCh38
rs751649683 Chr.8: 144519836-144519836 on Build GRCh38
rs558133272 Chr.8: 144520066-144520066 on Build GRCh38
rs763471849 Chr.8: 144520197-144520197 on Build GRCh38
rs755326075 Chr.8: 144520044-144520044 on Build GRCh38
rs371214247 Chr.8: 144519870-144519870 on Build GRCh38
rs753645447 Chr.8: 144520313-144520313 on Build GRCh38
rs753326377 Chr.8: 144520055-144520055 on Build GRCh38
rs375290563 Chr.8: 144519843-144519843 on Build GRCh38
rs979863054 Chr.8: 144520314-144520314 on Build GRCh38
rs140702712 Chr.8: 144519896-144519896 on Build GRCh38
rs367779446 Chr.8: 144519869-144519869 on Build GRCh38
rs750719753 Chr.8: 144519846-144519846 on Build GRCh38
rs751044918 Chr.8: 144520208-144520208 on Build GRCh38
rs1042446984 Chr.8: 144520129-144520129 on Build GRCh38
rs1003670511 Chr.8: 144520137-144520137 on Build GRCh38
rs761917070 Chr.8: 144519941-144519941 on Build GRCh38
rs761307574 Chr.8: 144520292-144520292 on Build GRCh38
rs778907033 Chr.8: 144520326-144520326 on Build GRCh38
rs755503759 Chr.8: 144519961-144519961 on Build GRCh38
rs776858537 Chr.8: 144520259-144520259 on Build GRCh38
rs761785047 Chr.8: 144520008-144520008 on Build GRCh38

View Full Product Details