Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98250864-98251415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL4
Entrez Gene ID
84171
Gene Name
lysyl oxidase like 4
Gene Aliases
LOXC
UniGene
Hs.306814
Gene Chromosome Location
Chr.10: 98247686-98268250 on Build GRCh38
Transcripts
NM_032211.6 XM_005270216.1 XM_006718020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2057015 c.2153G>A p.R718H Chr.10: 98251112-98251112 on Build GRCh38
1345171 c.2101C>T p.P701S Chr.10: 98251164-98251164 on Build GRCh38
913672 c.2096T>C p.V699A Chr.10: 98251169-98251169 on Build GRCh38
913671 c.2198C>T p.T733I Chr.10: 98251067-98251067 on Build GRCh38
1289294 c.2109T>G p.Y703* Chr.10: 98251156-98251156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151114097 Chr.10: 98251112-98251112 on Build GRCh38
rs1032028781 Chr.10: 98250968-98250968 on Build GRCh38
rs762034324 Chr.10: 98251108-98251109 on Build GRCh38
rs773858721 Chr.10: 98251015-98251015 on Build GRCh38
rs932404846 Chr.10: 98251138-98251138 on Build GRCh38
rs183273492 Chr.10: 98250977-98250977 on Build GRCh38
rs937143701 Chr.10: 98250963-98250966 on Build GRCh38
rs768096929 Chr.10: 98251141-98251142 on Build GRCh38
rs751666160 Chr.10: 98251184-98251184 on Build GRCh38
rs930071440 Chr.10: 98251342-98251342 on Build GRCh38
rs377486850 Chr.10: 98251273-98251273 on Build GRCh38
rs1008346989 Chr.10: 98251235-98251235 on Build GRCh38
rs974295239 Chr.10: 98251367-98251367 on Build GRCh38
rs749247729 Chr.10: 98250920-98250920 on Build GRCh38
rs371865670 Chr.10: 98251029-98251029 on Build GRCh38
rs758403032 Chr.10: 98251058-98251058 on Build GRCh38
rs151200395 Chr.10: 98251056-98251056 on Build GRCh38
rs891714120 Chr.10: 98251282-98251282 on Build GRCh38
rs141069497 Chr.10: 98251152-98251152 on Build GRCh38
rs111706784 Chr.10: 98251359-98251359 on Build GRCh38
rs752004075 Chr.10: 98251062-98251063 on Build GRCh38
rs566764302 Chr.10: 98251033-98251033 on Build GRCh38
rs1031326203 Chr.10: 98251260-98251260 on Build GRCh38
rs770312604 Chr.10: 98251014-98251014 on Build GRCh38
rs562259622 Chr.10: 98250894-98250894 on Build GRCh38
rs751756187 Chr.10: 98251079-98251079 on Build GRCh38
rs11189523 Chr.10: 98251320-98251320 on Build GRCh38
rs780424742 Chr.10: 98251106-98251106 on Build GRCh38
rs1008470822 Chr.10: 98250986-98250986 on Build GRCh38
rs753717983 Chr.10: 98251045-98251045 on Build GRCh38
rs767125364 Chr.10: 98251027-98251027 on Build GRCh38
rs759409455 Chr.10: 98251145-98251145 on Build GRCh38
rs761237350 Chr.10: 98251019-98251019 on Build GRCh38
rs776932143 Chr.10: 98251299-98251299 on Build GRCh38
rs890858839 Chr.10: 98251164-98251164 on Build GRCh38
rs757507970 Chr.10: 98251084-98251084 on Build GRCh38
rs777964914 Chr.10: 98251059-98251059 on Build GRCh38
rs955978343 Chr.10: 98250970-98250970 on Build GRCh38
rs776511822 Chr.10: 98251139-98251139 on Build GRCh38
rs990422312 Chr.10: 98250974-98250974 on Build GRCh38
rs760364697 Chr.10: 98251115-98251115 on Build GRCh38
rs759838931 Chr.10: 98251312-98251312 on Build GRCh38
rs762994699 Chr.10: 98251155-98251155 on Build GRCh38
rs1039996950 Chr.10: 98251380-98251380 on Build GRCh38
rs540108978 Chr.10: 98251287-98251287 on Build GRCh38
rs552835081 Chr.10: 98251038-98251038 on Build GRCh38
rs981391532 Chr.10: 98251272-98251272 on Build GRCh38
rs7082202 Chr.10: 98251357-98251357 on Build GRCh38
rs770272930 Chr.10: 98251092-98251092 on Build GRCh38
rs200513650 Chr.10: 98251035-98251035 on Build GRCh38
rs78500686 Chr.10: 98251383-98251383 on Build GRCh38
rs927122791 Chr.10: 98250942-98250942 on Build GRCh38
rs1007878613 Chr.10: 98250966-98250966 on Build GRCh38
rs117258652 Chr.10: 98251363-98251363 on Build GRCh38
rs943997254 Chr.10: 98251369-98251369 on Build GRCh38
rs940254028 Chr.10: 98251104-98251104 on Build GRCh38
rs773208500 Chr.10: 98250978-98250978 on Build GRCh38
rs182642078 Chr.10: 98251292-98251292 on Build GRCh38
rs375946407 Chr.10: 98251123-98251123 on Build GRCh38
rs556627959 Chr.10: 98251089-98251089 on Build GRCh38
rs565208117 Chr.10: 98251288-98251288 on Build GRCh38
rs768083287 Chr.10: 98251187-98251188 on Build GRCh38
rs529765548 Chr.10: 98250964-98250964 on Build GRCh38
rs766247903 Chr.10: 98251040-98251040 on Build GRCh38
rs1028012341 Chr.10: 98250892-98250892 on Build GRCh38
rs980798111 Chr.10: 98251000-98251000 on Build GRCh38
rs781400495 Chr.10: 98251212-98251212 on Build GRCh38
rs971755697 Chr.10: 98251166-98251166 on Build GRCh38
rs192574171 Chr.10: 98251165-98251165 on Build GRCh38
rs764763769 Chr.10: 98251028-98251029 on Build GRCh38
rs538252025 Chr.10: 98251057-98251057 on Build GRCh38
rs752561194 Chr.10: 98251154-98251154 on Build GRCh38
rs750714906 Chr.10: 98251214-98251214 on Build GRCh38
rs369472739 Chr.10: 98251091-98251091 on Build GRCh38
rs992577473 Chr.10: 98251349-98251349 on Build GRCh38
rs749868336 Chr.10: 98251109-98251109 on Build GRCh38
rs140156290 Chr.10: 98251327-98251327 on Build GRCh38
rs373991076 Chr.10: 98251113-98251113 on Build GRCh38
rs867647710 Chr.10: 98251254-98251254 on Build GRCh38
rs768536931 Chr.10: 98250925-98250925 on Build GRCh38
rs939371770 Chr.10: 98251049-98251049 on Build GRCh38
rs573005885 Chr.10: 98251195-98251195 on Build GRCh38

View Full Product Details