Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102152358-102152692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOLC1
Entrez Gene ID
9221
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
NOPP130, NOPP140, NS5ATP13, P130
UniGene
Hs.523238
Gene Chromosome Location
Chr.10: 102152176-102163871 on Build GRCh38
Transcripts
NM_001284388.1 NM_001284389.1 NM_004741.4 XM_005270273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6727738 c.33C>T p.P11P Chr.10: 102152443-102152443 on Build GRCh38
5796938 c.19C>T p.R7C Chr.10: 102152429-102152429 on Build GRCh38
5369958 c.46C>T p.P16S Chr.10: 102152456-102152456 on Build GRCh38
5621392 c.44A>T p.Y15F Chr.10: 102152454-102152454 on Build GRCh38
287793 c.9C>T p.D3D Chr.10: 102152419-102152419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765570698 Chr.10: 102152546-102152546 on Build GRCh38
rs530128970 Chr.10: 102152595-102152595 on Build GRCh38
rs921499549 Chr.10: 102152663-102152663 on Build GRCh38
rs766592829 Chr.10: 102152432-102152432 on Build GRCh38
rs376493530 Chr.10: 102152524-102152524 on Build GRCh38
rs755302816 Chr.10: 102152390-102152390 on Build GRCh38
rs779583472 Chr.10: 102152559-102152559 on Build GRCh38
rs757586785 Chr.10: 102152604-102152604 on Build GRCh38
rs921493498 Chr.10: 102152568-102152568 on Build GRCh38
rs906518443 Chr.10: 102152619-102152620 on Build GRCh38
rs756303017 Chr.10: 102152384-102152384 on Build GRCh38
rs780908545 Chr.10: 102152450-102152450 on Build GRCh38
rs749648695 Chr.10: 102152389-102152389 on Build GRCh38
rs540452525 Chr.10: 102152419-102152419 on Build GRCh38
rs763937815 Chr.10: 102152377-102152377 on Build GRCh38
rs548669059 Chr.10: 102152641-102152641 on Build GRCh38
rs962876058 Chr.10: 102152623-102152623 on Build GRCh38
rs116928523 Chr.10: 102152452-102152452 on Build GRCh38
rs751449728 Chr.10: 102152378-102152378 on Build GRCh38
rs747974987 Chr.10: 102152387-102152396 on Build GRCh38
rs768373388 Chr.10: 102152470-102152470 on Build GRCh38
rs367617568 Chr.10: 102152607-102152607 on Build GRCh38
rs573072484 Chr.10: 102152416-102152416 on Build GRCh38
rs760756089 Chr.10: 102152520-102152520 on Build GRCh38
rs1042530301 Chr.10: 102152616-102152616 on Build GRCh38
rs11540480 Chr.10: 102152457-102152457 on Build GRCh38
rs553466488 Chr.10: 102152402-102152402 on Build GRCh38
rs755318558 Chr.10: 102152439-102152439 on Build GRCh38
rs750484400 Chr.10: 102152429-102152429 on Build GRCh38
rs562694950 Chr.10: 102152570-102152570 on Build GRCh38
rs373081405 Chr.10: 102152447-102152447 on Build GRCh38
rs371012896 Chr.10: 102152540-102152540 on Build GRCh38
rs748939408 Chr.10: 102152560-102152560 on Build GRCh38
rs767543288 Chr.10: 102152380-102152380 on Build GRCh38
rs751926786 Chr.10: 102152553-102152553 on Build GRCh38
rs771573999 Chr.10: 102152570-102152570 on Build GRCh38
rs780450364 Chr.10: 102152385-102152385 on Build GRCh38
rs745566242 Chr.10: 102152404-102152404 on Build GRCh38
rs974335954 Chr.10: 102152661-102152661 on Build GRCh38
rs753093011 Chr.10: 102152551-102152551 on Build GRCh38
rs752057770 Chr.10: 102152556-102152556 on Build GRCh38
rs748924182 Chr.10: 102152465-102152465 on Build GRCh38
rs370235771 Chr.10: 102152577-102152577 on Build GRCh38
rs761776310 Chr.10: 102152423-102152423 on Build GRCh38
rs769365402 Chr.10: 102152456-102152456 on Build GRCh38
rs746907163 Chr.10: 102152582-102152582 on Build GRCh38
rs754035972 Chr.10: 102152539-102152539 on Build GRCh38
rs762663571 Chr.10: 102152411-102152411 on Build GRCh38
rs779299614 Chr.10: 102152394-102152394 on Build GRCh38
rs771988687 Chr.10: 102152505-102152505 on Build GRCh38
rs142011703 Chr.10: 102152426-102152426 on Build GRCh38
rs747979755 Chr.10: 102152572-102152572 on Build GRCh38
rs934735980 Chr.10: 102152393-102152393 on Build GRCh38
rs868683880 Chr.10: 102152424-102152424 on Build GRCh38
rs774170597 Chr.10: 102152497-102152497 on Build GRCh38
rs369643164 Chr.10: 102152436-102152436 on Build GRCh38
rs769561812 Chr.10: 102152407-102152407 on Build GRCh38
rs775497484 Chr.10: 102152408-102152408 on Build GRCh38
rs771796372 Chr.10: 102152576-102152576 on Build GRCh38
rs145813353 Chr.10: 102152441-102152441 on Build GRCh38
rs117993238 Chr.10: 102152461-102152461 on Build GRCh38
rs754641843 Chr.10: 102152562-102152562 on Build GRCh38
rs760954180 Chr.10: 102152431-102152431 on Build GRCh38
rs945441194 Chr.10: 102152612-102152612 on Build GRCh38
rs372877879 Chr.10: 102152397-102152397 on Build GRCh38
rs1016178942 Chr.10: 102152622-102152622 on Build GRCh38
rs761711255 Chr.10: 102152504-102152504 on Build GRCh38
rs757299628 Chr.10: 102152379-102152379 on Build GRCh38
rs375712357 Chr.10: 102152557-102152557 on Build GRCh38
rs1002518893 Chr.10: 102152522-102152522 on Build GRCh38
rs372638591 Chr.10: 102152554-102152554 on Build GRCh38
rs750680222 Chr.10: 102152381-102152381 on Build GRCh38

View Full Product Details