Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88160705-88161082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
UniGene
Hs.331268
Gene Chromosome Location
Chr.12: 88142296-88199887 on Build GRCh38
Transcripts
NM_181783.3 XM_011537980.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695514 c.652G>T p.A218S Chr.12: 88160706-88160706 on Build GRCh38
1229838 c.690T>G p.F230L Chr.12: 88160744-88160744 on Build GRCh38
241950 c.667C>T p.R223C Chr.12: 88160721-88160721 on Build GRCh38
3466241 c.715C>T p.L239F Chr.12: 88160769-88160769 on Build GRCh38
6804800 c.696G>A p.M232I Chr.12: 88160750-88160750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573987495 Chr.12: 88161015-88161015 on Build GRCh38
rs776941667 Chr.12: 88160795-88160795 on Build GRCh38
rs945007558 Chr.12: 88160958-88160958 on Build GRCh38
rs778731724 Chr.12: 88160740-88160740 on Build GRCh38
rs984151681 Chr.12: 88160735-88160735 on Build GRCh38
rs535911494 Chr.12: 88161026-88161026 on Build GRCh38
rs977005627 Chr.12: 88160765-88160765 on Build GRCh38
rs932852070 Chr.12: 88160772-88160772 on Build GRCh38
rs749735504 Chr.12: 88160791-88160792 on Build GRCh38
rs771461068 Chr.12: 88160800-88160802 on Build GRCh38
rs778537565 Chr.12: 88160855-88160855 on Build GRCh38
rs769822072 Chr.12: 88160799-88160799 on Build GRCh38
rs1013476182 Chr.12: 88160773-88160773 on Build GRCh38
rs961345809 Chr.12: 88160753-88160753 on Build GRCh38
rs913480599 Chr.12: 88160972-88160972 on Build GRCh38
rs761970793 Chr.12: 88160798-88160798 on Build GRCh38
rs748569528 Chr.12: 88160898-88160898 on Build GRCh38
rs557883369 Chr.12: 88160807-88160807 on Build GRCh38
rs747531268 Chr.12: 88160775-88160775 on Build GRCh38
rs774792538 Chr.12: 88161009-88161009 on Build GRCh38
rs950585761 Chr.12: 88160792-88160792 on Build GRCh38
rs371029436 Chr.12: 88161021-88161021 on Build GRCh38
rs550195464 Chr.12: 88160988-88160988 on Build GRCh38
rs1053443048 Chr.12: 88160909-88160909 on Build GRCh38
rs142506336 Chr.12: 88161027-88161027 on Build GRCh38
rs753670426 Chr.12: 88160845-88160845 on Build GRCh38
rs575682323 Chr.12: 88160730-88160730 on Build GRCh38
rs780353221 Chr.12: 88160874-88160874 on Build GRCh38
rs922947960 Chr.12: 88160951-88160951 on Build GRCh38
rs773460854 Chr.12: 88160804-88160804 on Build GRCh38
rs375952824 Chr.12: 88161041-88161041 on Build GRCh38
rs868253887 Chr.12: 88160842-88160842 on Build GRCh38
rs183127275 Chr.12: 88160747-88160747 on Build GRCh38
rs747409904 Chr.12: 88160879-88160879 on Build GRCh38
rs752532664 Chr.12: 88160820-88160820 on Build GRCh38
rs2471545 Chr.12: 88160968-88160968 on Build GRCh38
rs974868536 Chr.12: 88160942-88160942 on Build GRCh38
rs758113950 Chr.12: 88160860-88160860 on Build GRCh38
rs568466760 Chr.12: 88161023-88161023 on Build GRCh38
rs367570629 Chr.12: 88160780-88160780 on Build GRCh38
rs756859773 Chr.12: 88160848-88160848 on Build GRCh38
rs371324002 Chr.12: 88160865-88160865 on Build GRCh38
rs964477444 Chr.12: 88160934-88160934 on Build GRCh38
rs780546810 Chr.12: 88160770-88160770 on Build GRCh38
rs760320102 Chr.12: 88160840-88160840 on Build GRCh38
rs892132558 Chr.12: 88160939-88160939 on Build GRCh38
rs774851374 Chr.12: 88160866-88160869 on Build GRCh38
rs769206632 Chr.12: 88160781-88160781 on Build GRCh38
rs745479134 Chr.12: 88160741-88160741 on Build GRCh38
rs991089560 Chr.12: 88160986-88160986 on Build GRCh38
rs560496532 Chr.12: 88161003-88161003 on Build GRCh38
rs772683343 Chr.12: 88160862-88160862 on Build GRCh38
rs577575304 Chr.12: 88160891-88160891 on Build GRCh38
rs764106545 Chr.12: 88160841-88160841 on Build GRCh38

View Full Product Details