Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105194995-105195544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3998646 c.1096-4A>G p.? Chr.12: 105195505-105195505 on Build GRCh38
5453572 c.1237C>T p.P413S Chr.12: 105195265-105195265 on Build GRCh38
1476068 c.1173T>G p.N391K Chr.12: 105195329-105195329 on Build GRCh38
2064887 c.1160C>T p.A387V Chr.12: 105195342-105195342 on Build GRCh38
1628340 c.1139C>T p.T380I Chr.12: 105195458-105195458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573034685 Chr.12: 105195224-105195224 on Build GRCh38
rs546005776 Chr.12: 105195462-105195462 on Build GRCh38
rs202240683 Chr.12: 105195280-105195280 on Build GRCh38
rs767934517 Chr.12: 105195460-105195460 on Build GRCh38
rs979628502 Chr.12: 105195128-105195128 on Build GRCh38
rs558853233 Chr.12: 105195342-105195342 on Build GRCh38
rs1035371550 Chr.12: 105195063-105195063 on Build GRCh38
rs748589283 Chr.12: 105195296-105195296 on Build GRCh38
rs893191071 Chr.12: 105195161-105195161 on Build GRCh38
rs766145339 Chr.12: 105195219-105195219 on Build GRCh38
rs60785893 Chr.12: 105195417-105195417 on Build GRCh38
rs765337599 Chr.12: 105195265-105195265 on Build GRCh38
rs762607854 Chr.12: 105195214-105195214 on Build GRCh38
rs771822415 Chr.12: 105195510-105195510 on Build GRCh38
rs562472275 Chr.12: 105195513-105195513 on Build GRCh38
rs1050677231 Chr.12: 105195156-105195156 on Build GRCh38
rs967716280 Chr.12: 105195396-105195396 on Build GRCh38
rs780047811 Chr.12: 105195511-105195511 on Build GRCh38
rs374828814 Chr.12: 105195518-105195518 on Build GRCh38
rs913616827 Chr.12: 105195098-105195098 on Build GRCh38
rs1011690287 Chr.12: 105195206-105195206 on Build GRCh38
rs552048602 Chr.12: 105195196-105195196 on Build GRCh38
rs756793583 Chr.12: 105195509-105195509 on Build GRCh38
rs907336761 Chr.12: 105195109-105195109 on Build GRCh38
rs764876603 Chr.12: 105195327-105195327 on Build GRCh38
rs1001717164 Chr.12: 105195149-105195149 on Build GRCh38
rs752630132 Chr.12: 105195237-105195237 on Build GRCh38
rs2272491 Chr.12: 105195505-105195505 on Build GRCh38
rs113988162 Chr.12: 105195133-105195133 on Build GRCh38
rs199942405 Chr.12: 105195281-105195281 on Build GRCh38
rs553335042 Chr.12: 105195287-105195287 on Build GRCh38
rs149088030 Chr.12: 105195341-105195341 on Build GRCh38
rs767842675 Chr.12: 105195322-105195323 on Build GRCh38
rs560954651 Chr.12: 105195399-105195399 on Build GRCh38
rs201186683 Chr.12: 105195429-105195429 on Build GRCh38
rs769306446 Chr.12: 105195297-105195297 on Build GRCh38
rs759395555 Chr.12: 105195335-105195335 on Build GRCh38
rs1036366173 Chr.12: 105195243-105195244 on Build GRCh38
rs763125021 Chr.12: 105195441-105195441 on Build GRCh38
rs960338869 Chr.12: 105195087-105195087 on Build GRCh38
rs1046017830 Chr.12: 105195018-105195018 on Build GRCh38
rs751693864 Chr.12: 105195371-105195371 on Build GRCh38
rs529863663 Chr.12: 105195415-105195415 on Build GRCh38
rs760736623 Chr.12: 105195428-105195428 on Build GRCh38
rs766647330 Chr.12: 105195361-105195361 on Build GRCh38
rs775913540 Chr.12: 105195363-105195363 on Build GRCh38
rs144982082 Chr.12: 105195273-105195273 on Build GRCh38
rs766105326 Chr.12: 105195167-105195167 on Build GRCh38
rs759740021 Chr.12: 105195457-105195457 on Build GRCh38
rs537120689 Chr.12: 105195210-105195210 on Build GRCh38
rs987777064 Chr.12: 105195089-105195089 on Build GRCh38
rs912144421 Chr.12: 105195215-105195216 on Build GRCh38
rs774189317 Chr.12: 105195223-105195223 on Build GRCh38
rs774837672 Chr.12: 105195230-105195230 on Build GRCh38
rs34775780 Chr.12: 105195344-105195344 on Build GRCh38
rs776387697 Chr.12: 105195432-105195432 on Build GRCh38
rs746922622 Chr.12: 105195516-105195516 on Build GRCh38
rs772695100 Chr.12: 105195319-105195319 on Build GRCh38
rs745441287 Chr.12: 105195419-105195419 on Build GRCh38
rs756609466 Chr.12: 105195398-105195398 on Build GRCh38
rs201986206 Chr.12: 105195395-105195395 on Build GRCh38
rs974965282 Chr.12: 105195349-105195349 on Build GRCh38
rs748958008 Chr.12: 105195322-105195322 on Build GRCh38
rs545699838 Chr.12: 105195325-105195325 on Build GRCh38
rs763136683 Chr.12: 105195356-105195356 on Build GRCh38
rs759930326 Chr.12: 105195280-105195281 on Build GRCh38
rs765483373 Chr.12: 105195343-105195343 on Build GRCh38
rs780205282 Chr.12: 105195385-105195386 on Build GRCh38
rs181908396 Chr.12: 105195323-105195323 on Build GRCh38
rs749914562 Chr.12: 105195329-105195330 on Build GRCh38
rs775775868 Chr.12: 105195202-105195202 on Build GRCh38
rs185692201 Chr.12: 105195456-105195456 on Build GRCh38
rs756662669 Chr.12: 105195468-105195468 on Build GRCh38
rs749776476 Chr.12: 105195480-105195480 on Build GRCh38
rs766304916 Chr.12: 105195445-105195445 on Build GRCh38
rs375729310 Chr.12: 105195226-105195226 on Build GRCh38
rs755374676 Chr.12: 105195392-105195392 on Build GRCh38
rs781479774 Chr.12: 105195393-105195393 on Build GRCh38
rs879169429 Chr.12: 105195154-105195154 on Build GRCh38
rs750736519 Chr.12: 105195268-105195268 on Build GRCh38
rs757439228 Chr.12: 105195246-105195246 on Build GRCh38
rs757966820 Chr.12: 105195368-105195369 on Build GRCh38
rs775424321 Chr.12: 105195423-105195423 on Build GRCh38
rs756886785 Chr.12: 105195325-105195326 on Build GRCh38
rs753028284 Chr.12: 105195466-105195466 on Build GRCh38
rs967771630 Chr.12: 105195112-105195112 on Build GRCh38
rs189200579 Chr.12: 105195201-105195201 on Build GRCh38
rs781687745 Chr.12: 105195289-105195289 on Build GRCh38
rs772002595 Chr.12: 105195420-105195420 on Build GRCh38
rs753052240 Chr.12: 105195323-105195324 on Build GRCh38

View Full Product Details