Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155140233-155140826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPM3
Entrez Gene ID
54344
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
Gene Aliases
CDG1O
UniGene
Hs.110477
Gene Chromosome Location
Chr.1: 155139891-155140573 on Build GRCh38
Transcripts
NM_018973.3 NM_153741.1 XM_017001498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576635 c.55C>A p.L19I Chr.1: 155140276-155140276 on Build GRCh38
4554743 c.62G>A p.G21E Chr.1: 155140269-155140269 on Build GRCh38
7089216 c.49C>G p.L17V Chr.1: 155140282-155140282 on Build GRCh38
2209936 c.50T>G p.L17R Chr.1: 155140281-155140281 on Build GRCh38
4608105 c.35T>G p.V12G Chr.1: 155140296-155140296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866669538 Chr.1: 155140556-155140556 on Build GRCh38
rs913229463 Chr.1: 155140522-155140522 on Build GRCh38
rs979284366 Chr.1: 155140508-155140508 on Build GRCh38
rs564809508 Chr.1: 155140666-155140666 on Build GRCh38
rs4971073 Chr.1: 155140628-155140628 on Build GRCh38
rs774556478 Chr.1: 155140403-155140403 on Build GRCh38
rs1054884567 Chr.1: 155140336-155140336 on Build GRCh38
rs1029570861 Chr.1: 155140496-155140496 on Build GRCh38
rs769012553 Chr.1: 155140347-155140347 on Build GRCh38
rs750010864 Chr.1: 155140514-155140514 on Build GRCh38
rs554351545 Chr.1: 155140447-155140447 on Build GRCh38
rs148133518 Chr.1: 155140607-155140607 on Build GRCh38
rs1005837935 Chr.1: 155140647-155140647 on Build GRCh38
rs550299793 Chr.1: 155140687-155140687 on Build GRCh38
rs768131340 Chr.1: 155140501-155140501 on Build GRCh38
rs772583366 Chr.1: 155140268-155140268 on Build GRCh38
rs560690638 Chr.1: 155140611-155140611 on Build GRCh38
rs148961715 Chr.1: 155140281-155140281 on Build GRCh38
rs916916042 Chr.1: 155140425-155140425 on Build GRCh38
rs58776162 Chr.1: 155140773-155140773 on Build GRCh38
rs928682253 Chr.1: 155140721-155140721 on Build GRCh38
rs996273999 Chr.1: 155140630-155140630 on Build GRCh38
rs995170681 Chr.1: 155140606-155140606 on Build GRCh38
rs201280096 Chr.1: 155140548-155140548 on Build GRCh38
rs577202972 Chr.1: 155140499-155140499 on Build GRCh38
rs374637597 Chr.1: 155140326-155140326 on Build GRCh38
rs921924553 Chr.1: 155140325-155140325 on Build GRCh38
rs558745578 Chr.1: 155140485-155140485 on Build GRCh38
rs764549754 Chr.1: 155140277-155140277 on Build GRCh38
rs971641569 Chr.1: 155140553-155140553 on Build GRCh38
rs762421905 Chr.1: 155140427-155140427 on Build GRCh38
rs1020724209 Chr.1: 155140500-155140500 on Build GRCh38
rs1013049349 Chr.1: 155140665-155140665 on Build GRCh38
rs967402101 Chr.1: 155140673-155140673 on Build GRCh38
rs566459400 Chr.1: 155140366-155140366 on Build GRCh38
rs761262436 Chr.1: 155140275-155140275 on Build GRCh38
rs754885305 Chr.1: 155140328-155140328 on Build GRCh38
rs12745576 Chr.1: 155140598-155140598 on Build GRCh38
rs750055766 Chr.1: 155140303-155140303 on Build GRCh38
rs114720269 Chr.1: 155140426-155140426 on Build GRCh38
rs532183286 Chr.1: 155140677-155140677 on Build GRCh38
rs151144405 Chr.1: 155140309-155140309 on Build GRCh38
rs764905012 Chr.1: 155140299-155140299 on Build GRCh38
rs1003452112 Chr.1: 155140409-155140409 on Build GRCh38
rs766156566 Chr.1: 155140313-155140313 on Build GRCh38
rs990844268 Chr.1: 155140656-155140656 on Build GRCh38
rs891851972 Chr.1: 155140353-155140353 on Build GRCh38
rs754875421 Chr.1: 155140643-155140643 on Build GRCh38
rs113849439 Chr.1: 155140489-155140489 on Build GRCh38
rs1045881299 Chr.1: 155140535-155140535 on Build GRCh38
rs1042203219 Chr.1: 155140375-155140375 on Build GRCh38
rs1038549118 Chr.1: 155140602-155140602 on Build GRCh38
rs1054982449 Chr.1: 155140564-155140564 on Build GRCh38
rs754751387 Chr.1: 155140619-155140619 on Build GRCh38
rs894784396 Chr.1: 155140424-155140424 on Build GRCh38
rs922185869 Chr.1: 155140502-155140502 on Build GRCh38
rs779141404 Chr.1: 155140360-155140360 on Build GRCh38
rs781233759 Chr.1: 155140608-155140608 on Build GRCh38
rs556862543 Chr.1: 155140529-155140529 on Build GRCh38
rs772491588 Chr.1: 155140379-155140379 on Build GRCh38
rs917470272 Chr.1: 155140538-155140538 on Build GRCh38
rs975000766 Chr.1: 155140323-155140323 on Build GRCh38
rs751736339 Chr.1: 155140555-155140555 on Build GRCh38
rs781011454 Chr.1: 155140334-155140334 on Build GRCh38
rs939189303 Chr.1: 155140557-155140557 on Build GRCh38
rs958784901 Chr.1: 155140664-155140664 on Build GRCh38
rs1029585220 Chr.1: 155140634-155140634 on Build GRCh38
rs566282280 Chr.1: 155140448-155140448 on Build GRCh38
rs965787618 Chr.1: 155140629-155140629 on Build GRCh38
rs746172014 Chr.1: 155140258-155140258 on Build GRCh38
rs1020336650 Chr.1: 155140305-155140305 on Build GRCh38
rs990135103 Chr.1: 155140516-155140516 on Build GRCh38
rs913226629 Chr.1: 155140662-155140662 on Build GRCh38
rs776185360 Chr.1: 155140278-155140278 on Build GRCh38
rs1019063610 Chr.1: 155140492-155140492 on Build GRCh38
rs937564539 Chr.1: 155140418-155140418 on Build GRCh38
rs41264901 Chr.1: 155140528-155140528 on Build GRCh38
rs926128894 Chr.1: 155140406-155140406 on Build GRCh38
rs185236597 Chr.1: 155140641-155140641 on Build GRCh38
rs1033624250 Chr.1: 155140423-155140423 on Build GRCh38
rs764245196 Chr.1: 155140549-155140549 on Build GRCh38
rs1004628491 Chr.1: 155140481-155140481 on Build GRCh38
rs966211544 Chr.1: 155140494-155140494 on Build GRCh38
rs749344531 Chr.1: 155140344-155140344 on Build GRCh38
rs776057897 Chr.1: 155140269-155140269 on Build GRCh38
rs534320339 Chr.1: 155140468-155140468 on Build GRCh38
rs941125229 Chr.1: 155140463-155140463 on Build GRCh38
rs950462595 Chr.1: 155140470-155140470 on Build GRCh38
rs531934453 Chr.1: 155140472-155140472 on Build GRCh38
rs537764711 Chr.1: 155140676-155140676 on Build GRCh38

View Full Product Details