Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36592012-36592603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGIF2
Entrez Gene ID
60436
Gene Name
TGFB induced factor homeobox 2
Gene Aliases
-
UniGene
Hs.632264
Gene Chromosome Location
Chr.20: 36573473-36593952 on Build GRCh38
Transcripts
NM_001199513.1 NM_001199514.1 NM_001199515.1 NM_021809.6 NM_001199535.1

Gene Symbol
TGIF2-C20orf24
Entrez Gene ID
100527943
Gene Name
TGIF2-C20orf24 readthrough
Gene Aliases
-
UniGene
Hs.584985
Gene Chromosome Location
Chr.20: 36574554-36612557 on Build GRCh38
Transcripts
NM_001199513.1 NM_001199514.1 NM_001199515.1 NM_021809.6 NM_001199535.1

SNP Detail

SNP ID Chromosome Location
rs943730712 Chr.20: 36592404-36592404 on Build GRCh38
rs980723841 Chr.20: 36592172-36592172 on Build GRCh38
rs997875782 Chr.20: 36592061-36592061 on Build GRCh38
rs1032563291 Chr.20: 36592581-36592581 on Build GRCh38
rs6101371 Chr.20: 36592082-36592082 on Build GRCh38
rs1051256028 Chr.20: 36592268-36592268 on Build GRCh38
rs755821096 Chr.20: 36592169-36592169 on Build GRCh38
rs992056772 Chr.20: 36592282-36592282 on Build GRCh38
rs995665610 Chr.20: 36592205-36592205 on Build GRCh38
rs932370808 Chr.20: 36592349-36592349 on Build GRCh38
rs933507539 Chr.20: 36592262-36592262 on Build GRCh38
rs1002011148 Chr.20: 36592486-36592486 on Build GRCh38
rs548061480 Chr.20: 36592380-36592380 on Build GRCh38
rs150382819 Chr.20: 36592511-36592511 on Build GRCh38
rs897378029 Chr.20: 36592448-36592448 on Build GRCh38
rs906044119 Chr.20: 36592485-36592485 on Build GRCh38
rs540350914 Chr.20: 36592341-36592341 on Build GRCh38
rs549686628 Chr.20: 36592467-36592467 on Build GRCh38
rs960559402 Chr.20: 36592180-36592180 on Build GRCh38
rs1004246270 Chr.20: 36592361-36592361 on Build GRCh38
rs964703069 Chr.20: 36592503-36592503 on Build GRCh38
rs200293257 Chr.20: 36592165-36592166 on Build GRCh38
rs901998916 Chr.20: 36592244-36592244 on Build GRCh38
rs6101373 Chr.20: 36592410-36592410 on Build GRCh38
rs764215917 Chr.20: 36592169-36592169 on Build GRCh38
rs979643553 Chr.20: 36592316-36592316 on Build GRCh38
rs566834781 Chr.20: 36592380-36592380 on Build GRCh38
rs4812359 Chr.20: 36592351-36592351 on Build GRCh38
rs138103023 Chr.20: 36592525-36592525 on Build GRCh38
rs932092798 Chr.20: 36592419-36592419 on Build GRCh38
rs1037274799 Chr.20: 36592198-36592198 on Build GRCh38
rs944266180 Chr.20: 36592188-36592188 on Build GRCh38
rs925145052 Chr.20: 36592318-36592318 on Build GRCh38
rs960505976 Chr.20: 36592397-36592399 on Build GRCh38
rs71351028 Chr.20: 36592558-36592558 on Build GRCh38
rs74748470 Chr.20: 36592212-36592212 on Build GRCh38
rs905903180 Chr.20: 36592213-36592213 on Build GRCh38
rs60964912 Chr.20: 36592181-36592182 on Build GRCh38
rs376493075 Chr.20: 36592494-36592494 on Build GRCh38
rs1002108294 Chr.20: 36592395-36592396 on Build GRCh38
rs897311359 Chr.20: 36592199-36592199 on Build GRCh38
rs966528845 Chr.20: 36592314-36592314 on Build GRCh38
rs895815211 Chr.20: 36592519-36592519 on Build GRCh38
rs533429497 Chr.20: 36592207-36592207 on Build GRCh38
rs75484069 Chr.20: 36592126-36592126 on Build GRCh38
rs1037243524 Chr.20: 36592435-36592435 on Build GRCh38
rs1020308019 Chr.20: 36592538-36592538 on Build GRCh38
rs1000809647 Chr.20: 36592564-36592564 on Build GRCh38
rs931585376 Chr.20: 36592459-36592459 on Build GRCh38
rs1017228845 Chr.20: 36592130-36592131 on Build GRCh38
rs556291931 Chr.20: 36592182-36592182 on Build GRCh38
rs925060357 Chr.20: 36592047-36592047 on Build GRCh38
rs1033504856 Chr.20: 36592180-36592183 on Build GRCh38
rs1033619667 Chr.20: 36592397-36592398 on Build GRCh38
rs1001643983 Chr.20: 36592218-36592218 on Build GRCh38
rs985241745 Chr.20: 36592374-36592374 on Build GRCh38
rs186240598 Chr.20: 36592141-36592141 on Build GRCh38
rs933455012 Chr.20: 36592538-36592539 on Build GRCh38
rs923535715 Chr.20: 36592521-36592521 on Build GRCh38
rs75858122 Chr.20: 36592386-36592386 on Build GRCh38
rs992007666 Chr.20: 36592404-36592405 on Build GRCh38
rs890812086 Chr.20: 36592187-36592187 on Build GRCh38
rs1036070144 Chr.20: 36592246-36592246 on Build GRCh38
rs925490222 Chr.20: 36592214-36592214 on Build GRCh38
rs1020920179 Chr.20: 36592298-36592298 on Build GRCh38
rs1049015531 Chr.20: 36592464-36592464 on Build GRCh38
rs898189322 Chr.20: 36592385-36592386 on Build GRCh38
rs966580114 Chr.20: 36592549-36592549 on Build GRCh38
rs1029387768 Chr.20: 36592076-36592076 on Build GRCh38
rs113760296 Chr.20: 36592386-36592386 on Build GRCh38
rs960056196 Chr.20: 36592252-36592252 on Build GRCh38
rs1039288558 Chr.20: 36592242-36592243 on Build GRCh38

View Full Product Details