Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44905796-44906395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAB
Entrez Gene ID
7529
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
Gene Aliases
GW128, HEL-S-1, HS1, KCIP-1, YWHAA
UniGene
Hs.643544
Gene Chromosome Location
Chr.20: 44885599-44908532 on Build GRCh38
Transcripts
NM_003404.4 NM_139323.3 XM_017028039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725292 c.622C>T p.L208L Chr.20: 44906034-44906034 on Build GRCh38
1729288 c.649A>T p.T217S Chr.20: 44906061-44906061 on Build GRCh38
478189 c.696G>A p.S232S Chr.20: 44906393-44906393 on Build GRCh38
6224405 c.620C>T p.T207M Chr.20: 44906032-44906032 on Build GRCh38
1270648 c.684+1_684+2insT p.? Chr.20: 44906097-44906098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16989481 Chr.20: 44906262-44906262 on Build GRCh38
rs764128781 Chr.20: 44906368-44906368 on Build GRCh38
rs368087121 Chr.20: 44906132-44906132 on Build GRCh38
rs1010881944 Chr.20: 44906158-44906158 on Build GRCh38
rs376067295 Chr.20: 44905976-44905976 on Build GRCh38
rs780163745 Chr.20: 44906314-44906314 on Build GRCh38
rs776953403 Chr.20: 44906048-44906048 on Build GRCh38
rs1038325559 Chr.20: 44905944-44905944 on Build GRCh38
rs938777133 Chr.20: 44905891-44905891 on Build GRCh38
rs754976856 Chr.20: 44906144-44906144 on Build GRCh38
rs753129464 Chr.20: 44906115-44906115 on Build GRCh38
rs778178549 Chr.20: 44906350-44906350 on Build GRCh38
rs200677018 Chr.20: 44906332-44906332 on Build GRCh38
rs1041213830 Chr.20: 44906113-44906113 on Build GRCh38
rs370845746 Chr.20: 44906117-44906117 on Build GRCh38
rs374058745 Chr.20: 44906032-44906032 on Build GRCh38
rs747357320 Chr.20: 44905924-44905924 on Build GRCh38
rs755545276 Chr.20: 44906051-44906051 on Build GRCh38
rs771556324 Chr.20: 44905951-44905951 on Build GRCh38
rs901378097 Chr.20: 44906133-44906133 on Build GRCh38
rs982185343 Chr.20: 44906220-44906220 on Build GRCh38
rs573702386 Chr.20: 44906116-44906116 on Build GRCh38
rs1028580589 Chr.20: 44906154-44906154 on Build GRCh38
rs927830862 Chr.20: 44905931-44905931 on Build GRCh38
rs947365468 Chr.20: 44905861-44905861 on Build GRCh38
rs758539031 Chr.20: 44906340-44906340 on Build GRCh38
rs966495111 Chr.20: 44905907-44905907 on Build GRCh38
rs776670359 Chr.20: 44906121-44906121 on Build GRCh38
rs562545819 Chr.20: 44906149-44906149 on Build GRCh38
rs367989746 Chr.20: 44906034-44906034 on Build GRCh38
rs975694730 Chr.20: 44905831-44905831 on Build GRCh38
rs140330183 Chr.20: 44906042-44906042 on Build GRCh38
rs557983598 Chr.20: 44905901-44905901 on Build GRCh38
rs535450863 Chr.20: 44905854-44905854 on Build GRCh38
rs539522672 Chr.20: 44905858-44905858 on Build GRCh38
rs913992775 Chr.20: 44905906-44905906 on Build GRCh38
rs372139934 Chr.20: 44906348-44906348 on Build GRCh38
rs371792112 Chr.20: 44906353-44906353 on Build GRCh38
rs758197754 Chr.20: 44905926-44905926 on Build GRCh38
rs186987230 Chr.20: 44906162-44906162 on Build GRCh38
rs756407085 Chr.20: 44906099-44906099 on Build GRCh38
rs765587390 Chr.20: 44905984-44905984 on Build GRCh38
rs989491681 Chr.20: 44905900-44905900 on Build GRCh38
rs771621179 Chr.20: 44906110-44906110 on Build GRCh38
rs761703648 Chr.20: 44906124-44906124 on Build GRCh38
rs542237358 Chr.20: 44906135-44906135 on Build GRCh38
rs924670259 Chr.20: 44905889-44905889 on Build GRCh38
rs894624112 Chr.20: 44905932-44905932 on Build GRCh38
rs1035263516 Chr.20: 44906231-44906231 on Build GRCh38
rs990791192 Chr.20: 44905933-44905933 on Build GRCh38
rs991110913 Chr.20: 44906247-44906247 on Build GRCh38
rs758119196 Chr.20: 44906354-44906354 on Build GRCh38
rs531108222 Chr.20: 44906155-44906155 on Build GRCh38
rs777490924 Chr.20: 44905978-44905978 on Build GRCh38
rs368643545 Chr.20: 44906340-44906340 on Build GRCh38
rs948918316 Chr.20: 44905941-44905941 on Build GRCh38
rs758417751 Chr.20: 44906018-44906018 on Build GRCh38
rs763907268 Chr.20: 44905972-44905972 on Build GRCh38
rs781631165 Chr.20: 44906062-44906062 on Build GRCh38
rs553553500 Chr.20: 44906103-44906103 on Build GRCh38
rs376330601 Chr.20: 44905969-44905969 on Build GRCh38
rs200157783 Chr.20: 44905979-44905979 on Build GRCh38
rs577867752 Chr.20: 44905920-44905920 on Build GRCh38
rs138834380 Chr.20: 44905882-44905882 on Build GRCh38
rs753146828 Chr.20: 44906334-44906334 on Build GRCh38
rs779524963 Chr.20: 44906373-44906373 on Build GRCh38
rs745592677 Chr.20: 44906107-44906107 on Build GRCh38
rs374163232 Chr.20: 44906141-44906141 on Build GRCh38
rs997129564 Chr.20: 44905872-44905874 on Build GRCh38
rs566827347 Chr.20: 44906295-44906295 on Build GRCh38
rs770559888 Chr.20: 44906168-44906168 on Build GRCh38
rs750604097 Chr.20: 44906001-44906001 on Build GRCh38
rs893761792 Chr.20: 44906156-44906156 on Build GRCh38
rs141399243 Chr.20: 44906239-44906239 on Build GRCh38
rs774850833 Chr.20: 44905958-44905958 on Build GRCh38
rs920540098 Chr.20: 44906253-44906253 on Build GRCh38
rs2425675 Chr.20: 44906293-44906293 on Build GRCh38
rs966754332 Chr.20: 44906191-44906191 on Build GRCh38
rs538185332 Chr.20: 44905890-44905890 on Build GRCh38
rs748546824 Chr.20: 44906064-44906064 on Build GRCh38

View Full Product Details