Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39250548-39251069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP4K3
Entrez Gene ID
8491
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases
GLK, MAPKKKK3, MEKKK 3, MEKKK3, RAB8IPL1
UniGene
Hs.655750
Gene Chromosome Location
Chr.2: 39249266-39437312 on Build GRCh38
Transcripts
NM_001270425.1 NM_003618.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1020887 c.2650T>C p.L884L Chr.2: 39250653-39250653 on Build GRCh38
6446621 c.2663C>T p.A888V Chr.2: 39250640-39250640 on Build GRCh38
5445030 c.2601C>A p.V867V Chr.2: 39250702-39250702 on Build GRCh38
6010082 c.2598-8A>G p.? Chr.2: 39250713-39250713 on Build GRCh38
6707553 c.2650T>G p.L884V Chr.2: 39250653-39250653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766982652 Chr.2: 39251003-39251003 on Build GRCh38
rs755849059 Chr.2: 39250659-39250659 on Build GRCh38
rs984633002 Chr.2: 39251001-39251001 on Build GRCh38
rs767371652 Chr.2: 39250615-39250615 on Build GRCh38
rs770956938 Chr.2: 39250735-39250735 on Build GRCh38
rs546701368 Chr.2: 39250925-39250925 on Build GRCh38
rs199919551 Chr.2: 39250639-39250639 on Build GRCh38
rs1003836640 Chr.2: 39250573-39250573 on Build GRCh38
rs930453186 Chr.2: 39250778-39250778 on Build GRCh38
rs181761046 Chr.2: 39250860-39250860 on Build GRCh38
rs914979779 Chr.2: 39250681-39250681 on Build GRCh38
rs1031260369 Chr.2: 39250621-39250621 on Build GRCh38
rs766434633 Chr.2: 39250576-39250576 on Build GRCh38
rs541164780 Chr.2: 39250716-39250716 on Build GRCh38
rs930589009 Chr.2: 39250978-39250978 on Build GRCh38
rs535246631 Chr.2: 39250898-39250898 on Build GRCh38
rs776582643 Chr.2: 39250608-39250610 on Build GRCh38
rs953154695 Chr.2: 39250948-39250948 on Build GRCh38
rs956967586 Chr.2: 39250896-39250896 on Build GRCh38
rs989666593 Chr.2: 39250650-39250650 on Build GRCh38
rs368775338 Chr.2: 39250750-39250750 on Build GRCh38
rs146648340 Chr.2: 39250702-39250702 on Build GRCh38
rs772991510 Chr.2: 39250731-39250731 on Build GRCh38
rs144778542 Chr.2: 39250964-39250964 on Build GRCh38
rs371729838 Chr.2: 39250857-39250857 on Build GRCh38
rs145819279 Chr.2: 39251019-39251019 on Build GRCh38
rs373967246 Chr.2: 39250757-39250757 on Build GRCh38
rs753487366 Chr.2: 39250670-39250670 on Build GRCh38
rs932853814 Chr.2: 39250671-39250671 on Build GRCh38
rs765333351 Chr.2: 39250728-39250732 on Build GRCh38
rs777297625 Chr.2: 39250662-39250662 on Build GRCh38
rs888525069 Chr.2: 39250797-39250797 on Build GRCh38
rs763212958 Chr.2: 39250734-39250734 on Build GRCh38
rs531067576 Chr.2: 39250822-39250822 on Build GRCh38
rs1031234824 Chr.2: 39250863-39250863 on Build GRCh38
rs759531523 Chr.2: 39250740-39250740 on Build GRCh38
rs941770849 Chr.2: 39251002-39251002 on Build GRCh38
rs148411410 Chr.2: 39250693-39250693 on Build GRCh38
rs893870601 Chr.2: 39250859-39250859 on Build GRCh38
rs1011066565 Chr.2: 39250937-39250937 on Build GRCh38
rs906486932 Chr.2: 39250855-39250855 on Build GRCh38
rs967138312 Chr.2: 39250895-39250895 on Build GRCh38
rs1012779172 Chr.2: 39250805-39250805 on Build GRCh38
rs919155559 Chr.2: 39250971-39250971 on Build GRCh38
rs906635880 Chr.2: 39251044-39251044 on Build GRCh38
rs139585680 Chr.2: 39250940-39250940 on Build GRCh38
rs202190387 Chr.2: 39250736-39250736 on Build GRCh38
rs766168735 Chr.2: 39250645-39250645 on Build GRCh38
rs761689240 Chr.2: 39250724-39250725 on Build GRCh38
rs575419845 Chr.2: 39250611-39250611 on Build GRCh38
rs549689134 Chr.2: 39250830-39250830 on Build GRCh38
rs751501168 Chr.2: 39250584-39250584 on Build GRCh38
rs756737289 Chr.2: 39250680-39250680 on Build GRCh38
rs971720575 Chr.2: 39250967-39250967 on Build GRCh38
rs965651998 Chr.2: 39250965-39250965 on Build GRCh38
rs986247538 Chr.2: 39250950-39250950 on Build GRCh38
rs1049389116 Chr.2: 39250780-39250780 on Build GRCh38
rs748535236 Chr.2: 39250725-39250725 on Build GRCh38
rs770246454 Chr.2: 39250726-39250726 on Build GRCh38
rs985553608 Chr.2: 39250992-39250992 on Build GRCh38
rs115499149 Chr.2: 39250821-39250821 on Build GRCh38
rs573034476 Chr.2: 39250712-39250712 on Build GRCh38
rs559449542 Chr.2: 39250728-39250728 on Build GRCh38
rs928997231 Chr.2: 39251031-39251031 on Build GRCh38
rs1045521158 Chr.2: 39251028-39251028 on Build GRCh38

View Full Product Details