Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29131945-29132475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP4
Entrez Gene ID
79745
Gene Name
CAP-Gly domain containing linker protein family member 4
Gene Aliases
RSNL2
UniGene
Hs.122927
Gene Chromosome Location
Chr.2: 29097676-29183813 on Build GRCh38
Transcripts
NM_001287527.1 NM_001287528.1 NM_024692.5 NR_109844.1 XM_005264562.2 XM_005264563.1 XM_006712104.2 XM_006712105.3 XM_011533109.1 XM_011533110.1 XM_017004958.1 XM_017004959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019991 c.278T>C p.L93P Chr.2: 29132156-29132156 on Build GRCh38
7147500 c.297G>C p.V99V Chr.2: 29132175-29132175 on Build GRCh38
5717007 c.353G>A p.G118E Chr.2: 29132231-29132231 on Build GRCh38
7147499 c.297G>C p.V99V Chr.2: 29132175-29132175 on Build GRCh38
4751850 c.287G>A p.G96D Chr.2: 29132165-29132165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756363152 Chr.2: 29132360-29132360 on Build GRCh38
rs180843843 Chr.2: 29132031-29132031 on Build GRCh38
rs151063938 Chr.2: 29132061-29132061 on Build GRCh38
rs377595609 Chr.2: 29132372-29132372 on Build GRCh38
rs1019422553 Chr.2: 29132405-29132405 on Build GRCh38
rs552096947 Chr.2: 29132340-29132340 on Build GRCh38
rs1008910870 Chr.2: 29132402-29132402 on Build GRCh38
rs751088392 Chr.2: 29132157-29132157 on Build GRCh38
rs549775636 Chr.2: 29132306-29132306 on Build GRCh38
rs771672135 Chr.2: 29132104-29132104 on Build GRCh38
rs200932113 Chr.2: 29132107-29132107 on Build GRCh38
rs776743176 Chr.2: 29132225-29132225 on Build GRCh38
rs766492294 Chr.2: 29132264-29132264 on Build GRCh38
rs991063855 Chr.2: 29132269-29132269 on Build GRCh38
rs750419999 Chr.2: 29132143-29132143 on Build GRCh38
rs754469018 Chr.2: 29132271-29132271 on Build GRCh38
rs776056684 Chr.2: 29132215-29132215 on Build GRCh38
rs888485369 Chr.2: 29132072-29132072 on Build GRCh38
rs112800473 Chr.2: 29132007-29132007 on Build GRCh38
rs184949678 Chr.2: 29132330-29132330 on Build GRCh38
rs754507362 Chr.2: 29132158-29132158 on Build GRCh38
rs561000032 Chr.2: 29132070-29132070 on Build GRCh38
rs1022490454 Chr.2: 29132318-29132318 on Build GRCh38
rs779180511 Chr.2: 29132208-29132208 on Build GRCh38
rs4666167 Chr.2: 29132437-29132437 on Build GRCh38
rs934372421 Chr.2: 29132378-29132378 on Build GRCh38
rs976616867 Chr.2: 29132383-29132383 on Build GRCh38
rs138906439 Chr.2: 29132329-29132329 on Build GRCh38
rs989011559 Chr.2: 29132455-29132455 on Build GRCh38
rs1038218202 Chr.2: 29132308-29132308 on Build GRCh38
rs760705316 Chr.2: 29132222-29132222 on Build GRCh38
rs765130877 Chr.2: 29132242-29132242 on Build GRCh38
rs754063786 Chr.2: 29132109-29132109 on Build GRCh38
rs879544193 Chr.2: 29132022-29132022 on Build GRCh38
rs965210842 Chr.2: 29131968-29131968 on Build GRCh38
rs901099678 Chr.2: 29132036-29132036 on Build GRCh38
rs189282917 Chr.2: 29132391-29132391 on Build GRCh38
rs959562551 Chr.2: 29132229-29132229 on Build GRCh38
rs937131653 Chr.2: 29132272-29132272 on Build GRCh38
rs747787199 Chr.2: 29132172-29132172 on Build GRCh38
rs560056289 Chr.2: 29132018-29132018 on Build GRCh38
rs772623218 Chr.2: 29132213-29132213 on Build GRCh38
rs912883508 Chr.2: 29132165-29132165 on Build GRCh38
rs762828573 Chr.2: 29132261-29132261 on Build GRCh38
rs771314138 Chr.2: 29132201-29132201 on Build GRCh38
rs957560968 Chr.2: 29132445-29132445 on Build GRCh38
rs779726591 Chr.2: 29132153-29132153 on Build GRCh38
rs571241625 Chr.2: 29132316-29132316 on Build GRCh38
rs978372767 Chr.2: 29132346-29132346 on Build GRCh38
rs377382035 Chr.2: 29132151-29132151 on Build GRCh38
rs768763294 Chr.2: 29132224-29132224 on Build GRCh38
rs780755878 Chr.2: 29132160-29132160 on Build GRCh38
rs766203839 Chr.2: 29132406-29132406 on Build GRCh38
rs949776287 Chr.2: 29132185-29132185 on Build GRCh38
rs764259646 Chr.2: 29132379-29132379 on Build GRCh38
rs750186969 Chr.2: 29132258-29132258 on Build GRCh38
rs751492263 Chr.2: 29132266-29132266 on Build GRCh38
rs898365487 Chr.2: 29132312-29132312 on Build GRCh38
rs762090158 Chr.2: 29132233-29132233 on Build GRCh38
rs780711521 Chr.2: 29132281-29132281 on Build GRCh38
rs974834349 Chr.2: 29132424-29132424 on Build GRCh38
rs974897962 Chr.2: 29131974-29131974 on Build GRCh38
rs765078001 Chr.2: 29132115-29132115 on Build GRCh38
rs1005545384 Chr.2: 29132080-29132080 on Build GRCh38

View Full Product Details