Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24337571-24338162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIPK3
Entrez Gene ID
11035
Gene Name
receptor interacting serine/threonine kinase 3
Gene Aliases
RIP3
UniGene
Hs.268551
Gene Chromosome Location
Chr.14: 24336021-24340036 on Build GRCh38
Transcripts
NM_006871.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045961 c.832+9G>T p.? Chr.14: 24337864-24337864 on Build GRCh38
4847371 c.774G>A p.L258L Chr.14: 24337931-24337931 on Build GRCh38
6032766 c.716G>A p.R239Q Chr.14: 24337989-24337989 on Build GRCh38
5905969 c.833-7C>T p.? Chr.14: 24337769-24337769 on Build GRCh38
6766868 c.767_769delAAG p.E256delE Chr.14: 24337936-24337938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114413869 Chr.14: 24337989-24337989 on Build GRCh38
rs768125006 Chr.14: 24338072-24338072 on Build GRCh38
rs777309093 Chr.14: 24337593-24337593 on Build GRCh38
rs752461948 Chr.14: 24337645-24337645 on Build GRCh38
rs575662398 Chr.14: 24337903-24337903 on Build GRCh38
rs776458938 Chr.14: 24337968-24337968 on Build GRCh38
rs771638456 Chr.14: 24338109-24338109 on Build GRCh38
rs769847566 Chr.14: 24337639-24337639 on Build GRCh38
rs958023653 Chr.14: 24338028-24338028 on Build GRCh38
rs761512630 Chr.14: 24337755-24337755 on Build GRCh38
rs756762291 Chr.14: 24337649-24337649 on Build GRCh38
rs754119477 Chr.14: 24338138-24338138 on Build GRCh38
rs760490173 Chr.14: 24337739-24337739 on Build GRCh38
rs753498898 Chr.14: 24337752-24337752 on Build GRCh38
rs781680919 Chr.14: 24337781-24337781 on Build GRCh38
rs1046877212 Chr.14: 24338124-24338124 on Build GRCh38
rs761384731 Chr.14: 24338139-24338139 on Build GRCh38
rs769825688 Chr.14: 24337695-24337695 on Build GRCh38
rs747996310 Chr.14: 24337975-24337975 on Build GRCh38
rs190925372 Chr.14: 24338069-24338069 on Build GRCh38
rs930939956 Chr.14: 24338122-24338122 on Build GRCh38
rs761101910 Chr.14: 24338012-24338012 on Build GRCh38
rs764919774 Chr.14: 24337864-24337864 on Build GRCh38
rs752047201 Chr.14: 24337777-24337777 on Build GRCh38
rs114213493 Chr.14: 24337902-24337902 on Build GRCh38
rs762016289 Chr.14: 24337637-24337637 on Build GRCh38
rs374322185 Chr.14: 24337754-24337754 on Build GRCh38
rs1007815233 Chr.14: 24337722-24337722 on Build GRCh38
rs750949099 Chr.14: 24337655-24337655 on Build GRCh38
rs371790419 Chr.14: 24338087-24338087 on Build GRCh38
rs756134603 Chr.14: 24337799-24337801 on Build GRCh38
rs776405582 Chr.14: 24337854-24337854 on Build GRCh38
rs749917612 Chr.14: 24337761-24337761 on Build GRCh38
rs866753680 Chr.14: 24337885-24337885 on Build GRCh38
rs746888867 Chr.14: 24337843-24337843 on Build GRCh38
rs781160593 Chr.14: 24337867-24337867 on Build GRCh38
rs748219287 Chr.14: 24337890-24337890 on Build GRCh38
rs748451526 Chr.14: 24337810-24337810 on Build GRCh38
rs764583913 Chr.14: 24338136-24338136 on Build GRCh38
rs747014964 Chr.14: 24337960-24337960 on Build GRCh38
rs139820893 Chr.14: 24337644-24337644 on Build GRCh38
rs780322549 Chr.14: 24337662-24337662 on Build GRCh38
rs747354146 Chr.14: 24337669-24337669 on Build GRCh38
rs749012241 Chr.14: 24338079-24338079 on Build GRCh38
rs755408854 Chr.14: 24337779-24337779 on Build GRCh38
rs760172312 Chr.14: 24338004-24338004 on Build GRCh38
rs757340329 Chr.14: 24337946-24337946 on Build GRCh38
rs781571121 Chr.14: 24337686-24337686 on Build GRCh38
rs778424966 Chr.14: 24337654-24337654 on Build GRCh38
rs775353337 Chr.14: 24337724-24337724 on Build GRCh38
rs780451764 Chr.14: 24337601-24337601 on Build GRCh38
rs371715531 Chr.14: 24337826-24337826 on Build GRCh38
rs959899388 Chr.14: 24338106-24338106 on Build GRCh38
rs45537534 Chr.14: 24338096-24338096 on Build GRCh38
rs772028370 Chr.14: 24337708-24337708 on Build GRCh38
rs571024283 Chr.14: 24337821-24337821 on Build GRCh38
rs547625454 Chr.14: 24337811-24337811 on Build GRCh38
rs7153640 Chr.14: 24337926-24337926 on Build GRCh38
rs34106261 Chr.14: 24337696-24337696 on Build GRCh38
rs781220787 Chr.14: 24338033-24338033 on Build GRCh38
rs373898638 Chr.14: 24337990-24337990 on Build GRCh38
rs915911463 Chr.14: 24337981-24337981 on Build GRCh38
rs56337365 Chr.14: 24338015-24338015 on Build GRCh38
rs753443712 Chr.14: 24337648-24337648 on Build GRCh38
rs747302549 Chr.14: 24337611-24337611 on Build GRCh38
rs563544738 Chr.14: 24338098-24338098 on Build GRCh38
rs756388466 Chr.14: 24337921-24337921 on Build GRCh38
rs867455841 Chr.14: 24337880-24337880 on Build GRCh38
rs757463085 Chr.14: 24338017-24338017 on Build GRCh38
rs761071452 Chr.14: 24338117-24338117 on Build GRCh38
rs749400287 Chr.14: 24337945-24337945 on Build GRCh38
rs28620990 Chr.14: 24337646-24337646 on Build GRCh38
rs1000676766 Chr.14: 24337673-24337673 on Build GRCh38
rs148293187 Chr.14: 24337961-24337961 on Build GRCh38
rs970897515 Chr.14: 24338077-24338077 on Build GRCh38
rs753087053 Chr.14: 24337919-24337919 on Build GRCh38
rs368542701 Chr.14: 24337938-24337938 on Build GRCh38
rs769409655 Chr.14: 24337984-24337984 on Build GRCh38
rs768968523 Chr.14: 24337615-24337615 on Build GRCh38
rs761410881 Chr.14: 24337860-24337860 on Build GRCh38
rs370452891 Chr.14: 24337819-24337819 on Build GRCh38
rs908274636 Chr.14: 24338135-24338135 on Build GRCh38
rs267603967 Chr.14: 24337952-24337952 on Build GRCh38
rs993199113 Chr.14: 24337958-24337958 on Build GRCh38
rs748324721 Chr.14: 24337692-24337692 on Build GRCh38
rs199742695 Chr.14: 24338032-24338032 on Build GRCh38
rs762530127 Chr.14: 24337871-24337871 on Build GRCh38
rs770873022 Chr.14: 24337832-24337832 on Build GRCh38
rs769689175 Chr.14: 24338035-24338035 on Build GRCh38
rs368257721 Chr.14: 24337822-24337822 on Build GRCh38
rs931059849 Chr.14: 24337977-24337977 on Build GRCh38
rs1950501 Chr.14: 24337594-24337594 on Build GRCh38
rs758937355 Chr.14: 24337656-24337656 on Build GRCh38
rs371354196 Chr.14: 24337962-24337962 on Build GRCh38
rs1020965663 Chr.14: 24338055-24338055 on Build GRCh38
rs539787075 Chr.14: 24337835-24337835 on Build GRCh38
rs777568739 Chr.14: 24337958-24337958 on Build GRCh38
rs764304131 Chr.14: 24338014-24338014 on Build GRCh38
rs369990803 Chr.14: 24338020-24338020 on Build GRCh38
rs776075051 Chr.14: 24338115-24338115 on Build GRCh38
rs952956124 Chr.14: 24338084-24338084 on Build GRCh38
rs949531291 Chr.14: 24337818-24337818 on Build GRCh38
rs758877093 Chr.14: 24337766-24337766 on Build GRCh38
rs869212791 Chr.14: 24337817-24337817 on Build GRCh38
rs765466321 Chr.14: 24338140-24338140 on Build GRCh38
rs374696778 Chr.14: 24337992-24337992 on Build GRCh38
rs368370166 Chr.14: 24338061-24338061 on Build GRCh38
rs374237537 Chr.14: 24337918-24337918 on Build GRCh38
rs765929505 Chr.14: 24337883-24337883 on Build GRCh38
rs903689555 Chr.14: 24337672-24337672 on Build GRCh38
rs568424822 Chr.14: 24337675-24337675 on Build GRCh38
rs763808561 Chr.14: 24337740-24337740 on Build GRCh38
rs768494548 Chr.14: 24337853-24337853 on Build GRCh38
rs964087733 Chr.14: 24337789-24337789 on Build GRCh38
rs372654613 Chr.14: 24338021-24338021 on Build GRCh38
rs776944621 Chr.14: 24337622-24337622 on Build GRCh38
rs562904093 Chr.14: 24337595-24337595 on Build GRCh38
rs775406379 Chr.14: 24337836-24337836 on Build GRCh38
rs764718796 Chr.14: 24337758-24337758 on Build GRCh38
rs761571685 Chr.14: 24337920-24337920 on Build GRCh38
rs996941995 Chr.14: 24338047-24338047 on Build GRCh38
rs768165733 Chr.14: 24338007-24338007 on Build GRCh38

View Full Product Details