Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117048840-117049439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TESC
Entrez Gene ID
54997
Gene Name
tescalcin
Gene Aliases
CHP3, TSC
UniGene
Hs.525709
Gene Chromosome Location
Chr.12: 117038923-117099490 on Build GRCh38
Transcripts
NM_001168325.1 NM_017899.3 NR_031766.2 XM_011538503.1 XM_017019532.1 XM_017019533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398399 c.445C>T p.R149W Chr.12: 117049082-117049082 on Build GRCh38
5909309 c.382G>A p.G128R Chr.12: 117049145-117049145 on Build GRCh38
3731316 c.376C>T p.R126C Chr.12: 117049151-117049151 on Build GRCh38
5024644 c.382G>T p.G128* Chr.12: 117049145-117049145 on Build GRCh38
3811150 c.403G>C p.E135Q Chr.12: 117049124-117049124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893559659 Chr.12: 117049282-117049282 on Build GRCh38
rs572125124 Chr.12: 117049391-117049391 on Build GRCh38
rs527239655 Chr.12: 117048981-117048981 on Build GRCh38
rs772630732 Chr.12: 117049144-117049144 on Build GRCh38
rs868535508 Chr.12: 117049055-117049055 on Build GRCh38
rs1003361456 Chr.12: 117049327-117049327 on Build GRCh38
rs760780955 Chr.12: 117049415-117049415 on Build GRCh38
rs570252037 Chr.12: 117049113-117049113 on Build GRCh38
rs906390320 Chr.12: 117049316-117049316 on Build GRCh38
rs530406783 Chr.12: 117049094-117049094 on Build GRCh38
rs143285421 Chr.12: 117049081-117049081 on Build GRCh38
rs372715773 Chr.12: 117049182-117049182 on Build GRCh38
rs544806935 Chr.12: 117049368-117049368 on Build GRCh38
rs756953606 Chr.12: 117049029-117049031 on Build GRCh38
rs183572326 Chr.12: 117049193-117049193 on Build GRCh38
rs936468673 Chr.12: 117049060-117049060 on Build GRCh38
rs758234232 Chr.12: 117049101-117049101 on Build GRCh38
rs755734021 Chr.12: 117049205-117049205 on Build GRCh38
rs1041745866 Chr.12: 117049369-117049369 on Build GRCh38
rs376962711 Chr.12: 117048968-117048968 on Build GRCh38
rs145583765 Chr.12: 117049202-117049202 on Build GRCh38
rs750698318 Chr.12: 117049179-117049179 on Build GRCh38
rs754155709 Chr.12: 117049013-117049013 on Build GRCh38
rs757275678 Chr.12: 117049089-117049089 on Build GRCh38
rs745683243 Chr.12: 117049037-117049037 on Build GRCh38
rs540805043 Chr.12: 117049413-117049413 on Build GRCh38
rs756257303 Chr.12: 117049002-117049002 on Build GRCh38
rs377645691 Chr.12: 117049366-117049366 on Build GRCh38
rs977982079 Chr.12: 117049207-117049207 on Build GRCh38
rs193137390 Chr.12: 117049074-117049074 on Build GRCh38
rs749955911 Chr.12: 117049122-117049124 on Build GRCh38
rs148314787 Chr.12: 117049119-117049119 on Build GRCh38
rs148450268 Chr.12: 117049233-117049233 on Build GRCh38
rs368694775 Chr.12: 117049162-117049162 on Build GRCh38
rs1032964583 Chr.12: 117049393-117049393 on Build GRCh38
rs374122867 Chr.12: 117049112-117049112 on Build GRCh38
rs772952061 Chr.12: 117049076-117049076 on Build GRCh38
rs920660320 Chr.12: 117048945-117048945 on Build GRCh38
rs35765692 Chr.12: 117048871-117048872 on Build GRCh38
rs867618785 Chr.12: 117049038-117049039 on Build GRCh38
rs780004006 Chr.12: 117049058-117049058 on Build GRCh38
rs778631702 Chr.12: 117049103-117049103 on Build GRCh38
rs759924790 Chr.12: 117048991-117048991 on Build GRCh38
rs143884130 Chr.12: 117049077-117049077 on Build GRCh38
rs141471213 Chr.12: 117049152-117049152 on Build GRCh38
rs983910842 Chr.12: 117049402-117049402 on Build GRCh38
rs761081800 Chr.12: 117049085-117049085 on Build GRCh38
rs768274684 Chr.12: 117049068-117049068 on Build GRCh38
rs753875219 Chr.12: 117049087-117049087 on Build GRCh38
rs574767106 Chr.12: 117049241-117049241 on Build GRCh38
rs200120071 Chr.12: 117049188-117049188 on Build GRCh38
rs576130884 Chr.12: 117049409-117049409 on Build GRCh38
rs753592055 Chr.12: 117049026-117049028 on Build GRCh38
rs779047232 Chr.12: 117049016-117049016 on Build GRCh38
rs187524553 Chr.12: 117049367-117049367 on Build GRCh38
rs149789658 Chr.12: 117049196-117049196 on Build GRCh38
rs746893667 Chr.12: 117049059-117049059 on Build GRCh38
rs757923846 Chr.12: 117049168-117049170 on Build GRCh38
rs562320074 Chr.12: 117048877-117048877 on Build GRCh38
rs775721872 Chr.12: 117049149-117049149 on Build GRCh38
rs912022078 Chr.12: 117049206-117049206 on Build GRCh38
rs1040734474 Chr.12: 117049063-117049063 on Build GRCh38
rs373277209 Chr.12: 117049125-117049125 on Build GRCh38
rs748927568 Chr.12: 117049141-117049141 on Build GRCh38
rs201966544 Chr.12: 117049151-117049151 on Build GRCh38
rs201990334 Chr.12: 117048999-117048999 on Build GRCh38
rs138273556 Chr.12: 117049073-117049073 on Build GRCh38
rs899479719 Chr.12: 117049064-117049064 on Build GRCh38
rs755703311 Chr.12: 117049237-117049237 on Build GRCh38
rs932356996 Chr.12: 117049242-117049242 on Build GRCh38
rs544696175 Chr.12: 117048933-117048933 on Build GRCh38
rs370907489 Chr.12: 117048975-117048975 on Build GRCh38
rs761040708 Chr.12: 117049150-117049150 on Build GRCh38
rs564647990 Chr.12: 117048952-117048952 on Build GRCh38
rs1033637419 Chr.12: 117049349-117049349 on Build GRCh38
rs751979955 Chr.12: 117048985-117048985 on Build GRCh38
rs944990557 Chr.12: 117049232-117049232 on Build GRCh38
rs1009708493 Chr.12: 117049288-117049288 on Build GRCh38
rs372587214 Chr.12: 117048909-117048910 on Build GRCh38
rs910164450 Chr.12: 117049331-117049331 on Build GRCh38
rs974878295 Chr.12: 117049416-117049416 on Build GRCh38
rs929531154 Chr.12: 117048951-117048951 on Build GRCh38
rs745836237 Chr.12: 117049281-117049281 on Build GRCh38
rs957943383 Chr.12: 117049235-117049235 on Build GRCh38
rs748003825 Chr.12: 117048866-117048866 on Build GRCh38
rs370536389 Chr.12: 117049025-117049025 on Build GRCh38
rs758146184 Chr.12: 117049038-117049038 on Build GRCh38
rs916382975 Chr.12: 117049268-117049268 on Build GRCh38
rs938089998 Chr.12: 117048864-117048864 on Build GRCh38
rs757473384 Chr.12: 117049015-117049015 on Build GRCh38
rs752650641 Chr.12: 117049204-117049204 on Build GRCh38
rs1053352794 Chr.12: 117049272-117049272 on Build GRCh38
rs189129803 Chr.12: 117049071-117049071 on Build GRCh38
rs1015625886 Chr.12: 117048909-117048909 on Build GRCh38
rs1042236800 Chr.12: 117049307-117049307 on Build GRCh38
rs536545057 Chr.12: 117049356-117049356 on Build GRCh38
rs765274811 Chr.12: 117049163-117049163 on Build GRCh38
rs550237149 Chr.12: 117049095-117049095 on Build GRCh38
rs776122247 Chr.12: 117049082-117049082 on Build GRCh38
rs1000190171 Chr.12: 117049391-117049392 on Build GRCh38
rs942928541 Chr.12: 117049332-117049332 on Build GRCh38
rs564612608 Chr.12: 117049386-117049386 on Build GRCh38
rs562814104 Chr.12: 117049408-117049408 on Build GRCh38
rs925126653 Chr.12: 117049056-117049056 on Build GRCh38
rs929664435 Chr.12: 117049319-117049319 on Build GRCh38
rs531450647 Chr.12: 117048902-117048902 on Build GRCh38
rs201789594 Chr.12: 117049093-117049093 on Build GRCh38
rs370277198 Chr.12: 117049086-117049086 on Build GRCh38
rs779877135 Chr.12: 117049177-117049177 on Build GRCh38
rs768161931 Chr.12: 117048996-117048996 on Build GRCh38

View Full Product Details