Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2669857-2670338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAFAH1B1
Entrez Gene ID
5048
Gene Name
platelet activating factor acetylhydrolase 1b regulatory subunit 1
Gene Aliases
LIS1, LIS2, MDCR, MDS, NudF, PAFAH
UniGene
Hs.77318
Gene Chromosome Location
Chr.17: 2593210-2685617 on Build GRCh38
Transcripts
NM_000430.3 XM_011523901.1 XM_011523902.2 XM_011523903.2 XM_017024699.1 XM_017024700.1 XM_017024701.1 XM_017024702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448606 c.567C>T p.H189H Chr.17: 2670330-2670330 on Build GRCh38
4450077 c.437delT p.G148fs*25 Chr.17: 2670200-2670200 on Build GRCh38
178379 c.431G>A p.R144Q Chr.17: 2670194-2670194 on Build GRCh38
4975807 c.568+1G>A p.? Chr.17: 2670332-2670332 on Build GRCh38
5628046 c.552C>T p.C184C Chr.17: 2670315-2670315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757060868 Chr.17: 2670255-2670255 on Build GRCh38
rs760293751 Chr.17: 2670023-2670023 on Build GRCh38
rs200390886 Chr.17: 2670266-2670266 on Build GRCh38
rs752624395 Chr.17: 2670301-2670301 on Build GRCh38
rs1034841577 Chr.17: 2669961-2669961 on Build GRCh38
rs1017404460 Chr.17: 2669977-2669977 on Build GRCh38
rs587784271 Chr.17: 2670300-2670300 on Build GRCh38
rs184753127 Chr.17: 2669894-2669894 on Build GRCh38
rs147529883 Chr.17: 2669969-2669969 on Build GRCh38
rs763770941 Chr.17: 2670104-2670104 on Build GRCh38
rs897149222 Chr.17: 2669909-2669909 on Build GRCh38
rs797045061 Chr.17: 2670286-2670286 on Build GRCh38
rs121434482 Chr.17: 2670209-2670209 on Build GRCh38
rs147910544 Chr.17: 2670264-2670264 on Build GRCh38
rs955933755 Chr.17: 2670061-2670061 on Build GRCh38
rs797045862 Chr.17: 2670300-2670301 on Build GRCh38
rs797045861 Chr.17: 2670204-2670205 on Build GRCh38
rs587784266 Chr.17: 2670168-2670168 on Build GRCh38
rs779220451 Chr.17: 2670163-2670163 on Build GRCh38
rs951994378 Chr.17: 2669957-2669957 on Build GRCh38
rs185820538 Chr.17: 2670119-2670119 on Build GRCh38
rs768998763 Chr.17: 2670216-2670216 on Build GRCh38
rs532943486 Chr.17: 2670246-2670246 on Build GRCh38
rs1003764054 Chr.17: 2669951-2669951 on Build GRCh38
rs988494557 Chr.17: 2670073-2670073 on Build GRCh38
rs201754847 Chr.17: 2670153-2670153 on Build GRCh38
rs773975872 Chr.17: 2670269-2670269 on Build GRCh38
rs749040257 Chr.17: 2670142-2670142 on Build GRCh38
rs746003762 Chr.17: 2670165-2670165 on Build GRCh38
rs927461308 Chr.17: 2670147-2670147 on Build GRCh38
rs370126179 Chr.17: 2670154-2670154 on Build GRCh38
rs1010530047 Chr.17: 2669923-2669924 on Build GRCh38
rs775905950 Chr.17: 2670180-2670180 on Build GRCh38
rs189993201 Chr.17: 2669929-2669929 on Build GRCh38
rs759035644 Chr.17: 2670279-2670279 on Build GRCh38
rs777600143 Chr.17: 2670137-2670137 on Build GRCh38
rs1057244318 Chr.17: 2670183-2670183 on Build GRCh38
rs947653493 Chr.17: 2670105-2670105 on Build GRCh38
rs181384339 Chr.17: 2670058-2670058 on Build GRCh38
rs368096054 Chr.17: 2669939-2669939 on Build GRCh38
rs534090290 Chr.17: 2669988-2669988 on Build GRCh38
rs753583241 Chr.17: 2670238-2670238 on Build GRCh38
rs587784269 Chr.17: 2670223-2670223 on Build GRCh38
rs754449962 Chr.17: 2670130-2670130 on Build GRCh38
rs755900926 Chr.17: 2670135-2670135 on Build GRCh38
rs772188120 Chr.17: 2670177-2670177 on Build GRCh38
rs11078302 Chr.17: 2669879-2669879 on Build GRCh38
rs1002969109 Chr.17: 2669888-2669888 on Build GRCh38
rs587784270 Chr.17: 2670287-2670291 on Build GRCh38
rs986163463 Chr.17: 2670095-2670095 on Build GRCh38
rs78897692 Chr.17: 2670042-2670042 on Build GRCh38
rs375418643 Chr.17: 2670148-2670148 on Build GRCh38
rs121434487 Chr.17: 2670247-2670247 on Build GRCh38
rs575432164 Chr.17: 2670145-2670145 on Build GRCh38
rs1026682996 Chr.17: 2669950-2669950 on Build GRCh38
rs562369565 Chr.17: 2670133-2670133 on Build GRCh38
rs544993391 Chr.17: 2670093-2670093 on Build GRCh38
rs997298510 Chr.17: 2670314-2670314 on Build GRCh38
rs897228864 Chr.17: 2670206-2670206 on Build GRCh38
rs201740734 Chr.17: 2670198-2670198 on Build GRCh38
rs767021387 Chr.17: 2670291-2670291 on Build GRCh38
rs964491570 Chr.17: 2670014-2670014 on Build GRCh38
rs587784267 Chr.17: 2670193-2670193 on Build GRCh38
rs587784268 Chr.17: 2670218-2670219 on Build GRCh38
rs761226380 Chr.17: 2670213-2670213 on Build GRCh38
rs121434484 Chr.17: 2670268-2670268 on Build GRCh38
rs201201710 Chr.17: 2670215-2670215 on Build GRCh38
rs116237011 Chr.17: 2670237-2670237 on Build GRCh38

View Full Product Details