Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4940080-4940656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF167
Entrez Gene ID
26001
Gene Name
ring finger protein 167
Gene Aliases
5730408C10Rik, LP2254, RING105
UniGene
Hs.7158
Gene Chromosome Location
Chr.17: 4940008-4945223 on Build GRCh38
Transcripts
NM_001320356.1 NM_001320357.1 NM_001320358.1 NM_001320359.1 NM_001320360.1 NM_001320361.1 NM_001320362.1 NM_001320363.1 NM_001320364.1 NM_001320365.1 NM_015528.2 XM_017024427.1 NM_001165417.1 NM_001165418.1 NM_003562.4

Gene Symbol
SLC25A11
Entrez Gene ID
8402
Gene Name
solute carrier family 25 member 11
Gene Aliases
OGC, SLC20A4
UniGene
Hs.184877
Gene Chromosome Location
Chr.17: 4937130-4940167 on Build GRCh38
Transcripts
NM_001320356.1 NM_001320357.1 NM_001320358.1 NM_001320359.1 NM_001320360.1 NM_001320361.1 NM_001320362.1 NM_001320363.1 NM_001320364.1 NM_001320365.1 NM_015528.2 XM_017024427.1 NM_001165417.1 NM_001165418.1 NM_003562.4

SNP Detail

SNP ID Chromosome Location
rs926817012 Chr.17: 4940246-4940246 on Build GRCh38
rs901258088 Chr.17: 4940317-4940317 on Build GRCh38
rs971759837 Chr.17: 4940158-4940158 on Build GRCh38
rs978597703 Chr.17: 4940486-4940486 on Build GRCh38
rs368570381 Chr.17: 4940386-4940386 on Build GRCh38
rs760100165 Chr.17: 4940511-4940511 on Build GRCh38
rs947167904 Chr.17: 4940249-4940249 on Build GRCh38
rs993126146 Chr.17: 4940410-4940410 on Build GRCh38
rs886676262 Chr.17: 4940569-4940569 on Build GRCh38
rs145971506 Chr.17: 4940271-4940271 on Build GRCh38
rs897462904 Chr.17: 4940393-4940393 on Build GRCh38
rs541012328 Chr.17: 4940456-4940456 on Build GRCh38
rs1041113858 Chr.17: 4940307-4940307 on Build GRCh38
rs533901700 Chr.17: 4940156-4940156 on Build GRCh38
rs1042735209 Chr.17: 4940104-4940104 on Build GRCh38
rs996179266 Chr.17: 4940627-4940627 on Build GRCh38
rs200349118 Chr.17: 4940270-4940271 on Build GRCh38
rs910357201 Chr.17: 4940549-4940549 on Build GRCh38
rs144016137 Chr.17: 4940187-4940187 on Build GRCh38
rs1033832648 Chr.17: 4940479-4940479 on Build GRCh38
rs965047764 Chr.17: 4940188-4940188 on Build GRCh38
rs954621498 Chr.17: 4940201-4940201 on Build GRCh38
rs1047893547 Chr.17: 4940345-4940345 on Build GRCh38
rs923221254 Chr.17: 4940197-4940199 on Build GRCh38
rs1037541098 Chr.17: 4940572-4940572 on Build GRCh38
rs913019068 Chr.17: 4940215-4940215 on Build GRCh38
rs1056803399 Chr.17: 4940298-4940298 on Build GRCh38
rs894901724 Chr.17: 4940140-4940152 on Build GRCh38
rs916878401 Chr.17: 4940509-4940509 on Build GRCh38
rs771042331 Chr.17: 4940491-4940491 on Build GRCh38
rs74989743 Chr.17: 4940489-4940489 on Build GRCh38
rs974697025 Chr.17: 4940192-4940192 on Build GRCh38
rs779913451 Chr.17: 4940150-4940150 on Build GRCh38
rs896720586 Chr.17: 4940152-4940152 on Build GRCh38
rs1030299558 Chr.17: 4940628-4940629 on Build GRCh38
rs756504541 Chr.17: 4940178-4940178 on Build GRCh38
rs1013842295 Chr.17: 4940320-4940320 on Build GRCh38
rs766885358 Chr.17: 4940592-4940592 on Build GRCh38
rs1056856238 Chr.17: 4940501-4940501 on Build GRCh38
rs563073401 Chr.17: 4940359-4940359 on Build GRCh38
rs139794857 Chr.17: 4940519-4940519 on Build GRCh38
rs528173283 Chr.17: 4940293-4940293 on Build GRCh38
rs985240588 Chr.17: 4940178-4940178 on Build GRCh38
rs570590546 Chr.17: 4940203-4940203 on Build GRCh38
rs932734378 Chr.17: 4940530-4940530 on Build GRCh38
rs937011846 Chr.17: 4940298-4940299 on Build GRCh38
rs80274549 Chr.17: 4940500-4940500 on Build GRCh38
rs534232688 Chr.17: 4940478-4940478 on Build GRCh38
rs898687288 Chr.17: 4940247-4940247 on Build GRCh38
rs371950478 Chr.17: 4940273-4940274 on Build GRCh38
rs976743119 Chr.17: 4940515-4940515 on Build GRCh38
rs889032001 Chr.17: 4940142-4940142 on Build GRCh38
rs913878216 Chr.17: 4940291-4940291 on Build GRCh38
rs568140341 Chr.17: 4940151-4940151 on Build GRCh38
rs1041166418 Chr.17: 4940125-4940125 on Build GRCh38
rs182820663 Chr.17: 4940451-4940451 on Build GRCh38
rs896774479 Chr.17: 4940301-4940301 on Build GRCh38
rs901645719 Chr.17: 4940129-4940129 on Build GRCh38
rs1006114998 Chr.17: 4940369-4940369 on Build GRCh38
rs1029282926 Chr.17: 4940135-4940135 on Build GRCh38
rs73973662 Chr.17: 4940114-4940114 on Build GRCh38
rs1024635912 Chr.17: 4940415-4940415 on Build GRCh38
rs900635012 Chr.17: 4940600-4940600 on Build GRCh38
rs988656855 Chr.17: 4940389-4940389 on Build GRCh38
rs990160945 Chr.17: 4940284-4940284 on Build GRCh38
rs945370779 Chr.17: 4940304-4940304 on Build GRCh38
rs747201584 Chr.17: 4940450-4940450 on Build GRCh38
rs1050259305 Chr.17: 4940348-4940348 on Build GRCh38
rs968930757 Chr.17: 4940484-4940484 on Build GRCh38
rs560523322 Chr.17: 4940609-4940609 on Build GRCh38
rs1050290231 Chr.17: 4940544-4940544 on Build GRCh38
rs970442460 Chr.17: 4940422-4940422 on Build GRCh38
rs941866289 Chr.17: 4940569-4940569 on Build GRCh38
rs556067469 Chr.17: 4940449-4940449 on Build GRCh38
rs564334131 Chr.17: 4940132-4940132 on Build GRCh38
rs576521431 Chr.17: 4940370-4940370 on Build GRCh38
rs994268371 Chr.17: 4940362-4940362 on Build GRCh38
rs746715790 Chr.17: 4940164-4940164 on Build GRCh38
rs1017055980 Chr.17: 4940244-4940244 on Build GRCh38
rs932764253 Chr.17: 4940337-4940337 on Build GRCh38

View Full Product Details