Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82419411-82420004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXDC
Entrez Gene ID
284004
Gene Name
hexosaminidase D
Gene Aliases
-
UniGene
Hs.744207
Gene Chromosome Location
Chr.17: 82418376-82442645 on Build GRCh38
Transcripts
NM_173620.2 XM_005256346.3 XM_005256347.4 XM_006722280.3 XM_017024484.1 XM_017024485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921164 c.35T>C p.V12A Chr.17: 82419834-82419834 on Build GRCh38
3523857 c.37C>T p.H13Y Chr.17: 82419836-82419836 on Build GRCh38
3756045 c.15T>G p.T5T Chr.17: 82419814-82419814 on Build GRCh38
1662129 c.52G>T p.G18* Chr.17: 82419851-82419851 on Build GRCh38
4839721 c.31T>A p.L11I Chr.17: 82419830-82419830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754585361 Chr.17: 82419932-82419932 on Build GRCh38
rs1030094107 Chr.17: 82419731-82419731 on Build GRCh38
rs763503533 Chr.17: 82419891-82419891 on Build GRCh38
rs560711599 Chr.17: 82419578-82419578 on Build GRCh38
rs770312378 Chr.17: 82419650-82419650 on Build GRCh38
rs532581327 Chr.17: 82419972-82419972 on Build GRCh38
rs985781491 Chr.17: 82419466-82419466 on Build GRCh38
rs773473528 Chr.17: 82419845-82419845 on Build GRCh38
rs924707007 Chr.17: 82419938-82419938 on Build GRCh38
rs904937051 Chr.17: 82419730-82419730 on Build GRCh38
rs997343165 Chr.17: 82419714-82419714 on Build GRCh38
rs771094745 Chr.17: 82419868-82419868 on Build GRCh38
rs755629914 Chr.17: 82419915-82419915 on Build GRCh38
rs1032034433 Chr.17: 82419794-82419794 on Build GRCh38
rs190114977 Chr.17: 82419811-82419811 on Build GRCh38
rs984601120 Chr.17: 82419961-82419961 on Build GRCh38
rs956507242 Chr.17: 82419963-82419964 on Build GRCh38
rs921341204 Chr.17: 82419511-82419511 on Build GRCh38
rs370204353 Chr.17: 82419537-82419537 on Build GRCh38
rs149948770 Chr.17: 82419877-82419877 on Build GRCh38
rs926725922 Chr.17: 82419902-82419902 on Build GRCh38
rs113708942 Chr.17: 82419679-82419679 on Build GRCh38
rs4074069 Chr.17: 82419530-82419530 on Build GRCh38
rs949552419 Chr.17: 82419588-82419588 on Build GRCh38
rs559242663 Chr.17: 82419513-82419513 on Build GRCh38
rs953197166 Chr.17: 82419461-82419461 on Build GRCh38
rs759975649 Chr.17: 82419755-82419755 on Build GRCh38
rs764609570 Chr.17: 82419904-82419904 on Build GRCh38
rs749916025 Chr.17: 82419788-82419788 on Build GRCh38
rs748186108 Chr.17: 82419817-82419817 on Build GRCh38
rs926673332 Chr.17: 82419643-82419643 on Build GRCh38
rs765449911 Chr.17: 82419766-82419766 on Build GRCh38
rs986874819 Chr.17: 82419923-82419923 on Build GRCh38
rs935163204 Chr.17: 82419587-82419587 on Build GRCh38
rs988001295 Chr.17: 82419965-82419965 on Build GRCh38
rs779716286 Chr.17: 82419800-82419800 on Build GRCh38
rs550183935 Chr.17: 82419870-82419870 on Build GRCh38
rs1017160831 Chr.17: 82419465-82419465 on Build GRCh38
rs778810861 Chr.17: 82419933-82419933 on Build GRCh38
rs547586773 Chr.17: 82419452-82419452 on Build GRCh38
rs540011463 Chr.17: 82419916-82419916 on Build GRCh38
rs150759806 Chr.17: 82419780-82419780 on Build GRCh38
rs988784789 Chr.17: 82419573-82419573 on Build GRCh38
rs753957586 Chr.17: 82419754-82419754 on Build GRCh38
rs1056631895 Chr.17: 82419813-82419813 on Build GRCh38
rs370631233 Chr.17: 82419871-82419871 on Build GRCh38
rs368300914 Chr.17: 82419887-82419887 on Build GRCh38
rs982195107 Chr.17: 82419601-82419601 on Build GRCh38
rs1052328697 Chr.17: 82419631-82419631 on Build GRCh38
rs753256682 Chr.17: 82419772-82419772 on Build GRCh38
rs117071519 Chr.17: 82419576-82419576 on Build GRCh38
rs1017129921 Chr.17: 82419875-82419875 on Build GRCh38
rs372614862 Chr.17: 82419912-82419912 on Build GRCh38
rs139212069 Chr.17: 82419835-82419835 on Build GRCh38
rs897703548 Chr.17: 82419683-82419683 on Build GRCh38
rs776257638 Chr.17: 82419459-82419459 on Build GRCh38
rs1048365888 Chr.17: 82419648-82419648 on Build GRCh38
rs975415889 Chr.17: 82419483-82419483 on Build GRCh38
rs941989503 Chr.17: 82419661-82419661 on Build GRCh38
rs1141463 Chr.17: 82419814-82419814 on Build GRCh38
rs747108453 Chr.17: 82419860-82419860 on Build GRCh38
rs915151915 Chr.17: 82419966-82419966 on Build GRCh38
rs755574839 Chr.17: 82419795-82419795 on Build GRCh38
rs1021155156 Chr.17: 82419852-82419852 on Build GRCh38
rs563977476 Chr.17: 82419951-82419951 on Build GRCh38
rs938010494 Chr.17: 82419645-82419645 on Build GRCh38

View Full Product Details