Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10293575-10294148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM5
Entrez Gene ID
7087
Gene Name
intercellular adhesion molecule 5
Gene Aliases
TLCN, TLN
UniGene
Hs.465862
Gene Chromosome Location
Chr.19: 10289352-10296778 on Build GRCh38
Transcripts
NM_003259.3 XM_011528229.1 XM_017027185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174548 c.1621C>T p.R541C Chr.19: 10293853-10293853 on Build GRCh38
4884794 c.1795G>T p.D599Y Chr.19: 10294123-10294123 on Build GRCh38
4884034 c.1672C>T p.R558W Chr.19: 10293904-10293904 on Build GRCh38
6683540 c.1697C>T p.A566V Chr.19: 10293929-10293929 on Build GRCh38
5981438 c.1579G>A p.V527M Chr.19: 10293811-10293811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769964078 Chr.19: 10294001-10294001 on Build GRCh38
rs10418913 Chr.19: 10293718-10293718 on Build GRCh38
rs368565932 Chr.19: 10294031-10294031 on Build GRCh38
rs915642535 Chr.19: 10293597-10293597 on Build GRCh38
rs765447145 Chr.19: 10293658-10293658 on Build GRCh38
rs971552940 Chr.19: 10294043-10294043 on Build GRCh38
rs376924939 Chr.19: 10294057-10294057 on Build GRCh38
rs760097857 Chr.19: 10293699-10293699 on Build GRCh38
rs747493164 Chr.19: 10293974-10293974 on Build GRCh38
rs1052667533 Chr.19: 10293740-10293740 on Build GRCh38
rs537423937 Chr.19: 10293845-10293845 on Build GRCh38
rs767567981 Chr.19: 10293712-10293712 on Build GRCh38
rs946717878 Chr.19: 10293965-10293965 on Build GRCh38
rs762029745 Chr.19: 10293888-10293888 on Build GRCh38
rs765026177 Chr.19: 10293835-10293835 on Build GRCh38
rs962376378 Chr.19: 10294018-10294018 on Build GRCh38
rs369467933 Chr.19: 10294010-10294010 on Build GRCh38
rs1006735132 Chr.19: 10294059-10294059 on Build GRCh38
rs147604357 Chr.19: 10293905-10293905 on Build GRCh38
rs781531482 Chr.19: 10294119-10294119 on Build GRCh38
rs201672754 Chr.19: 10293685-10293685 on Build GRCh38
rs772476365 Chr.19: 10293865-10293865 on Build GRCh38
rs933883880 Chr.19: 10293993-10293993 on Build GRCh38
rs1042270165 Chr.19: 10293670-10293670 on Build GRCh38
rs753531594 Chr.19: 10293955-10293955 on Build GRCh38
rs751902593 Chr.19: 10294098-10294098 on Build GRCh38
rs372704058 Chr.19: 10294044-10294044 on Build GRCh38
rs774890992 Chr.19: 10293692-10293692 on Build GRCh38
rs17852402 Chr.19: 10293843-10293843 on Build GRCh38
rs766440041 Chr.19: 10293841-10293841 on Build GRCh38
rs759445262 Chr.19: 10293930-10293930 on Build GRCh38
rs201577589 Chr.19: 10293829-10293829 on Build GRCh38
rs1038135744 Chr.19: 10293916-10293916 on Build GRCh38
rs779646880 Chr.19: 10293760-10293760 on Build GRCh38
rs780511464 Chr.19: 10293872-10293872 on Build GRCh38
rs747796224 Chr.19: 10293784-10293784 on Build GRCh38
rs748726167 Chr.19: 10294125-10294125 on Build GRCh38
rs771249059 Chr.19: 10293691-10293691 on Build GRCh38
rs752150067 Chr.19: 10293940-10293940 on Build GRCh38
rs753480140 Chr.19: 10294064-10294064 on Build GRCh38
rs755749166 Chr.19: 10293949-10293949 on Build GRCh38
rs766315764 Chr.19: 10293915-10293915 on Build GRCh38
rs745837260 Chr.19: 10293656-10293656 on Build GRCh38
rs762946340 Chr.19: 10293840-10293840 on Build GRCh38
rs542345526 Chr.19: 10293954-10293954 on Build GRCh38
rs776587690 Chr.19: 10293882-10293882 on Build GRCh38
rs374601160 Chr.19: 10294030-10294030 on Build GRCh38
rs750250084 Chr.19: 10293837-10293837 on Build GRCh38
rs148788922 Chr.19: 10294083-10294083 on Build GRCh38
rs367687699 Chr.19: 10294015-10294015 on Build GRCh38
rs763664362 Chr.19: 10294061-10294061 on Build GRCh38
rs374282567 Chr.19: 10293770-10293770 on Build GRCh38
rs777384734 Chr.19: 10293953-10293953 on Build GRCh38
rs771319048 Chr.19: 10293789-10293789 on Build GRCh38
rs768429598 Chr.19: 10293684-10293684 on Build GRCh38
rs199664255 Chr.19: 10293795-10293795 on Build GRCh38
rs991101459 Chr.19: 10294019-10294019 on Build GRCh38
rs765013341 Chr.19: 10293736-10293736 on Build GRCh38
rs766699029 Chr.19: 10293958-10293958 on Build GRCh38
rs1006643364 Chr.19: 10293723-10293723 on Build GRCh38
rs373149643 Chr.19: 10293674-10293674 on Build GRCh38
rs779466051 Chr.19: 10293662-10293663 on Build GRCh38
rs756254678 Chr.19: 10294128-10294128 on Build GRCh38
rs144234155 Chr.19: 10294074-10294074 on Build GRCh38
rs746261178 Chr.19: 10293860-10293860 on Build GRCh38
rs775502255 Chr.19: 10293821-10293821 on Build GRCh38
rs751538371 Chr.19: 10293917-10293917 on Build GRCh38
rs769942517 Chr.19: 10293902-10293902 on Build GRCh38
rs780458607 Chr.19: 10293969-10293969 on Build GRCh38
rs761999669 Chr.19: 10293834-10293834 on Build GRCh38
rs577573555 Chr.19: 10293873-10293873 on Build GRCh38
rs749684915 Chr.19: 10293687-10293687 on Build GRCh38
rs754060140 Chr.19: 10293734-10293734 on Build GRCh38
rs768771634 Chr.19: 10293875-10293875 on Build GRCh38
rs543369803 Chr.19: 10294102-10294102 on Build GRCh38
rs770835595 Chr.19: 10294023-10294023 on Build GRCh38
rs370603381 Chr.19: 10293717-10293717 on Build GRCh38
rs750361196 Chr.19: 10293746-10293746 on Build GRCh38
rs948250786 Chr.19: 10293634-10293634 on Build GRCh38
rs757049124 Chr.19: 10293735-10293735 on Build GRCh38
rs915331641 Chr.19: 10293948-10293948 on Build GRCh38
rs902542187 Chr.19: 10293992-10293992 on Build GRCh38
rs748201501 Chr.19: 10293991-10293991 on Build GRCh38
rs36045754 Chr.19: 10293936-10293936 on Build GRCh38
rs200251866 Chr.19: 10293776-10293776 on Build GRCh38
rs200631718 Chr.19: 10293978-10293978 on Build GRCh38
rs889617465 Chr.19: 10294026-10294026 on Build GRCh38
rs200192686 Chr.19: 10294103-10294103 on Build GRCh38
rs775013785 Chr.19: 10294056-10294056 on Build GRCh38
rs758425056 Chr.19: 10293752-10293752 on Build GRCh38
rs757977282 Chr.19: 10293661-10293661 on Build GRCh38
rs760725113 Chr.19: 10293828-10293828 on Build GRCh38
rs774297418 Chr.19: 10294036-10294036 on Build GRCh38
rs368250065 Chr.19: 10293794-10293794 on Build GRCh38
rs766664471 Chr.19: 10294077-10294077 on Build GRCh38
rs559003853 Chr.19: 10293859-10293859 on Build GRCh38
rs746546706 Chr.19: 10293765-10293765 on Build GRCh38
rs116688659 Chr.19: 10293912-10293912 on Build GRCh38
rs772570246 Chr.19: 10293807-10293807 on Build GRCh38
rs147074144 Chr.19: 10293854-10293854 on Build GRCh38
rs536342739 Chr.19: 10293805-10293805 on Build GRCh38
rs919493019 Chr.19: 10293817-10293817 on Build GRCh38
rs779769643 Chr.19: 10293662-10293662 on Build GRCh38
rs371136676 Chr.19: 10294017-10294017 on Build GRCh38
rs781293487 Chr.19: 10293981-10293981 on Build GRCh38

View Full Product Details