Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48716541-48717140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
UniGene
Hs.191815
Gene Chromosome Location
Chr.19: 48705718-48719898 on Build GRCh38
Transcripts
NM_001130915.1 NM_001297753.1 NM_182574.2 XM_011526807.2 XM_011526808.2 XM_011526809.2 XM_017026640.1 XM_017026641.1 XM_017026642.1 XM_017026643.1 XM_017026644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5511098 c.79A>C p.R27R Chr.19: 48716723-48716723 on Build GRCh38
6311498 c.64C>A p.Q22K Chr.19: 48716738-48716738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044333517 Chr.19: 48716858-48716858 on Build GRCh38
rs918110971 Chr.19: 48716592-48716592 on Build GRCh38
rs192544749 Chr.19: 48716597-48716597 on Build GRCh38
rs568296122 Chr.19: 48717044-48717044 on Build GRCh38
rs910238172 Chr.19: 48716760-48716760 on Build GRCh38
rs746874608 Chr.19: 48716770-48716770 on Build GRCh38
rs1020801445 Chr.19: 48716915-48716915 on Build GRCh38
rs377178039 Chr.19: 48716885-48716885 on Build GRCh38
rs188150257 Chr.19: 48716678-48716678 on Build GRCh38
rs779669262 Chr.19: 48716732-48716732 on Build GRCh38
rs979064522 Chr.19: 48716731-48716731 on Build GRCh38
rs1035347333 Chr.19: 48716895-48716895 on Build GRCh38
rs964366619 Chr.19: 48717005-48717005 on Build GRCh38
rs11672900 Chr.19: 48717066-48717066 on Build GRCh38
rs1040657930 Chr.19: 48716779-48716779 on Build GRCh38
rs967855411 Chr.19: 48716684-48716684 on Build GRCh38
rs1034780319 Chr.19: 48716587-48716587 on Build GRCh38
rs192702490 Chr.19: 48716789-48716789 on Build GRCh38
rs12611143 Chr.19: 48717113-48717113 on Build GRCh38
rs183885617 Chr.19: 48716846-48716846 on Build GRCh38
rs184932902 Chr.19: 48716598-48716598 on Build GRCh38
rs887830253 Chr.19: 48716843-48716843 on Build GRCh38
rs967926825 Chr.19: 48716853-48716853 on Build GRCh38
rs187458596 Chr.19: 48716588-48716588 on Build GRCh38
rs777929258 Chr.19: 48716666-48716666 on Build GRCh38
rs960681087 Chr.19: 48716903-48716903 on Build GRCh38
rs1002861841 Chr.19: 48716888-48716888 on Build GRCh38
rs969252139 Chr.19: 48716572-48716572 on Build GRCh38
rs909382635 Chr.19: 48717096-48717096 on Build GRCh38
rs549686146 Chr.19: 48717034-48717034 on Build GRCh38
rs909485307 Chr.19: 48716989-48716989 on Build GRCh38
rs910187255 Chr.19: 48716942-48716942 on Build GRCh38
rs745350527 Chr.19: 48716720-48716720 on Build GRCh38
rs372744870 Chr.19: 48716670-48716670 on Build GRCh38
rs968179769 Chr.19: 48716923-48716923 on Build GRCh38
rs1048490380 Chr.19: 48716662-48716662 on Build GRCh38
rs746455829 Chr.19: 48716618-48716618 on Build GRCh38
rs903672096 Chr.19: 48717076-48717076 on Build GRCh38
rs529192428 Chr.19: 48717058-48717058 on Build GRCh38
rs943563429 Chr.19: 48716766-48716766 on Build GRCh38
rs933331764 Chr.19: 48717021-48717021 on Build GRCh38
rs975769564 Chr.19: 48717006-48717006 on Build GRCh38
rs998038111 Chr.19: 48716719-48716719 on Build GRCh38
rs979179088 Chr.19: 48716933-48716933 on Build GRCh38
rs894843329 Chr.19: 48717114-48717114 on Build GRCh38
rs752398503 Chr.19: 48716836-48716836 on Build GRCh38
rs909431118 Chr.19: 48716664-48716665 on Build GRCh38
rs1009804922 Chr.19: 48716907-48716907 on Build GRCh38
rs370140427 Chr.19: 48716914-48716914 on Build GRCh38
rs921970325 Chr.19: 48717011-48717011 on Build GRCh38
rs531455108 Chr.19: 48716965-48716965 on Build GRCh38
rs565990285 Chr.19: 48717073-48717073 on Build GRCh38
rs1033371060 Chr.19: 48716884-48716884 on Build GRCh38
rs771759578 Chr.19: 48716727-48716727 on Build GRCh38
rs929483630 Chr.19: 48716628-48716628 on Build GRCh38
rs1002076372 Chr.19: 48716578-48716578 on Build GRCh38
rs929444867 Chr.19: 48716804-48716804 on Build GRCh38
rs959360970 Chr.19: 48716569-48716569 on Build GRCh38
rs539417399 Chr.19: 48717082-48717082 on Build GRCh38

View Full Product Details