Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55059170-55059452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RDH13
Entrez Gene ID
112724
Gene Name
retinol dehydrogenase 13 (all-trans/9-cis)
Gene Aliases
SDR7C3
UniGene
Hs.327631
Gene Chromosome Location
Chr.19: 55038895-55069774 on Build GRCh38
Transcripts
NM_001145971.1 NM_138412.3 NR_027381.1 NR_027382.1 XM_005258473.1 XM_011526407.1 XM_011526408.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458805 c.109C>T p.P37S Chr.19: 55059232-55059232 on Build GRCh38
7204793 c.132G>C p.T44T Chr.19: 55059209-55059209 on Build GRCh38
4798947 c.163G>T p.A55S Chr.19: 55059178-55059178 on Build GRCh38
1223461 c.136G>A p.A46T Chr.19: 55059205-55059205 on Build GRCh38
6003750 c.131C>T p.T44M Chr.19: 55059210-55059210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768563875 Chr.19: 55059304-55059304 on Build GRCh38
rs774342222 Chr.19: 55059315-55059315 on Build GRCh38
rs934058149 Chr.19: 55059280-55059280 on Build GRCh38
rs868746379 Chr.19: 55059415-55059415 on Build GRCh38
rs780441519 Chr.19: 55059199-55059199 on Build GRCh38
rs556307522 Chr.19: 55059215-55059215 on Build GRCh38
rs754364002 Chr.19: 55059263-55059263 on Build GRCh38
rs974455125 Chr.19: 55059338-55059341 on Build GRCh38
rs749691661 Chr.19: 55059200-55059200 on Build GRCh38
rs370160638 Chr.19: 55059196-55059196 on Build GRCh38
rs375991167 Chr.19: 55059282-55059282 on Build GRCh38
rs142936402 Chr.19: 55059247-55059247 on Build GRCh38
rs550029082 Chr.19: 55059397-55059397 on Build GRCh38
rs998441015 Chr.19: 55059198-55059198 on Build GRCh38
rs770407762 Chr.19: 55059209-55059209 on Build GRCh38
rs970034481 Chr.19: 55059270-55059270 on Build GRCh38
rs762070711 Chr.19: 55059317-55059317 on Build GRCh38
rs577167797 Chr.19: 55059393-55059393 on Build GRCh38
rs767619962 Chr.19: 55059233-55059233 on Build GRCh38
rs746377480 Chr.19: 55059191-55059191 on Build GRCh38
rs539892847 Chr.19: 55059335-55059335 on Build GRCh38
rs758447165 Chr.19: 55059358-55059358 on Build GRCh38
rs751761732 Chr.19: 55059223-55059223 on Build GRCh38
rs114180071 Chr.19: 55059357-55059357 on Build GRCh38
rs756612576 Chr.19: 55059262-55059262 on Build GRCh38
rs369685132 Chr.19: 55059221-55059221 on Build GRCh38
rs1022690889 Chr.19: 55059416-55059416 on Build GRCh38
rs200371141 Chr.19: 55059222-55059222 on Build GRCh38
rs745412780 Chr.19: 55059288-55059288 on Build GRCh38
rs781132193 Chr.19: 55059190-55059190 on Build GRCh38
rs762743328 Chr.19: 55059245-55059246 on Build GRCh38
rs755276891 Chr.19: 55059275-55059275 on Build GRCh38
rs759505965 Chr.19: 55059214-55059214 on Build GRCh38
rs970645047 Chr.19: 55059424-55059424 on Build GRCh38
rs376421701 Chr.19: 55059210-55059210 on Build GRCh38
rs907753165 Chr.19: 55059423-55059423 on Build GRCh38
rs769040588 Chr.19: 55059201-55059201 on Build GRCh38
rs966219598 Chr.19: 55059316-55059316 on Build GRCh38
rs1000627854 Chr.19: 55059274-55059274 on Build GRCh38
rs574362838 Chr.19: 55059327-55059327 on Build GRCh38
rs369999147 Chr.19: 55059291-55059291 on Build GRCh38
rs779254228 Chr.19: 55059277-55059277 on Build GRCh38
rs757484287 Chr.19: 55059231-55059231 on Build GRCh38
rs546133348 Chr.19: 55059398-55059398 on Build GRCh38
rs764100382 Chr.19: 55059315-55059316 on Build GRCh38
rs537923298 Chr.19: 55059295-55059295 on Build GRCh38
rs910107891 Chr.19: 55059246-55059246 on Build GRCh38
rs372582333 Chr.19: 55059296-55059296 on Build GRCh38
rs1671176 Chr.19: 55059414-55059414 on Build GRCh38
rs772801401 Chr.19: 55059205-55059205 on Build GRCh38
rs1654447 Chr.19: 55059206-55059206 on Build GRCh38
rs780732136 Chr.19: 55059284-55059284 on Build GRCh38

View Full Product Details