Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130854612-130855167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL1
Entrez Gene ID
25
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
UniGene
Hs.431048
Gene Chromosome Location
Chr.9: 130713881-130887675 on Build GRCh38
Transcripts
NM_005157.5 NM_007313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993192 c.307C>G p.Q103E Chr.9: 130854854-130854854 on Build GRCh38
7291892 c.562G>T p.G188W Chr.9: 130855052-130855052 on Build GRCh38
5977597 c.283delC p.H95fs*47 Chr.9: 130854830-130854830 on Build GRCh38
6591044 c.387G>A p.W129* Chr.9: 130854877-130854877 on Build GRCh38
1460512 c.454C>T p.R152C Chr.9: 130855001-130855001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143666455 Chr.9: 130855091-130855091 on Build GRCh38
rs772948197 Chr.9: 130854750-130854750 on Build GRCh38
rs587778016 Chr.9: 130854834-130854834 on Build GRCh38
rs750774244 Chr.9: 130855021-130855021 on Build GRCh38
rs781070135 Chr.9: 130854919-130854919 on Build GRCh38
rs774952482 Chr.9: 130854984-130854984 on Build GRCh38
rs757245158 Chr.9: 130855066-130855066 on Build GRCh38
rs770884302 Chr.9: 130854913-130854913 on Build GRCh38
rs1042889963 Chr.9: 130854914-130854914 on Build GRCh38
rs890170477 Chr.9: 130854995-130854995 on Build GRCh38
rs762158910 Chr.9: 130854994-130854994 on Build GRCh38
rs370776549 Chr.9: 130854814-130854814 on Build GRCh38
rs35095038 Chr.9: 130854751-130854751 on Build GRCh38
rs775748939 Chr.9: 130854956-130854956 on Build GRCh38
rs372671026 Chr.9: 130854774-130854774 on Build GRCh38
rs780251727 Chr.9: 130854789-130854792 on Build GRCh38
rs759794704 Chr.9: 130854779-130854779 on Build GRCh38
rs555918283 Chr.9: 130855122-130855122 on Build GRCh38
rs746875249 Chr.9: 130854910-130854910 on Build GRCh38
rs980417111 Chr.9: 130855141-130855141 on Build GRCh38
rs758308579 Chr.9: 130854892-130854892 on Build GRCh38
rs996137245 Chr.9: 130854968-130854968 on Build GRCh38
rs370897942 Chr.9: 130854838-130854838 on Build GRCh38
rs757304525 Chr.9: 130854796-130854796 on Build GRCh38
rs770132360 Chr.9: 130854955-130854955 on Build GRCh38
rs1002305388 Chr.9: 130854925-130854925 on Build GRCh38
rs764838466 Chr.9: 130855038-130855038 on Build GRCh38
rs777111369 Chr.9: 130855053-130855053 on Build GRCh38
rs767883505 Chr.9: 130855007-130855007 on Build GRCh38
rs368863447 Chr.9: 130855119-130855119 on Build GRCh38
rs375582718 Chr.9: 130854812-130854812 on Build GRCh38
rs770004598 Chr.9: 130854784-130854784 on Build GRCh38
rs941264264 Chr.9: 130854771-130854771 on Build GRCh38
rs1064152 Chr.9: 130854966-130854966 on Build GRCh38
rs146963680 Chr.9: 130855004-130855004 on Build GRCh38
rs201483141 Chr.9: 130855143-130855143 on Build GRCh38
rs987261935 Chr.9: 130854637-130854637 on Build GRCh38
rs748750300 Chr.9: 130855126-130855126 on Build GRCh38
rs781197426 Chr.9: 130855072-130855072 on Build GRCh38
rs867238359 Chr.9: 130854662-130854662 on Build GRCh38
rs778454302 Chr.9: 130854847-130854847 on Build GRCh38
rs368589654 Chr.9: 130854825-130854825 on Build GRCh38
rs757951694 Chr.9: 130855049-130855049 on Build GRCh38
rs752248660 Chr.9: 130854874-130854874 on Build GRCh38
rs61749011 Chr.9: 130854802-130854802 on Build GRCh38
rs199515879 Chr.9: 130854976-130854976 on Build GRCh38
rs961572795 Chr.9: 130854739-130854739 on Build GRCh38
rs373877460 Chr.9: 130854815-130854815 on Build GRCh38
rs1020988088 Chr.9: 130855039-130855039 on Build GRCh38
rs958337829 Chr.9: 130854973-130854973 on Build GRCh38
rs972865875 Chr.9: 130855116-130855116 on Build GRCh38
rs767993079 Chr.9: 130854656-130854656 on Build GRCh38
rs546918599 Chr.9: 130855063-130855063 on Build GRCh38
rs762122930 Chr.9: 130855110-130855110 on Build GRCh38
rs199551701 Chr.9: 130855107-130855107 on Build GRCh38
rs775695843 Chr.9: 130854808-130854808 on Build GRCh38
rs185209149 Chr.9: 130855138-130855138 on Build GRCh38
rs141072111 Chr.9: 130854898-130854898 on Build GRCh38
rs770185520 Chr.9: 130855105-130855105 on Build GRCh38
rs759251749 Chr.9: 130855034-130855034 on Build GRCh38
rs746147456 Chr.9: 130854937-130854937 on Build GRCh38
rs1019790995 Chr.9: 130854667-130854667 on Build GRCh38
rs11788639 Chr.9: 130854786-130854786 on Build GRCh38
rs535980798 Chr.9: 130855120-130855120 on Build GRCh38
rs148161029 Chr.9: 130855048-130855048 on Build GRCh38
rs764626784 Chr.9: 130854813-130854813 on Build GRCh38
rs1064153 Chr.9: 130855022-130855022 on Build GRCh38
rs753490709 Chr.9: 130854829-130854829 on Build GRCh38
rs1049690307 Chr.9: 130854693-130854693 on Build GRCh38

View Full Product Details