Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197817004-197817574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH3
Entrez Gene ID
84859
Gene Name
leucine rich repeats and calponin homology domain containing 3
Gene Aliases
-
UniGene
Hs.518414
Gene Chromosome Location
Chr.3: 197790855-197889346 on Build GRCh38
Transcripts
NM_032773.3 XM_005269362.2 XM_005269363.2 XM_005269365.2 XM_005269367.2 XM_006713791.3 XM_006713792.3 XM_006713793.2 XM_011513242.2 XM_011513243.2 XM_017007350.1 XM_017007351.1 XM_017007352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6702784 c.409C>T p.R137W Chr.3: 197817177-197817177 on Build GRCh38
6702787 c.518G>T p.R173I Chr.3: 197817286-197817286 on Build GRCh38
7098386 c.525G>C p.L175F Chr.3: 197817293-197817293 on Build GRCh38
6290672 c.462C>T p.V154V Chr.3: 197817230-197817230 on Build GRCh38
2948022 c.521A>G p.H174R Chr.3: 197817289-197817289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892390673 Chr.3: 197817177-197817177 on Build GRCh38
rs895636720 Chr.3: 197817083-197817083 on Build GRCh38
rs750327815 Chr.3: 197817074-197817074 on Build GRCh38
rs750322668 Chr.3: 197817339-197817340 on Build GRCh38
rs746387277 Chr.3: 197817200-197817200 on Build GRCh38
rs747167526 Chr.3: 197817221-197817221 on Build GRCh38
rs527947480 Chr.3: 197817121-197817121 on Build GRCh38
rs766297519 Chr.3: 197817352-197817353 on Build GRCh38
rs112378706 Chr.3: 197817039-197817039 on Build GRCh38
rs753672053 Chr.3: 197817138-197817138 on Build GRCh38
rs1052923976 Chr.3: 197817056-197817056 on Build GRCh38
rs58866690 Chr.3: 197817379-197817380 on Build GRCh38
rs796077481 Chr.3: 197817345-197817346 on Build GRCh38
rs548378972 Chr.3: 197817366-197817367 on Build GRCh38
rs547407719 Chr.3: 197817368-197817368 on Build GRCh38
rs779678253 Chr.3: 197817330-197817331 on Build GRCh38
rs868180027 Chr.3: 197817255-197817255 on Build GRCh38
rs929564566 Chr.3: 197817452-197817452 on Build GRCh38
rs759478091 Chr.3: 197817164-197817165 on Build GRCh38
rs749724892 Chr.3: 197817194-197817194 on Build GRCh38
rs367618377 Chr.3: 197817340-197817341 on Build GRCh38
rs1017773786 Chr.3: 197817418-197817418 on Build GRCh38
rs375750226 Chr.3: 197817360-197817360 on Build GRCh38
rs761726716 Chr.3: 197817142-197817142 on Build GRCh38
rs55896070 Chr.3: 197817329-197817330 on Build GRCh38
rs775668460 Chr.3: 197817219-197817219 on Build GRCh38
rs878951925 Chr.3: 197817054-197817054 on Build GRCh38
rs756117513 Chr.3: 197817165-197817165 on Build GRCh38
rs539752975 Chr.3: 197817489-197817489 on Build GRCh38
rs774948248 Chr.3: 197817151-197817154 on Build GRCh38
rs533289240 Chr.3: 197817154-197817154 on Build GRCh38
rs139714175 Chr.3: 197817338-197817339 on Build GRCh38
rs767642889 Chr.3: 197817266-197817268 on Build GRCh38
rs905254944 Chr.3: 197817129-197817129 on Build GRCh38
rs371061888 Chr.3: 197817353-197817353 on Build GRCh38
rs751135158 Chr.3: 197817178-197817178 on Build GRCh38
rs368027052 Chr.3: 197817185-197817185 on Build GRCh38
rs775688797 Chr.3: 197817334-197817335 on Build GRCh38
rs573533559 Chr.3: 197817106-197817106 on Build GRCh38
rs150956786 Chr.3: 197817211-197817211 on Build GRCh38
rs779975279 Chr.3: 197817322-197817322 on Build GRCh38
rs551929841 Chr.3: 197817346-197817347 on Build GRCh38
rs143325022 Chr.3: 197817387-197817387 on Build GRCh38
rs559654313 Chr.3: 197817136-197817136 on Build GRCh38
rs764924938 Chr.3: 197817349-197817350 on Build GRCh38
rs745831520 Chr.3: 197817330-197817349 on Build GRCh38
rs754421014 Chr.3: 197817354-197817355 on Build GRCh38
rs201576282 Chr.3: 197817362-197817362 on Build GRCh38
rs941068741 Chr.3: 197817518-197817518 on Build GRCh38
rs760159655 Chr.3: 197817333-197817334 on Build GRCh38
rs910272003 Chr.3: 197817037-197817037 on Build GRCh38
rs879374759 Chr.3: 197817539-197817539 on Build GRCh38
rs757635482 Chr.3: 197817277-197817277 on Build GRCh38
rs748393430 Chr.3: 197817349-197817349 on Build GRCh38
rs752398902 Chr.3: 197817274-197817274 on Build GRCh38
rs375478596 Chr.3: 197817244-197817244 on Build GRCh38
rs767341764 Chr.3: 197817260-197817260 on Build GRCh38
rs369081077 Chr.3: 197817344-197817344 on Build GRCh38
rs762765719 Chr.3: 197817351-197817352 on Build GRCh38
rs752861453 Chr.3: 197817327-197817328 on Build GRCh38
rs934569725 Chr.3: 197817051-197817051 on Build GRCh38
rs55705919 Chr.3: 197817331-197817332 on Build GRCh38
rs200477482 Chr.3: 197817346-197817346 on Build GRCh38
rs757830121 Chr.3: 197817196-197817196 on Build GRCh38
rs747806070 Chr.3: 197817345-197817346 on Build GRCh38
rs987611549 Chr.3: 197817036-197817036 on Build GRCh38
rs764372892 Chr.3: 197817336-197817337 on Build GRCh38
rs879222040 Chr.3: 197817364-197817364 on Build GRCh38
rs939153525 Chr.3: 197817552-197817552 on Build GRCh38
rs987632477 Chr.3: 197817400-197817400 on Build GRCh38
rs55955867 Chr.3: 197817361-197817362 on Build GRCh38
rs558068562 Chr.3: 197817505-197817505 on Build GRCh38
rs762982358 Chr.3: 197817237-197817237 on Build GRCh38
rs1046931217 Chr.3: 197817122-197817122 on Build GRCh38
rs758984138 Chr.3: 197817305-197817305 on Build GRCh38
rs545179844 Chr.3: 197817192-197817192 on Build GRCh38
rs756463093 Chr.3: 197817328-197817329 on Build GRCh38
rs372167489 Chr.3: 197817341-197817342 on Build GRCh38
rs765777629 Chr.3: 197817240-197817240 on Build GRCh38
rs768622993 Chr.3: 197817344-197817344 on Build GRCh38
rs796089573 Chr.3: 197817344-197817348 on Build GRCh38
rs765225697 Chr.3: 197817146-197817146 on Build GRCh38
rs750366399 Chr.3: 197817151-197817151 on Build GRCh38
rs760950980 Chr.3: 197817335-197817336 on Build GRCh38
rs1044847155 Chr.3: 197817088-197817088 on Build GRCh38
rs779493256 Chr.3: 197817199-197817199 on Build GRCh38
rs747690865 Chr.3: 197817360-197817361 on Build GRCh38
rs779440004 Chr.3: 197817279-197817279 on Build GRCh38
rs376394506 Chr.3: 197817342-197817342 on Build GRCh38
rs765031585 Chr.3: 197817228-197817228 on Build GRCh38
rs556661678 Chr.3: 197817535-197817535 on Build GRCh38
rs754512268 Chr.3: 197817342-197817343 on Build GRCh38
rs919483471 Chr.3: 197817388-197817389 on Build GRCh38
rs373639538 Chr.3: 197817358-197817358 on Build GRCh38
rs1002621377 Chr.3: 197817132-197817132 on Build GRCh38
rs769541383 Chr.3: 197817351-197817351 on Build GRCh38
rs555482985 Chr.3: 197817105-197817105 on Build GRCh38
rs199672959 Chr.3: 197817226-197817226 on Build GRCh38
rs748786868 Chr.3: 197817370-197817371 on Build GRCh38
rs768971597 Chr.3: 197817222-197817222 on Build GRCh38
rs773086327 Chr.3: 197817230-197817230 on Build GRCh38
rs768868671 Chr.3: 197817347-197817348 on Build GRCh38
rs978412171 Chr.3: 197817040-197817040 on Build GRCh38
rs565985700 Chr.3: 197817370-197817370 on Build GRCh38
rs746335994 Chr.3: 197817289-197817289 on Build GRCh38
rs754134261 Chr.3: 197817337-197817338 on Build GRCh38
rs888180546 Chr.3: 197817058-197817059 on Build GRCh38
rs1273552 Chr.3: 197817330-197817330 on Build GRCh38
rs754577411 Chr.3: 197817249-197817249 on Build GRCh38
rs985086017 Chr.3: 197817491-197817491 on Build GRCh38
rs978868593 Chr.3: 197817506-197817506 on Build GRCh38
rs925879473 Chr.3: 197817042-197817042 on Build GRCh38
rs189900093 Chr.3: 197817538-197817538 on Build GRCh38

View Full Product Details