Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87261193-87261713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU1F1
Entrez Gene ID
5449
Gene Name
POU class 1 homeobox 1
Gene Aliases
CPHD1, GHF-1, PIT1, POU1F1a, Pit-1
UniGene
Hs.591654
Gene Chromosome Location
Chr.3: 87259633-87276587 on Build GRCh38
Transcripts
NM_000306.3 NM_001122757.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1425581 c.665+2T>C p.? Chr.3: 87261271-87261271 on Build GRCh38
1425580 c.743+2T>C p.? Chr.3: 87261271-87261271 on Build GRCh38
1594124 c.728G>A p.R243Q Chr.3: 87261288-87261288 on Build GRCh38
137857 c.650G>A p.R217Q Chr.3: 87261288-87261288 on Build GRCh38
2828068 c.722G>T p.R241I Chr.3: 87261294-87261294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542209531 Chr.3: 87261468-87261468 on Build GRCh38
rs950061283 Chr.3: 87261408-87261408 on Build GRCh38
rs934831031 Chr.3: 87261563-87261563 on Build GRCh38
rs535867874 Chr.3: 87261602-87261605 on Build GRCh38
rs575910906 Chr.3: 87261249-87261249 on Build GRCh38
rs760045491 Chr.3: 87261327-87261327 on Build GRCh38
rs1026047555 Chr.3: 87261463-87261463 on Build GRCh38
rs515726221 Chr.3: 87261272-87261272 on Build GRCh38
rs777030970 Chr.3: 87261323-87261323 on Build GRCh38
rs1026565959 Chr.3: 87261629-87261629 on Build GRCh38
rs188183412 Chr.3: 87261509-87261509 on Build GRCh38
rs768261627 Chr.3: 87261250-87261250 on Build GRCh38
rs371899469 Chr.3: 87261342-87261342 on Build GRCh38
rs775878145 Chr.3: 87261304-87261304 on Build GRCh38
rs970517323 Chr.3: 87261284-87261284 on Build GRCh38
rs954589943 Chr.3: 87261471-87261471 on Build GRCh38
rs1002040266 Chr.3: 87261440-87261440 on Build GRCh38
rs945023910 Chr.3: 87261469-87261469 on Build GRCh38
rs769407182 Chr.3: 87261443-87261443 on Build GRCh38
rs1053201870 Chr.3: 87261390-87261390 on Build GRCh38
rs777263442 Chr.3: 87261360-87261365 on Build GRCh38
rs772390221 Chr.3: 87261296-87261300 on Build GRCh38
rs572547745 Chr.3: 87261608-87261608 on Build GRCh38
rs747045488 Chr.3: 87261347-87261348 on Build GRCh38
rs927255535 Chr.3: 87261624-87261624 on Build GRCh38
rs201406891 Chr.3: 87261338-87261338 on Build GRCh38
rs995038159 Chr.3: 87261557-87261557 on Build GRCh38
rs766603627 Chr.3: 87261295-87261295 on Build GRCh38
rs756693012 Chr.3: 87261445-87261445 on Build GRCh38
rs139649367 Chr.3: 87261406-87261406 on Build GRCh38
rs993683243 Chr.3: 87261457-87261457 on Build GRCh38
rs752983390 Chr.3: 87261340-87261340 on Build GRCh38
rs1044350722 Chr.3: 87261419-87261419 on Build GRCh38
rs978933736 Chr.3: 87261335-87261335 on Build GRCh38
rs905130774 Chr.3: 87261420-87261420 on Build GRCh38
rs745825368 Chr.3: 87261237-87261237 on Build GRCh38
rs886715980 Chr.3: 87261661-87261661 on Build GRCh38
rs113615664 Chr.3: 87261474-87261474 on Build GRCh38
rs764268838 Chr.3: 87261365-87261365 on Build GRCh38
rs1017247562 Chr.3: 87261493-87261493 on Build GRCh38
rs898992649 Chr.3: 87261449-87261449 on Build GRCh38
rs773523196 Chr.3: 87261287-87261287 on Build GRCh38
rs762450166 Chr.3: 87261377-87261378 on Build GRCh38
rs899296061 Chr.3: 87261529-87261529 on Build GRCh38
rs769438429 Chr.3: 87261240-87261240 on Build GRCh38
rs546396818 Chr.3: 87261651-87261651 on Build GRCh38
rs937966584 Chr.3: 87261663-87261663 on Build GRCh38
rs750282021 Chr.3: 87261373-87261373 on Build GRCh38
rs368358102 Chr.3: 87261612-87261612 on Build GRCh38
rs768862427 Chr.3: 87261349-87261351 on Build GRCh38
rs560344199 Chr.3: 87261508-87261508 on Build GRCh38
rs936514360 Chr.3: 87261401-87261401 on Build GRCh38
rs761275346 Chr.3: 87261289-87261289 on Build GRCh38
rs374853152 Chr.3: 87261288-87261288 on Build GRCh38
rs536984508 Chr.3: 87261254-87261254 on Build GRCh38
rs926143330 Chr.3: 87261561-87261561 on Build GRCh38
rs886037826 Chr.3: 87261300-87261304 on Build GRCh38
rs547129320 Chr.3: 87261481-87261481 on Build GRCh38
rs989324347 Chr.3: 87261595-87261595 on Build GRCh38
rs779925350 Chr.3: 87261247-87261247 on Build GRCh38
rs943091840 Chr.3: 87261482-87261482 on Build GRCh38
rs917356541 Chr.3: 87261607-87261607 on Build GRCh38
rs771581828 Chr.3: 87261277-87261277 on Build GRCh38
rs964961649 Chr.3: 87261500-87261500 on Build GRCh38
rs757365762 Chr.3: 87261264-87261264 on Build GRCh38

View Full Product Details