Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 170068500-170068783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
UniGene
Hs.529735
Gene Chromosome Location
Chr.4: 170060222-170091699 on Build GRCh38
Transcripts
NM_001286682.1 NM_001286683.1 NM_016228.3 NM_182662.1 XM_006714231.2 XM_011532020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419707 c.892T>C p.F298L Chr.4: 170068599-170068599 on Build GRCh38
5918326 c.898C>T p.Q300* Chr.4: 170068593-170068593 on Build GRCh38
5948609 c.868G>T p.V290F Chr.4: 170068623-170068623 on Build GRCh38
733119 c.860A>T p.H287L Chr.4: 170068631-170068631 on Build GRCh38
1181147 c.834A>C p.K278N Chr.4: 170068657-170068657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751698301 Chr.4: 170068730-170068730 on Build GRCh38
rs780401971 Chr.4: 170068595-170068595 on Build GRCh38
rs753739719 Chr.4: 170068565-170068566 on Build GRCh38
rs762489440 Chr.4: 170068632-170068632 on Build GRCh38
rs151231482 Chr.4: 170068641-170068641 on Build GRCh38
rs3214945 Chr.4: 170068557-170068558 on Build GRCh38
rs114038917 Chr.4: 170068710-170068710 on Build GRCh38
rs922205561 Chr.4: 170068569-170068569 on Build GRCh38
rs773871959 Chr.4: 170068644-170068644 on Build GRCh38
rs112229990 Chr.4: 170068606-170068606 on Build GRCh38
rs111447506 Chr.4: 170068700-170068700 on Build GRCh38
rs773656274 Chr.4: 170068627-170068627 on Build GRCh38
rs370431862 Chr.4: 170068615-170068615 on Build GRCh38
rs770317871 Chr.4: 170068624-170068624 on Build GRCh38
rs397750190 Chr.4: 170068567-170068568 on Build GRCh38
rs1030363229 Chr.4: 170068633-170068633 on Build GRCh38
rs757032802 Chr.4: 170068641-170068644 on Build GRCh38
rs374550256 Chr.4: 170068597-170068597 on Build GRCh38
rs375944075 Chr.4: 170068692-170068692 on Build GRCh38
rs759149170 Chr.4: 170068657-170068657 on Build GRCh38
rs1011800568 Chr.4: 170068539-170068539 on Build GRCh38
rs770474107 Chr.4: 170068637-170068637 on Build GRCh38
rs777630366 Chr.4: 170068570-170068570 on Build GRCh38
rs397704645 Chr.4: 170068566-170068567 on Build GRCh38
rs748477745 Chr.4: 170068619-170068619 on Build GRCh38
rs908047481 Chr.4: 170068695-170068695 on Build GRCh38
rs780244518 Chr.4: 170068712-170068712 on Build GRCh38
rs751766775 Chr.4: 170068558-170068558 on Build GRCh38
rs777732766 Chr.4: 170068572-170068572 on Build GRCh38
rs1020558687 Chr.4: 170068540-170068540 on Build GRCh38
rs767159360 Chr.4: 170068688-170068688 on Build GRCh38
rs781637765 Chr.4: 170068610-170068610 on Build GRCh38
rs372275480 Chr.4: 170068711-170068711 on Build GRCh38
rs777812492 Chr.4: 170068564-170068565 on Build GRCh38
rs371382324 Chr.4: 170068691-170068691 on Build GRCh38
rs758448288 Chr.4: 170068586-170068586 on Build GRCh38
rs750513609 Chr.4: 170068584-170068584 on Build GRCh38
rs755196147 Chr.4: 170068735-170068735 on Build GRCh38
rs756011377 Chr.4: 170068550-170068550 on Build GRCh38
rs752465113 Chr.4: 170068548-170068548 on Build GRCh38
rs756012907 Chr.4: 170068562-170068563 on Build GRCh38

View Full Product Details