Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26043380-26043916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2BB
Entrez Gene ID
3018
Gene Name
histone cluster 1, H2bb
Gene Aliases
H2B.1, H2B/f, H2BFF
UniGene
Hs.553494
Gene Chromosome Location
Chr.6: 26043227-26043657 on Build GRCh38
Transcripts
NM_021062.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6264749 c.90T>C p.R30R Chr.6: 26043568-26043568 on Build GRCh38
2857622 c.270C>A p.I90I Chr.6: 26043388-26043388 on Build GRCh38
6279345 c.95G>A p.R32H Chr.6: 26043563-26043563 on Build GRCh38
2857624 c.254A>C p.N85T Chr.6: 26043404-26043404 on Build GRCh38
6331992 c.264G>T p.S88S Chr.6: 26043394-26043394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774251549 Chr.6: 26043508-26043508 on Build GRCh38
rs1014408471 Chr.6: 26043838-26043838 on Build GRCh38
rs560518508 Chr.6: 26043818-26043818 on Build GRCh38
rs762266912 Chr.6: 26043802-26043802 on Build GRCh38
rs572599234 Chr.6: 26043684-26043684 on Build GRCh38
rs751683070 Chr.6: 26043655-26043655 on Build GRCh38
rs186144617 Chr.6: 26043663-26043663 on Build GRCh38
rs540610695 Chr.6: 26043797-26043797 on Build GRCh38
rs778158634 Chr.6: 26043548-26043548 on Build GRCh38
rs1031164937 Chr.6: 26043717-26043717 on Build GRCh38
rs776112468 Chr.6: 26043419-26043419 on Build GRCh38
rs955872994 Chr.6: 26043779-26043779 on Build GRCh38
rs768195915 Chr.6: 26043502-26043502 on Build GRCh38
rs371953695 Chr.6: 26043692-26043692 on Build GRCh38
rs972551538 Chr.6: 26043670-26043670 on Build GRCh38
rs765585164 Chr.6: 26043594-26043594 on Build GRCh38
rs139386667 Chr.6: 26043648-26043648 on Build GRCh38
rs747681265 Chr.6: 26043631-26043631 on Build GRCh38
rs909287289 Chr.6: 26043466-26043466 on Build GRCh38
rs764130486 Chr.6: 26043653-26043653 on Build GRCh38
rs971946993 Chr.6: 26043737-26043737 on Build GRCh38
rs144272993 Chr.6: 26043450-26043450 on Build GRCh38
rs768320945 Chr.6: 26043562-26043562 on Build GRCh38
rs780141769 Chr.6: 26043460-26043460 on Build GRCh38
rs894754724 Chr.6: 26043577-26043577 on Build GRCh38
rs746263633 Chr.6: 26043613-26043613 on Build GRCh38
rs764833010 Chr.6: 26043437-26043437 on Build GRCh38
rs903372916 Chr.6: 26043586-26043586 on Build GRCh38
rs775762574 Chr.6: 26043678-26043678 on Build GRCh38
rs752769182 Chr.6: 26043595-26043595 on Build GRCh38
rs1014522036 Chr.6: 26043574-26043574 on Build GRCh38
rs747341311 Chr.6: 26043487-26043487 on Build GRCh38
rs750994469 Chr.6: 26043685-26043685 on Build GRCh38
rs1028586777 Chr.6: 26043723-26043723 on Build GRCh38
rs12663349 Chr.6: 26043524-26043524 on Build GRCh38
rs778548350 Chr.6: 26043629-26043629 on Build GRCh38
rs779674665 Chr.6: 26043703-26043703 on Build GRCh38
rs1052949437 Chr.6: 26043420-26043420 on Build GRCh38
rs183493414 Chr.6: 26043848-26043848 on Build GRCh38
rs149617319 Chr.6: 26043755-26043755 on Build GRCh38
rs367562049 Chr.6: 26043687-26043687 on Build GRCh38
rs1029450521 Chr.6: 26043881-26043881 on Build GRCh38
rs756539532 Chr.6: 26043592-26043593 on Build GRCh38
rs372198960 Chr.6: 26043644-26043644 on Build GRCh38
rs575246050 Chr.6: 26043790-26043790 on Build GRCh38
rs750977180 Chr.6: 26043827-26043827 on Build GRCh38
rs1049647748 Chr.6: 26043454-26043454 on Build GRCh38
rs760932354 Chr.6: 26043708-26043708 on Build GRCh38
rs762465445 Chr.6: 26043442-26043442 on Build GRCh38
rs941263873 Chr.6: 26043814-26043814 on Build GRCh38
rs558377112 Chr.6: 26043720-26043720 on Build GRCh38
rs754556609 Chr.6: 26043628-26043628 on Build GRCh38
rs767395554 Chr.6: 26043455-26043455 on Build GRCh38
rs760399920 Chr.6: 26043521-26043521 on Build GRCh38
rs776966855 Chr.6: 26043589-26043589 on Build GRCh38
rs997577379 Chr.6: 26043722-26043722 on Build GRCh38
rs747391525 Chr.6: 26043550-26043550 on Build GRCh38
rs557191032 Chr.6: 26043649-26043649 on Build GRCh38
rs1010298248 Chr.6: 26043870-26043870 on Build GRCh38
rs915877682 Chr.6: 26043760-26043760 on Build GRCh38
rs772628669 Chr.6: 26043414-26043414 on Build GRCh38
rs761315898 Chr.6: 26043570-26043570 on Build GRCh38
rs775067316 Chr.6: 26043496-26043496 on Build GRCh38
rs999066193 Chr.6: 26043710-26043710 on Build GRCh38
rs953067702 Chr.6: 26043726-26043726 on Build GRCh38
rs757666983 Chr.6: 26043656-26043656 on Build GRCh38
rs999641372 Chr.6: 26043633-26043633 on Build GRCh38
rs775010729 Chr.6: 26043405-26043405 on Build GRCh38
rs767811340 Chr.6: 26043544-26043545 on Build GRCh38
rs763036603 Chr.6: 26043680-26043680 on Build GRCh38
rs532643029 Chr.6: 26043830-26043830 on Build GRCh38
rs774690751 Chr.6: 26043707-26043707 on Build GRCh38
rs535828942 Chr.6: 26043672-26043672 on Build GRCh38
rs903474190 Chr.6: 26043804-26043804 on Build GRCh38
rs775001760 Chr.6: 26043441-26043441 on Build GRCh38
rs779554627 Chr.6: 26043472-26043472 on Build GRCh38
rs756896467 Chr.6: 26043697-26043697 on Build GRCh38
rs566958517 Chr.6: 26043459-26043459 on Build GRCh38
rs774770463 Chr.6: 26043498-26043498 on Build GRCh38
rs766610233 Chr.6: 26043523-26043523 on Build GRCh38
rs575748263 Chr.6: 26043736-26043736 on Build GRCh38
rs1044549641 Chr.6: 26043845-26043845 on Build GRCh38
rs201616602 Chr.6: 26043527-26043527 on Build GRCh38
rs530695650 Chr.6: 26043863-26043863 on Build GRCh38
rs769755321 Chr.6: 26043676-26043676 on Build GRCh38
rs558872598 Chr.6: 26043875-26043875 on Build GRCh38
rs776603317 Chr.6: 26043852-26043852 on Build GRCh38
rs374021869 Chr.6: 26043694-26043694 on Build GRCh38
rs755073705 Chr.6: 26043538-26043538 on Build GRCh38
rs151259814 Chr.6: 26043603-26043603 on Build GRCh38
rs997251205 Chr.6: 26043821-26043821 on Build GRCh38
rs759780461 Chr.6: 26043593-26043593 on Build GRCh38
rs749291945 Chr.6: 26043660-26043660 on Build GRCh38
rs1056448275 Chr.6: 26043547-26043547 on Build GRCh38
rs768018204 Chr.6: 26043657-26043657 on Build GRCh38
rs748700936 Chr.6: 26043475-26043475 on Build GRCh38
rs1041243894 Chr.6: 26043811-26043811 on Build GRCh38
rs970275523 Chr.6: 26043744-26043744 on Build GRCh38
rs748310697 Chr.6: 26043630-26043630 on Build GRCh38
rs779650601 Chr.6: 26043558-26043558 on Build GRCh38
rs960461004 Chr.6: 26043556-26043556 on Build GRCh38
rs1016117306 Chr.6: 26043869-26043869 on Build GRCh38
rs778146944 Chr.6: 26043477-26043477 on Build GRCh38
rs758876516 Chr.6: 26043545-26043545 on Build GRCh38
rs769691103 Chr.6: 26043870-26043870 on Build GRCh38
rs550535211 Chr.6: 26043892-26043892 on Build GRCh38
rs199902704 Chr.6: 26043407-26043407 on Build GRCh38
rs781490038 Chr.6: 26043611-26043611 on Build GRCh38
rs750164760 Chr.6: 26043517-26043517 on Build GRCh38
rs925685844 Chr.6: 26043404-26043404 on Build GRCh38
rs752844404 Chr.6: 26043542-26043542 on Build GRCh38
rs751986897 Chr.6: 26043535-26043536 on Build GRCh38
rs921903749 Chr.6: 26043426-26043426 on Build GRCh38
rs190950786 Chr.6: 26043682-26043682 on Build GRCh38
rs758468487 Chr.6: 26043598-26043598 on Build GRCh38
rs769875529 Chr.6: 26043702-26043702 on Build GRCh38
rs768596438 Chr.6: 26043667-26043667 on Build GRCh38
rs781248502 Chr.6: 26043699-26043699 on Build GRCh38
rs985285420 Chr.6: 26043806-26043806 on Build GRCh38
rs372065604 Chr.6: 26043430-26043430 on Build GRCh38
rs760513217 Chr.6: 26043583-26043583 on Build GRCh38
rs761691872 Chr.6: 26043514-26043514 on Build GRCh38
rs895887049 Chr.6: 26043837-26043837 on Build GRCh38
rs1016169623 Chr.6: 26043735-26043735 on Build GRCh38
rs199714846 Chr.6: 26043581-26043581 on Build GRCh38
rs779181397 Chr.6: 26043541-26043541 on Build GRCh38
rs922330995 Chr.6: 26043784-26043784 on Build GRCh38
rs748989913 Chr.6: 26043560-26043560 on Build GRCh38
rs891486738 Chr.6: 26043865-26043865 on Build GRCh38
rs771547734 Chr.6: 26043632-26043632 on Build GRCh38
rs746964834 Chr.6: 26043643-26043643 on Build GRCh38
rs11751732 Chr.6: 26043836-26043836 on Build GRCh38
rs769055166 Chr.6: 26043747-26043747 on Build GRCh38
rs779161052 Chr.6: 26043606-26043606 on Build GRCh38
rs180934723 Chr.6: 26043471-26043471 on Build GRCh38
rs567381335 Chr.6: 26043661-26043661 on Build GRCh38
rs764661719 Chr.6: 26043599-26043599 on Build GRCh38
rs776523524 Chr.6: 26043646-26043646 on Build GRCh38
rs953323196 Chr.6: 26043659-26043659 on Build GRCh38
rs368020868 Chr.6: 26043575-26043575 on Build GRCh38
rs144359238 Chr.6: 26043774-26043774 on Build GRCh38
rs199522923 Chr.6: 26043673-26043673 on Build GRCh38
rs911025578 Chr.6: 26043810-26043810 on Build GRCh38
rs984638612 Chr.6: 26043727-26043727 on Build GRCh38
rs201553217 Chr.6: 26043666-26043666 on Build GRCh38
rs145254404 Chr.6: 26043411-26043411 on Build GRCh38
rs768955458 Chr.6: 26043681-26043681 on Build GRCh38
rs757602618 Chr.6: 26043535-26043538 on Build GRCh38
rs745854481 Chr.6: 26043700-26043700 on Build GRCh38
rs368153259 Chr.6: 26043695-26043695 on Build GRCh38
rs1043319616 Chr.6: 26043782-26043782 on Build GRCh38
rs371643209 Chr.6: 26043704-26043704 on Build GRCh38
rs773812336 Chr.6: 26043564-26043564 on Build GRCh38
rs933787906 Chr.6: 26043665-26043665 on Build GRCh38
rs374608319 Chr.6: 26043553-26043553 on Build GRCh38
rs576497987 Chr.6: 26043805-26043805 on Build GRCh38
rs767385566 Chr.6: 26043516-26043516 on Build GRCh38
rs955256319 Chr.6: 26043803-26043803 on Build GRCh38
rs764008129 Chr.6: 26043445-26043445 on Build GRCh38
rs375309995 Chr.6: 26043683-26043683 on Build GRCh38
rs938753430 Chr.6: 26043749-26043749 on Build GRCh38
rs549642579 Chr.6: 26043702-26043703 on Build GRCh38
rs547034704 Chr.6: 26043451-26043451 on Build GRCh38
rs753910669 Chr.6: 26043464-26043464 on Build GRCh38
rs200869185 Chr.6: 26043605-26043605 on Build GRCh38
rs750933462 Chr.6: 26043583-26043585 on Build GRCh38
rs976368694 Chr.6: 26043400-26043400 on Build GRCh38
rs139397968 Chr.6: 26043647-26043647 on Build GRCh38
rs757782831 Chr.6: 26043701-26043710 on Build GRCh38
rs750208563 Chr.6: 26043456-26043456 on Build GRCh38
rs773165770 Chr.6: 26043638-26043638 on Build GRCh38

View Full Product Details