Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 159036623-159037101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPR2
Entrez Gene ID
7434
Gene Name
vasoactive intestinal peptide receptor 2
Gene Aliases
C16DUPq36.3, DUP7q36.3, PACAP-R-3, PACAP-R3, VIP-R-2, VPAC2, VPAC2R, VPCAP2R
UniGene
Hs.585052
Gene Chromosome Location
Chr.7: 159028175-159144957 on Build GRCh38
Transcripts
NM_001304522.1 NM_001308259.1 NM_003382.4 NR_130758.1 XM_005249561.3 XM_006716107.2 XM_006716108.3 XM_011516550.2 XM_017012580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3636886 c.723C>T p.F241F Chr.7: 159036777-159036777 on Build GRCh38
3636885 c.738G>A p.L246L Chr.7: 159036762-159036762 on Build GRCh38
5450367 c.673C>A p.L225I Chr.7: 159036827-159036827 on Build GRCh38
4505258 c.687C>T p.T229T Chr.7: 159036813-159036813 on Build GRCh38
1292478 c.698C>T p.A233V Chr.7: 159036802-159036802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747398205 Chr.7: 159037015-159037015 on Build GRCh38
rs768649678 Chr.7: 159036807-159036807 on Build GRCh38
rs931052491 Chr.7: 159037042-159037042 on Build GRCh38
rs374945318 Chr.7: 159036945-159036945 on Build GRCh38
rs151055829 Chr.7: 159036656-159036656 on Build GRCh38
rs570429614 Chr.7: 159036989-159036989 on Build GRCh38
rs769456799 Chr.7: 159036800-159036800 on Build GRCh38
rs1035610096 Chr.7: 159037076-159037076 on Build GRCh38
rs932183953 Chr.7: 159037024-159037024 on Build GRCh38
rs1001148965 Chr.7: 159037063-159037063 on Build GRCh38
rs201305770 Chr.7: 159036793-159036793 on Build GRCh38
rs752470525 Chr.7: 159036761-159036761 on Build GRCh38
rs185052423 Chr.7: 159036662-159036662 on Build GRCh38
rs762435762 Chr.7: 159036789-159036790 on Build GRCh38
rs990704035 Chr.7: 159036922-159036922 on Build GRCh38
rs781392198 Chr.7: 159036795-159036795 on Build GRCh38
rs775262765 Chr.7: 159036725-159036725 on Build GRCh38
rs189906099 Chr.7: 159036974-159036974 on Build GRCh38
rs778962771 Chr.7: 159036906-159036906 on Build GRCh38
rs1036500003 Chr.7: 159037005-159037005 on Build GRCh38
rs765977202 Chr.7: 159036825-159036825 on Build GRCh38
rs978187931 Chr.7: 159036973-159036973 on Build GRCh38
rs773960364 Chr.7: 159036734-159036734 on Build GRCh38
rs370531320 Chr.7: 159036733-159036733 on Build GRCh38
rs143276573 Chr.7: 159036801-159036801 on Build GRCh38
rs767637413 Chr.7: 159036874-159036874 on Build GRCh38
rs772155938 Chr.7: 159036912-159036912 on Build GRCh38
rs767221133 Chr.7: 159036740-159036740 on Build GRCh38
rs900733226 Chr.7: 159036785-159036785 on Build GRCh38
rs757299940 Chr.7: 159036872-159036872 on Build GRCh38
rs141052651 Chr.7: 159036661-159036661 on Build GRCh38
rs772976425 Chr.7: 159036819-159036819 on Build GRCh38
rs778134660 Chr.7: 159036706-159036706 on Build GRCh38
rs374435223 Chr.7: 159036879-159036879 on Build GRCh38
rs936789808 Chr.7: 159036977-159036977 on Build GRCh38
rs566325667 Chr.7: 159037035-159037035 on Build GRCh38
rs758895423 Chr.7: 159036790-159036790 on Build GRCh38
rs890657160 Chr.7: 159036676-159036676 on Build GRCh38
rs370111841 Chr.7: 159036911-159036911 on Build GRCh38
rs978744920 Chr.7: 159036648-159036648 on Build GRCh38
rs763296133 Chr.7: 159036941-159036941 on Build GRCh38
rs372399968 Chr.7: 159036728-159036728 on Build GRCh38
rs1020789513 Chr.7: 159036902-159036902 on Build GRCh38
rs761395005 Chr.7: 159036809-159036809 on Build GRCh38
rs540938977 Chr.7: 159036699-159036699 on Build GRCh38
rs764886905 Chr.7: 159036937-159036937 on Build GRCh38
rs754878327 Chr.7: 159036900-159036900 on Build GRCh38
rs763238183 Chr.7: 159036850-159036850 on Build GRCh38
rs567755241 Chr.7: 159036934-159036934 on Build GRCh38
rs144784720 Chr.7: 159036677-159036677 on Build GRCh38
rs555812541 Chr.7: 159037080-159037080 on Build GRCh38
rs912148786 Chr.7: 159036984-159036984 on Build GRCh38
rs1054023368 Chr.7: 159036739-159036739 on Build GRCh38
rs781402383 Chr.7: 159036719-159036719 on Build GRCh38
rs116647211 Chr.7: 159036694-159036694 on Build GRCh38
rs753627946 Chr.7: 159036706-159036710 on Build GRCh38
rs535291535 Chr.7: 159037039-159037039 on Build GRCh38
rs776200872 Chr.7: 159036935-159036935 on Build GRCh38
rs183188951 Chr.7: 159036990-159036990 on Build GRCh38
rs113343782 Chr.7: 159037020-159037020 on Build GRCh38
rs777779063 Chr.7: 159036705-159036705 on Build GRCh38
rs752122787 Chr.7: 159036865-159036865 on Build GRCh38
rs1052058115 Chr.7: 159037032-159037032 on Build GRCh38
rs759787939 Chr.7: 159036758-159036758 on Build GRCh38
rs769701404 Chr.7: 159036723-159036723 on Build GRCh38
rs772737308 Chr.7: 159036921-159036921 on Build GRCh38
rs749070438 Chr.7: 159036804-159036804 on Build GRCh38
rs896908956 Chr.7: 159037021-159037021 on Build GRCh38
rs745644799 Chr.7: 159036720-159036720 on Build GRCh38
rs530646947 Chr.7: 159036944-159036944 on Build GRCh38
rs78564798 Chr.7: 159036791-159036791 on Build GRCh38
rs905562951 Chr.7: 159037062-159037062 on Build GRCh38
rs940859240 Chr.7: 159037000-159037000 on Build GRCh38
rs765681011 Chr.7: 159036759-159036759 on Build GRCh38
rs149197032 Chr.7: 159036797-159036797 on Build GRCh38
rs200149573 Chr.7: 159036736-159036736 on Build GRCh38
rs746503068 Chr.7: 159036926-159036926 on Build GRCh38
rs368871135 Chr.7: 159036895-159036895 on Build GRCh38
rs767590939 Chr.7: 159036743-159036743 on Build GRCh38
rs780145643 Chr.7: 159036890-159036890 on Build GRCh38
rs924044833 Chr.7: 159036976-159036976 on Build GRCh38
rs375889196 Chr.7: 159036708-159036708 on Build GRCh38
rs759245467 Chr.7: 159036821-159036821 on Build GRCh38
rs764568382 Chr.7: 159036855-159036855 on Build GRCh38
rs771748798 Chr.7: 159036818-159036818 on Build GRCh38
rs1048112721 Chr.7: 159037048-159037048 on Build GRCh38
rs201210089 Chr.7: 159036822-159036822 on Build GRCh38
rs750166509 Chr.7: 159036745-159036745 on Build GRCh38
rs949436723 Chr.7: 159036687-159036687 on Build GRCh38
rs753035890 Chr.7: 159036836-159036836 on Build GRCh38

View Full Product Details