Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150546300-150546856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
UniGene
Hs.655782
Gene Chromosome Location
Chr.3: 150546678-150586016 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 NM_014445.3 XM_011512698.2

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 NM_014445.3 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365037 c.18G>C p.P6P Chr.3: 150546820-150546820 on Build GRCh38
479573 c.1A>G p.M1V Chr.3: 150546803-150546803 on Build GRCh38
5974594 c.26C>T p.T9I Chr.3: 150546828-150546828 on Build GRCh38
3588638 c.21C>T p.L7L Chr.3: 150546823-150546823 on Build GRCh38
5398380 c.13A>G p.T5A Chr.3: 150546815-150546815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374744533 Chr.3: 150546809-150546809 on Build GRCh38
rs747640825 Chr.3: 150546807-150546807 on Build GRCh38
rs573055668 Chr.3: 150546525-150546525 on Build GRCh38
rs756611096 Chr.3: 150546776-150546776 on Build GRCh38
rs369746328 Chr.3: 150546818-150546818 on Build GRCh38
rs1044392830 Chr.3: 150546588-150546588 on Build GRCh38
rs925869326 Chr.3: 150546530-150546530 on Build GRCh38
rs1003335820 Chr.3: 150546594-150546594 on Build GRCh38
rs6801769 Chr.3: 150546443-150546443 on Build GRCh38
rs556042063 Chr.3: 150546454-150546454 on Build GRCh38
rs1042989597 Chr.3: 150546432-150546432 on Build GRCh38
rs778567149 Chr.3: 150546593-150546593 on Build GRCh38
rs984450558 Chr.3: 150546492-150546492 on Build GRCh38
rs374657010 Chr.3: 150546479-150546479 on Build GRCh38
rs559623521 Chr.3: 150546515-150546515 on Build GRCh38
rs867596217 Chr.3: 150546493-150546493 on Build GRCh38
rs746701306 Chr.3: 150546796-150546796 on Build GRCh38
rs771583168 Chr.3: 150546813-150546813 on Build GRCh38
rs545149907 Chr.3: 150546771-150546771 on Build GRCh38
rs982445797 Chr.3: 150546705-150546712 on Build GRCh38
rs770906744 Chr.3: 150546815-150546815 on Build GRCh38
rs943090141 Chr.3: 150546494-150546494 on Build GRCh38
rs563529297 Chr.3: 150546595-150546595 on Build GRCh38
rs938735590 Chr.3: 150546710-150546710 on Build GRCh38
rs570266468 Chr.3: 150546328-150546328 on Build GRCh38
rs1002183890 Chr.3: 150546510-150546510 on Build GRCh38
rs535656712 Chr.3: 150546341-150546341 on Build GRCh38
rs373140920 Chr.3: 150546649-150546649 on Build GRCh38
rs913092901 Chr.3: 150546372-150546372 on Build GRCh38
rs534858765 Chr.3: 150546647-150546647 on Build GRCh38
rs1018518010 Chr.3: 150546759-150546759 on Build GRCh38
rs535308008 Chr.3: 150546610-150546610 on Build GRCh38
rs1046879009 Chr.3: 150546568-150546568 on Build GRCh38
rs201012452 Chr.3: 150546783-150546783 on Build GRCh38
rs923086037 Chr.3: 150546671-150546671 on Build GRCh38
rs905863112 Chr.3: 150546589-150546589 on Build GRCh38
rs375256643 Chr.3: 150546761-150546761 on Build GRCh38
rs749230981 Chr.3: 150546811-150546811 on Build GRCh38
rs745406269 Chr.3: 150546798-150546798 on Build GRCh38
rs192973743 Chr.3: 150546777-150546777 on Build GRCh38
rs947245573 Chr.3: 150546692-150546692 on Build GRCh38
rs369847308 Chr.3: 150546531-150546531 on Build GRCh38
rs551801632 Chr.3: 150546653-150546653 on Build GRCh38
rs188629338 Chr.3: 150546560-150546560 on Build GRCh38
rs556960651 Chr.3: 150546696-150546696 on Build GRCh38
rs905433246 Chr.3: 150546587-150546587 on Build GRCh38
rs973386630 Chr.3: 150546462-150546462 on Build GRCh38
rs780594445 Chr.3: 150546806-150546806 on Build GRCh38
rs796813108 Chr.3: 150546469-150546469 on Build GRCh38
rs905850779 Chr.3: 150546703-150546703 on Build GRCh38
rs575542256 Chr.3: 150546789-150546789 on Build GRCh38
rs1038563698 Chr.3: 150546582-150546582 on Build GRCh38
rs754322867 Chr.3: 150546780-150546780 on Build GRCh38
rs967150949 Chr.3: 150546526-150546526 on Build GRCh38
rs549048644 Chr.3: 150546621-150546621 on Build GRCh38
rs537013007 Chr.3: 150546445-150546445 on Build GRCh38
rs752858521 Chr.3: 150546693-150546694 on Build GRCh38
rs1020957161 Chr.3: 150546628-150546628 on Build GRCh38
rs750588027 Chr.3: 150546474-150546474 on Build GRCh38
rs6790726 Chr.3: 150546641-150546641 on Build GRCh38
rs759789637 Chr.3: 150546764-150546764 on Build GRCh38
rs914425420 Chr.3: 150546691-150546691 on Build GRCh38
rs988225809 Chr.3: 150546522-150546522 on Build GRCh38
rs201718601 Chr.3: 150546802-150546802 on Build GRCh38
rs934547452 Chr.3: 150546554-150546554 on Build GRCh38
rs11548122 Chr.3: 150546320-150546320 on Build GRCh38
rs897311071 Chr.3: 150546607-150546607 on Build GRCh38
rs762887230 Chr.3: 150546757-150546757 on Build GRCh38
rs550103045 Chr.3: 150546574-150546574 on Build GRCh38
rs764129238 Chr.3: 150546828-150546828 on Build GRCh38
rs536382837 Chr.3: 150546739-150546739 on Build GRCh38
rs544997476 Chr.3: 150546541-150546541 on Build GRCh38
rs1027088119 Chr.3: 150546741-150546741 on Build GRCh38
rs565925718 Chr.3: 150546351-150546351 on Build GRCh38
rs767738799 Chr.3: 150546824-150546824 on Build GRCh38
rs574287509 Chr.3: 150546498-150546498 on Build GRCh38
rs1054004859 Chr.3: 150546406-150546417 on Build GRCh38
rs1035462811 Chr.3: 150546511-150546511 on Build GRCh38
rs975798899 Chr.3: 150546775-150546775 on Build GRCh38
rs758745886 Chr.3: 150546770-150546770 on Build GRCh38
rs960806781 Chr.3: 150546519-150546519 on Build GRCh38
rs914372974 Chr.3: 150546821-150546821 on Build GRCh38
rs1035812963 Chr.3: 150546604-150546604 on Build GRCh38
rs1053052754 Chr.3: 150546344-150546344 on Build GRCh38
rs961090453 Chr.3: 150546360-150546360 on Build GRCh38
rs867223026 Chr.3: 150546672-150546672 on Build GRCh38
rs774229838 Chr.3: 150546817-150546817 on Build GRCh38
rs978568875 Chr.3: 150546529-150546529 on Build GRCh38
rs201751272 Chr.3: 150546754-150546754 on Build GRCh38
rs975774914 Chr.3: 150546679-150546679 on Build GRCh38
rs146864908 Chr.3: 150546667-150546667 on Build GRCh38
rs573830754 Chr.3: 150546477-150546477 on Build GRCh38
rs773320355 Chr.3: 150546810-150546810 on Build GRCh38
rs1017185397 Chr.3: 150546645-150546645 on Build GRCh38
rs529388128 Chr.3: 150546615-150546615 on Build GRCh38
rs1047242860 Chr.3: 150546471-150546471 on Build GRCh38
rs952937513 Chr.3: 150546627-150546627 on Build GRCh38
rs775454510 Chr.3: 150546826-150546826 on Build GRCh38
rs757201665 Chr.3: 150546740-150546740 on Build GRCh38
rs573090669 Chr.3: 150546770-150546770 on Build GRCh38
rs913830305 Chr.3: 150546570-150546571 on Build GRCh38
rs756358484 Chr.3: 150546831-150546839 on Build GRCh38
rs377526748 Chr.3: 150546730-150546730 on Build GRCh38
rs558407433 Chr.3: 150546367-150546367 on Build GRCh38
rs536643019 Chr.3: 150546687-150546687 on Build GRCh38
rs11548120 Chr.3: 150546353-150546353 on Build GRCh38
rs746468990 Chr.3: 150546794-150546794 on Build GRCh38
rs11548121 Chr.3: 150546331-150546331 on Build GRCh38
rs951695700 Chr.3: 150546480-150546480 on Build GRCh38
rs970099912 Chr.3: 150546366-150546366 on Build GRCh38
rs368910885 Chr.3: 150546564-150546564 on Build GRCh38
rs766548799 Chr.3: 150546509-150546509 on Build GRCh38
rs934472622 Chr.3: 150546683-150546683 on Build GRCh38
rs1025831171 Chr.3: 150546337-150546337 on Build GRCh38
rs771404523 Chr.3: 150546744-150546744 on Build GRCh38
rs368151033 Chr.3: 150546762-150546762 on Build GRCh38
rs1007064482 Chr.3: 150546746-150546746 on Build GRCh38
rs760859545 Chr.3: 150546827-150546827 on Build GRCh38
rs1049947 Chr.3: 150546349-150546349 on Build GRCh38

View Full Product Details