Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 55786728-55787292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGS1
Entrez Gene ID
96764
Gene Name
trimethylguanosine synthase 1
Gene Aliases
NCOA6IP, PIMT, PIPMT
UniGene
Hs.335068
Gene Chromosome Location
Chr.8: 55773232-55826445 on Build GRCh38
Transcripts
NM_001317902.1 NM_024831.7 XM_005251328.2 XM_006716485.2 XM_017014004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834846 c.1114G>A p.E372K Chr.8: 55787012-55787012 on Build GRCh38
4733461 c.854delA p.K286fs*3 Chr.8: 55786752-55786752 on Build GRCh38
2719402 c.989A>G p.E330G Chr.8: 55786887-55786887 on Build GRCh38
5039633 c.1147G>A p.D383N Chr.8: 55787045-55787045 on Build GRCh38
1292712 c.970A>G p.I324V Chr.8: 55786868-55786868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895486108 Chr.8: 55787072-55787072 on Build GRCh38
rs1028692409 Chr.8: 55787234-55787234 on Build GRCh38
rs139281321 Chr.8: 55787014-55787014 on Build GRCh38
rs144081619 Chr.8: 55787028-55787028 on Build GRCh38
rs568913195 Chr.8: 55787086-55787086 on Build GRCh38
rs771882048 Chr.8: 55786847-55786847 on Build GRCh38
rs1052749555 Chr.8: 55787048-55787048 on Build GRCh38
rs767655821 Chr.8: 55786800-55786800 on Build GRCh38
rs944312053 Chr.8: 55786884-55786884 on Build GRCh38
rs1002292083 Chr.8: 55787167-55787167 on Build GRCh38
rs143171305 Chr.8: 55786782-55786782 on Build GRCh38
rs151066817 Chr.8: 55787029-55787029 on Build GRCh38
rs142667771 Chr.8: 55786979-55786979 on Build GRCh38
rs762062615 Chr.8: 55787055-55787055 on Build GRCh38
rs749203929 Chr.8: 55786966-55786966 on Build GRCh38
rs373681202 Chr.8: 55786927-55786927 on Build GRCh38
rs149583415 Chr.8: 55787232-55787234 on Build GRCh38
rs769804925 Chr.8: 55786958-55786958 on Build GRCh38
rs931462364 Chr.8: 55786804-55786804 on Build GRCh38
rs756263597 Chr.8: 55787262-55787262 on Build GRCh38
rs552414411 Chr.8: 55786965-55786965 on Build GRCh38
rs768177307 Chr.8: 55786952-55786952 on Build GRCh38
rs759576494 Chr.8: 55786899-55786899 on Build GRCh38
rs773088367 Chr.8: 55786963-55786963 on Build GRCh38
rs777059199 Chr.8: 55786986-55786986 on Build GRCh38
rs762309774 Chr.8: 55787228-55787228 on Build GRCh38
rs760858088 Chr.8: 55786915-55786915 on Build GRCh38
rs747827086 Chr.8: 55786954-55786954 on Build GRCh38
rs775561738 Chr.8: 55787031-55787031 on Build GRCh38
rs760507437 Chr.8: 55786765-55786765 on Build GRCh38
rs948151781 Chr.8: 55787134-55787134 on Build GRCh38
rs369761488 Chr.8: 55786876-55786876 on Build GRCh38
rs1979131 Chr.8: 55787087-55787087 on Build GRCh38
rs767752146 Chr.8: 55786903-55786903 on Build GRCh38
rs11986329 Chr.8: 55786793-55786793 on Build GRCh38
rs373134336 Chr.8: 55787090-55787090 on Build GRCh38
rs905381470 Chr.8: 55786916-55786916 on Build GRCh38
rs763986488 Chr.8: 55787036-55787045 on Build GRCh38
rs61735671 Chr.8: 55787001-55787001 on Build GRCh38
rs781050854 Chr.8: 55786953-55786953 on Build GRCh38
rs368055377 Chr.8: 55786772-55786772 on Build GRCh38
rs888507214 Chr.8: 55787213-55787213 on Build GRCh38
rs985594767 Chr.8: 55787021-55787021 on Build GRCh38
rs771836880 Chr.8: 55786759-55786759 on Build GRCh38
rs369461578 Chr.8: 55787004-55787004 on Build GRCh38
rs965294077 Chr.8: 55786997-55786997 on Build GRCh38
rs752425414 Chr.8: 55787000-55787000 on Build GRCh38
rs868289707 Chr.8: 55786816-55786816 on Build GRCh38
rs757796292 Chr.8: 55786920-55786920 on Build GRCh38
rs765762068 Chr.8: 55787057-55787057 on Build GRCh38
rs766550963 Chr.8: 55786781-55786781 on Build GRCh38
rs767035448 Chr.8: 55787101-55787101 on Build GRCh38
rs759129424 Chr.8: 55786783-55786784 on Build GRCh38
rs756419314 Chr.8: 55786807-55786807 on Build GRCh38
rs750566792 Chr.8: 55787246-55787246 on Build GRCh38
rs530404869 Chr.8: 55786995-55786995 on Build GRCh38
rs764965388 Chr.8: 55786858-55786858 on Build GRCh38
rs758228346 Chr.8: 55787028-55787034 on Build GRCh38
rs554939598 Chr.8: 55787145-55787145 on Build GRCh38
rs140022488 Chr.8: 55786892-55786892 on Build GRCh38
rs371581459 Chr.8: 55786868-55786868 on Build GRCh38
rs751457792 Chr.8: 55787040-55787041 on Build GRCh38
rs375517082 Chr.8: 55786870-55786870 on Build GRCh38
rs765611540 Chr.8: 55786994-55786994 on Build GRCh38
rs553224427 Chr.8: 55787229-55787229 on Build GRCh38
rs768945072 Chr.8: 55787036-55787036 on Build GRCh38
rs202091137 Chr.8: 55786978-55786978 on Build GRCh38
rs769557194 Chr.8: 55786885-55786885 on Build GRCh38
rs371657989 Chr.8: 55786990-55786990 on Build GRCh38
rs866092575 Chr.8: 55786763-55786763 on Build GRCh38
rs752335971 Chr.8: 55787108-55787108 on Build GRCh38
rs748002193 Chr.8: 55787020-55787020 on Build GRCh38
rs777515351 Chr.8: 55787026-55787026 on Build GRCh38
rs773696362 Chr.8: 55787075-55787075 on Build GRCh38
rs757699524 Chr.8: 55786827-55786827 on Build GRCh38
rs1042852046 Chr.8: 55786766-55786766 on Build GRCh38
rs961019607 Chr.8: 55786999-55786999 on Build GRCh38
rs144255211 Chr.8: 55786882-55786882 on Build GRCh38
rs1979130 Chr.8: 55787222-55787222 on Build GRCh38
rs763371313 Chr.8: 55786945-55786945 on Build GRCh38
rs139249885 Chr.8: 55786758-55786758 on Build GRCh38
rs776765729 Chr.8: 55787041-55787041 on Build GRCh38
rs746995608 Chr.8: 55787032-55787032 on Build GRCh38
rs754401497 Chr.8: 55787093-55787093 on Build GRCh38
rs779677869 Chr.8: 55786943-55786943 on Build GRCh38
rs752968715 Chr.8: 55786908-55786908 on Build GRCh38
rs932407133 Chr.8: 55786761-55786761 on Build GRCh38
rs569412580 Chr.8: 55787010-55787010 on Build GRCh38
rs764587945 Chr.8: 55786919-55786919 on Build GRCh38
rs1015534411 Chr.8: 55787224-55787224 on Build GRCh38
rs573000754 Chr.8: 55786760-55786760 on Build GRCh38
rs778543133 Chr.8: 55786841-55786841 on Build GRCh38
rs758030179 Chr.8: 55786938-55786938 on Build GRCh38
rs761627593 Chr.8: 55786777-55786777 on Build GRCh38
rs185514267 Chr.8: 55787200-55787200 on Build GRCh38
rs777975868 Chr.8: 55786811-55786811 on Build GRCh38
rs552066189 Chr.8: 55787059-55787059 on Build GRCh38
rs905349839 Chr.8: 55787129-55787129 on Build GRCh38
rs375438967 Chr.8: 55786930-55786930 on Build GRCh38
rs149584240 Chr.8: 55786863-55786863 on Build GRCh38
rs769535706 Chr.8: 55786810-55786810 on Build GRCh38
rs755915533 Chr.8: 55787025-55787025 on Build GRCh38
rs138233590 Chr.8: 55786842-55786842 on Build GRCh38
rs768737718 Chr.8: 55786767-55786767 on Build GRCh38
rs767450804 Chr.8: 55787230-55787230 on Build GRCh38
rs564885473 Chr.8: 55786825-55786825 on Build GRCh38
rs146017987 Chr.8: 55786998-55786998 on Build GRCh38

View Full Product Details