Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 79383881-79384456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC24
Entrez Gene ID
219654
Gene Name
zinc finger CCHC-type containing 24
Gene Aliases
C10orf56, Z3CXXC8
UniGene
Hs.523080
Gene Chromosome Location
Chr.10: 79382325-79445627 on Build GRCh38
Transcripts
NM_153367.3 XM_011539452.2

SNP Detail

SNP ID Chromosome Location
rs1009140619 Chr.10: 79384250-79384250 on Build GRCh38
rs1010632145 Chr.10: 79384296-79384296 on Build GRCh38
rs757592244 Chr.10: 79383933-79383933 on Build GRCh38
rs12853 Chr.10: 79383910-79383910 on Build GRCh38
rs575117779 Chr.10: 79384280-79384280 on Build GRCh38
rs780909665 Chr.10: 79384106-79384106 on Build GRCh38
rs538737909 Chr.10: 79384170-79384170 on Build GRCh38
rs769459347 Chr.10: 79384311-79384311 on Build GRCh38
rs942930891 Chr.10: 79384226-79384226 on Build GRCh38
rs1012126766 Chr.10: 79384404-79384404 on Build GRCh38
rs930966999 Chr.10: 79384016-79384016 on Build GRCh38
rs779286803 Chr.10: 79384030-79384030 on Build GRCh38
rs970333016 Chr.10: 79384076-79384076 on Build GRCh38
rs948526057 Chr.10: 79384403-79384403 on Build GRCh38
rs148823023 Chr.10: 79384264-79384264 on Build GRCh38
rs545875386 Chr.10: 79384416-79384416 on Build GRCh38
rs566609048 Chr.10: 79384072-79384072 on Build GRCh38
rs915867929 Chr.10: 79384386-79384386 on Build GRCh38
rs1046695564 Chr.10: 79384028-79384028 on Build GRCh38
rs532497552 Chr.10: 79384408-79384408 on Build GRCh38
rs890467511 Chr.10: 79384222-79384222 on Build GRCh38
rs566144889 Chr.10: 79384374-79384374 on Build GRCh38
rs1049071605 Chr.10: 79384206-79384206 on Build GRCh38
rs964880033 Chr.10: 79383926-79383926 on Build GRCh38
rs1043157502 Chr.10: 79384103-79384103 on Build GRCh38
rs1041107557 Chr.10: 79383924-79383924 on Build GRCh38
rs560310052 Chr.10: 79384407-79384407 on Build GRCh38
rs866616535 Chr.10: 79384343-79384343 on Build GRCh38
rs142485283 Chr.10: 79384281-79384281 on Build GRCh38
rs73293507 Chr.10: 79383925-79383925 on Build GRCh38
rs911389080 Chr.10: 79384221-79384221 on Build GRCh38
rs931063980 Chr.10: 79384205-79384205 on Build GRCh38
rs546158167 Chr.10: 79384338-79384338 on Build GRCh38
rs908206946 Chr.10: 79384051-79384051 on Build GRCh38
rs773235848 Chr.10: 79384320-79384320 on Build GRCh38
rs116543832 Chr.10: 79384419-79384419 on Build GRCh38
rs1012776983 Chr.10: 79384081-79384081 on Build GRCh38
rs554987692 Chr.10: 79384285-79384285 on Build GRCh38
rs1041703296 Chr.10: 79384334-79384334 on Build GRCh38
rs113517478 Chr.10: 79383932-79383932 on Build GRCh38
rs879590737 Chr.10: 79384287-79384287 on Build GRCh38
rs553160987 Chr.10: 79383942-79383942 on Build GRCh38
rs1053643999 Chr.10: 79384430-79384430 on Build GRCh38
rs901189552 Chr.10: 79384282-79384282 on Build GRCh38
rs934735825 Chr.10: 79384426-79384426 on Build GRCh38
rs1031650063 Chr.10: 79384193-79384193 on Build GRCh38
rs746667390 Chr.10: 79384071-79384071 on Build GRCh38
rs908172393 Chr.10: 79384340-79384340 on Build GRCh38
rs959317638 Chr.10: 79383985-79383985 on Build GRCh38
rs919716715 Chr.10: 79383943-79383943 on Build GRCh38
rs543302660 Chr.10: 79384067-79384067 on Build GRCh38
rs575031156 Chr.10: 79384344-79384344 on Build GRCh38

View Full Product Details