Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73243566-73244118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC9
Entrez Gene ID
23234
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
HDJC9, JDD1, SB73
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73241954-73247331 on Build GRCh38
Transcripts
NM_015190.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465940 c.588G>C p.E196D Chr.10: 73243918-73243918 on Build GRCh38
7365123 c.637G>A p.V213M Chr.10: 73243869-73243869 on Build GRCh38
1603706 c.627T>C p.L209L Chr.10: 73243879-73243879 on Build GRCh38
6220647 c.636C>T p.G212G Chr.10: 73243870-73243870 on Build GRCh38
6647426 c.620T>C p.L207S Chr.10: 73243886-73243886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925543453 Chr.10: 73243752-73243756 on Build GRCh38
rs1007986754 Chr.10: 73244017-73244017 on Build GRCh38
rs188683190 Chr.10: 73243738-73243738 on Build GRCh38
rs889495946 Chr.10: 73243983-73243983 on Build GRCh38
rs752709372 Chr.10: 73243898-73243898 on Build GRCh38
rs760415559 Chr.10: 73243833-73243833 on Build GRCh38
rs765110686 Chr.10: 73243923-73243928 on Build GRCh38
rs146921292 Chr.10: 73243869-73243869 on Build GRCh38
rs367712008 Chr.10: 73243682-73243682 on Build GRCh38
rs757399233 Chr.10: 73243870-73243870 on Build GRCh38
rs560667394 Chr.10: 73243695-73243695 on Build GRCh38
rs768354123 Chr.10: 73243844-73243844 on Build GRCh38
rs975767541 Chr.10: 73243669-73243669 on Build GRCh38
rs761522007 Chr.10: 73243973-73243973 on Build GRCh38
rs752601567 Chr.10: 73243976-73243976 on Build GRCh38
rs746354880 Chr.10: 73243818-73243818 on Build GRCh38
rs771547794 Chr.10: 73243821-73243821 on Build GRCh38
rs965821152 Chr.10: 73243958-73243958 on Build GRCh38
rs560067839 Chr.10: 73243959-73243959 on Build GRCh38
rs1028754266 Chr.10: 73244008-73244008 on Build GRCh38
rs546476110 Chr.10: 73244012-73244013 on Build GRCh38
rs988303971 Chr.10: 73244073-73244073 on Build GRCh38
rs767510500 Chr.10: 73243890-73243890 on Build GRCh38
rs543162418 Chr.10: 73244071-73244071 on Build GRCh38
rs374658313 Chr.10: 73243673-73243673 on Build GRCh38
rs541844405 Chr.10: 73243664-73243664 on Build GRCh38
rs779331291 Chr.10: 73243935-73243935 on Build GRCh38
rs918100613 Chr.10: 73244082-73244082 on Build GRCh38
rs772571747 Chr.10: 73243951-73243951 on Build GRCh38
rs553467405 Chr.10: 73243956-73243956 on Build GRCh38
rs539528863 Chr.10: 73244061-73244061 on Build GRCh38
rs966195553 Chr.10: 73243668-73243668 on Build GRCh38
rs1000168196 Chr.10: 73244053-73244053 on Build GRCh38
rs756648925 Chr.10: 73243988-73243988 on Build GRCh38
rs780569451 Chr.10: 73244039-73244039 on Build GRCh38
rs757659619 Chr.10: 73243932-73243932 on Build GRCh38
rs552394659 Chr.10: 73243740-73243740 on Build GRCh38
rs570672243 Chr.10: 73243741-73243741 on Build GRCh38
rs983267534 Chr.10: 73243604-73243604 on Build GRCh38
rs749614502 Chr.10: 73243795-73243795 on Build GRCh38
rs771451161 Chr.10: 73243797-73243797 on Build GRCh38
rs541331218 Chr.10: 73243823-73243823 on Build GRCh38
rs200313574 Chr.10: 73243948-73243948 on Build GRCh38
rs138208732 Chr.10: 73243914-73243914 on Build GRCh38
rs949538081 Chr.10: 73243787-73243787 on Build GRCh38
rs776468235 Chr.10: 73243960-73243960 on Build GRCh38
rs765054910 Chr.10: 73243871-73243871 on Build GRCh38
rs779432879 Chr.10: 73243954-73243954 on Build GRCh38
rs146570416 Chr.10: 73243771-73243771 on Build GRCh38
rs771431510 Chr.10: 73243813-73243814 on Build GRCh38
rs192441500 Chr.10: 73243742-73243742 on Build GRCh38
rs966923183 Chr.10: 73244047-73244047 on Build GRCh38
rs951855229 Chr.10: 73243603-73243603 on Build GRCh38
rs762937556 Chr.10: 73243889-73243889 on Build GRCh38
rs1004639886 Chr.10: 73243920-73243920 on Build GRCh38
rs777924498 Chr.10: 73243910-73243910 on Build GRCh38
rs113840742 Chr.10: 73243592-73243592 on Build GRCh38
rs763100087 Chr.10: 73243627-73243627 on Build GRCh38
rs1032624396 Chr.10: 73244077-73244077 on Build GRCh38
rs376845849 Chr.10: 73243864-73243864 on Build GRCh38
rs907592489 Chr.10: 73243650-73243650 on Build GRCh38
rs750398432 Chr.10: 73243882-73243882 on Build GRCh38
rs921632680 Chr.10: 73243729-73243729 on Build GRCh38
rs1052611575 Chr.10: 73243768-73243768 on Build GRCh38
rs935601196 Chr.10: 73244054-73244054 on Build GRCh38
rs779338360 Chr.10: 73243799-73243799 on Build GRCh38
rs935445730 Chr.10: 73243759-73243759 on Build GRCh38
rs756362271 Chr.10: 73243908-73243908 on Build GRCh38
rs976260697 Chr.10: 73244004-73244004 on Build GRCh38

View Full Product Details