Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128937703-128938245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53AIP1
Entrez Gene ID
63970
Gene Name
tumor protein p53 regulated apoptosis inducing protein 1
Gene Aliases
P53AIP1
UniGene
Hs.160953
Gene Chromosome Location
Chr.11: 128934732-128944233 on Build GRCh38
Transcripts
NM_001195194.1 NM_001195195.1 NM_001251964.1 NM_022112.2 XM_017018113.1 XM_017018114.1 XM_017018115.1 XM_017018116.1 XM_017018117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
392766 c.74delG p.G25fs*9 Chr.11: 128937745-128937745 on Build GRCh38
4636558 c.61A>C p.R21R Chr.11: 128937758-128937758 on Build GRCh38
3666186 c.75C>T p.G25G Chr.11: 128937744-128937744 on Build GRCh38
3445218 c.24C>T p.S8S Chr.11: 128937795-128937795 on Build GRCh38
3415681 c.29G>T p.R10I Chr.11: 128937790-128937790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537076644 Chr.11: 128937746-128937746 on Build GRCh38
rs931286886 Chr.11: 128937831-128937831 on Build GRCh38
rs944151282 Chr.11: 128937775-128937775 on Build GRCh38
rs112432537 Chr.11: 128937885-128937885 on Build GRCh38
rs971707789 Chr.11: 128938107-128938107 on Build GRCh38
rs35942033 Chr.11: 128937799-128937799 on Build GRCh38
rs140061223 Chr.11: 128938139-128938139 on Build GRCh38
rs761705959 Chr.11: 128937880-128937880 on Build GRCh38
rs764633518 Chr.11: 128937844-128937844 on Build GRCh38
rs200155588 Chr.11: 128937758-128937758 on Build GRCh38
rs77790882 Chr.11: 128937804-128937804 on Build GRCh38
rs1034570149 Chr.11: 128938121-128938121 on Build GRCh38
rs776968241 Chr.11: 128937837-128937837 on Build GRCh38
rs552324040 Chr.11: 128938202-128938202 on Build GRCh38
rs1015495731 Chr.11: 128938083-128938083 on Build GRCh38
rs761059884 Chr.11: 128937827-128937827 on Build GRCh38
rs749416440 Chr.11: 128937756-128937756 on Build GRCh38
rs957211524 Chr.11: 128938127-128938127 on Build GRCh38
rs935517148 Chr.11: 128937744-128937744 on Build GRCh38
rs187874729 Chr.11: 128938008-128938008 on Build GRCh38
rs965497254 Chr.11: 128938166-128938166 on Build GRCh38
rs145522015 Chr.11: 128937748-128937748 on Build GRCh38
rs995136479 Chr.11: 128938014-128938014 on Build GRCh38
rs972039049 Chr.11: 128937726-128937726 on Build GRCh38
rs1022696895 Chr.11: 128938048-128938048 on Build GRCh38
rs140191758 Chr.11: 128937724-128937724 on Build GRCh38
rs772630262 Chr.11: 128937817-128937817 on Build GRCh38
rs943844545 Chr.11: 128938214-128938214 on Build GRCh38
rs999824636 Chr.11: 128938101-128938101 on Build GRCh38
rs1005079349 Chr.11: 128938054-128938054 on Build GRCh38
rs945898148 Chr.11: 128937852-128937852 on Build GRCh38
rs151337184 Chr.11: 128937723-128937723 on Build GRCh38
rs943943256 Chr.11: 128937941-128937941 on Build GRCh38
rs765084280 Chr.11: 128938047-128938047 on Build GRCh38
rs990081577 Chr.11: 128938132-128938132 on Build GRCh38
rs367595853 Chr.11: 128937729-128937729 on Build GRCh38
rs537153650 Chr.11: 128937818-128937818 on Build GRCh38
rs758240242 Chr.11: 128937740-128937740 on Build GRCh38
rs749996500 Chr.11: 128937734-128937734 on Build GRCh38
rs371444093 Chr.11: 128937833-128937833 on Build GRCh38
rs560935124 Chr.11: 128938114-128938114 on Build GRCh38
rs1039407869 Chr.11: 128938006-128938006 on Build GRCh38
rs565461039 Chr.11: 128937967-128937967 on Build GRCh38
rs545585080 Chr.11: 128937919-128937919 on Build GRCh38
rs1004128316 Chr.11: 128938003-128938003 on Build GRCh38
rs532195260 Chr.11: 128938186-128938186 on Build GRCh38
rs573486718 Chr.11: 128937917-128937917 on Build GRCh38
rs78994885 Chr.11: 128938133-128938133 on Build GRCh38
rs755683470 Chr.11: 128937817-128937818 on Build GRCh38
rs541601465 Chr.11: 128938187-128938187 on Build GRCh38
rs192636062 Chr.11: 128938016-128938016 on Build GRCh38
rs185711859 Chr.11: 128938177-128938177 on Build GRCh38
rs201878956 Chr.11: 128937752-128937752 on Build GRCh38
rs141395772 Chr.11: 128937755-128937756 on Build GRCh38
rs958968505 Chr.11: 128938129-128938129 on Build GRCh38
rs751565537 Chr.11: 128937747-128937747 on Build GRCh38
rs780923717 Chr.11: 128937754-128937754 on Build GRCh38
rs761410644 Chr.11: 128937839-128937839 on Build GRCh38
rs769293901 Chr.11: 128937832-128937832 on Build GRCh38

View Full Product Details