Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4627847-4628417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
UniGene
Hs.98397
Gene Chromosome Location
Chr.12: 4615508-4649047 on Build GRCh38
Transcripts
NM_001278309.1 NM_006422.3 XM_005253664.2 XM_011520908.1 XM_011520909.1 XM_011520910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1147017 c.806G>T p.R269L Chr.12: 4628096-4628096 on Build GRCh38
3384375 c.491A>T p.E164V Chr.12: 4628411-4628411 on Build GRCh38
1586497 c.631A>C p.N211H Chr.12: 4628271-4628271 on Build GRCh38
693867 c.806G>T p.R269L Chr.12: 4628096-4628096 on Build GRCh38
3461096 c.688C>T p.P230S Chr.12: 4628214-4628214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370012630 Chr.12: 4628288-4628288 on Build GRCh38
rs781778383 Chr.12: 4627909-4627909 on Build GRCh38
rs745982761 Chr.12: 4628290-4628290 on Build GRCh38
rs148501283 Chr.12: 4628114-4628114 on Build GRCh38
rs748232602 Chr.12: 4627910-4627910 on Build GRCh38
rs754233969 Chr.12: 4628277-4628277 on Build GRCh38
rs761811214 Chr.12: 4628212-4628212 on Build GRCh38
rs750027446 Chr.12: 4628125-4628125 on Build GRCh38
rs758136276 Chr.12: 4628134-4628134 on Build GRCh38
rs765777754 Chr.12: 4628371-4628371 on Build GRCh38
rs748606663 Chr.12: 4628050-4628050 on Build GRCh38
rs267603481 Chr.12: 4628021-4628021 on Build GRCh38
rs911468623 Chr.12: 4628017-4628017 on Build GRCh38
rs772869615 Chr.12: 4628229-4628229 on Build GRCh38
rs753084527 Chr.12: 4628255-4628255 on Build GRCh38
rs757388760 Chr.12: 4627972-4627972 on Build GRCh38
rs878881590 Chr.12: 4628029-4628029 on Build GRCh38
rs773177262 Chr.12: 4627927-4627927 on Build GRCh38
rs753313428 Chr.12: 4628004-4628004 on Build GRCh38
rs756897893 Chr.12: 4628025-4628025 on Build GRCh38
rs71579261 Chr.12: 4628238-4628238 on Build GRCh38
rs757778089 Chr.12: 4628285-4628285 on Build GRCh38
rs576355339 Chr.12: 4628069-4628069 on Build GRCh38
rs768265184 Chr.12: 4628315-4628315 on Build GRCh38
rs7960207 Chr.12: 4628152-4628152 on Build GRCh38
rs752700778 Chr.12: 4627958-4627958 on Build GRCh38
rs750484299 Chr.12: 4628382-4628382 on Build GRCh38
rs535694364 Chr.12: 4628031-4628031 on Build GRCh38
rs1037569414 Chr.12: 4628024-4628024 on Build GRCh38
rs7972737 Chr.12: 4627876-4627876 on Build GRCh38
rs749367824 Chr.12: 4628068-4628068 on Build GRCh38
rs140759485 Chr.12: 4628355-4628355 on Build GRCh38
rs1024772121 Chr.12: 4628160-4628160 on Build GRCh38
rs199862713 Chr.12: 4627869-4627869 on Build GRCh38
rs775279789 Chr.12: 4628094-4628094 on Build GRCh38
rs199990710 Chr.12: 4628115-4628115 on Build GRCh38
rs866329418 Chr.12: 4628170-4628170 on Build GRCh38
rs757348544 Chr.12: 4628161-4628161 on Build GRCh38
rs761284232 Chr.12: 4628122-4628122 on Build GRCh38
rs756494563 Chr.12: 4628260-4628260 on Build GRCh38
rs774102073 Chr.12: 4627940-4627940 on Build GRCh38
rs781477351 Chr.12: 4628102-4628103 on Build GRCh38
rs929095215 Chr.12: 4628319-4628319 on Build GRCh38
rs767430945 Chr.12: 4627949-4627949 on Build GRCh38
rs545087055 Chr.12: 4628096-4628096 on Build GRCh38
rs778475229 Chr.12: 4628026-4628026 on Build GRCh38
rs748309378 Chr.12: 4628104-4628105 on Build GRCh38
rs751955492 Chr.12: 4628030-4628030 on Build GRCh38
rs955522374 Chr.12: 4627996-4627996 on Build GRCh38
rs149898758 Chr.12: 4628342-4628342 on Build GRCh38
rs771790484 Chr.12: 4628079-4628079 on Build GRCh38
rs1017472614 Chr.12: 4627913-4627913 on Build GRCh38
rs746824078 Chr.12: 4628201-4628201 on Build GRCh38
rs369931580 Chr.12: 4627872-4627872 on Build GRCh38
rs769881921 Chr.12: 4628323-4628323 on Build GRCh38
rs775229840 Chr.12: 4628189-4628189 on Build GRCh38
rs768620386 Chr.12: 4628204-4628204 on Build GRCh38
rs376611820 Chr.12: 4628232-4628232 on Build GRCh38
rs774614928 Chr.12: 4628074-4628074 on Build GRCh38
rs773938534 Chr.12: 4628039-4628039 on Build GRCh38
rs759323301 Chr.12: 4628077-4628077 on Build GRCh38
rs202231561 Chr.12: 4628147-4628147 on Build GRCh38
rs764779667 Chr.12: 4628227-4628227 on Build GRCh38
rs561420027 Chr.12: 4628177-4628177 on Build GRCh38
rs528938252 Chr.12: 4628261-4628261 on Build GRCh38
rs771089765 Chr.12: 4627931-4627931 on Build GRCh38
rs774264124 Chr.12: 4628156-4628156 on Build GRCh38
rs199664599 Chr.12: 4627985-4627985 on Build GRCh38
rs770163349 Chr.12: 4628137-4628137 on Build GRCh38
rs71579260 Chr.12: 4628356-4628356 on Build GRCh38
rs770694375 Chr.12: 4628340-4628340 on Build GRCh38
rs201978763 Chr.12: 4628294-4628294 on Build GRCh38
rs374071910 Chr.12: 4627981-4627981 on Build GRCh38
rs776495333 Chr.12: 4628209-4628209 on Build GRCh38
rs755005394 Chr.12: 4628041-4628041 on Build GRCh38
rs749375251 Chr.12: 4628157-4628157 on Build GRCh38
rs763258102 Chr.12: 4628364-4628364 on Build GRCh38
rs762600685 Chr.12: 4628333-4628333 on Build GRCh38
rs151192998 Chr.12: 4628308-4628308 on Build GRCh38
rs769843586 Chr.12: 4627917-4627917 on Build GRCh38
rs753033294 Chr.12: 4628151-4628151 on Build GRCh38
rs139111565 Chr.12: 4628322-4628322 on Build GRCh38
rs776495822 Chr.12: 4628316-4628316 on Build GRCh38
rs772816579 Chr.12: 4628329-4628329 on Build GRCh38
rs890134517 Chr.12: 4628046-4628046 on Build GRCh38
rs868201722 Chr.12: 4628351-4628351 on Build GRCh38
rs756876609 Chr.12: 4627873-4627873 on Build GRCh38
rs758536949 Chr.12: 4628395-4628395 on Build GRCh38
rs574180493 Chr.12: 4627875-4627875 on Build GRCh38
rs769260852 Chr.12: 4627900-4627900 on Build GRCh38
rs144694533 Chr.12: 4627942-4627942 on Build GRCh38
rs746035800 Chr.12: 4628172-4628172 on Build GRCh38
rs2072356 Chr.12: 4628293-4628293 on Build GRCh38
rs149052688 Chr.12: 4627928-4627928 on Build GRCh38
rs778969713 Chr.12: 4628169-4628169 on Build GRCh38
rs35530750 Chr.12: 4628344-4628344 on Build GRCh38
rs781691588 Chr.12: 4628042-4628042 on Build GRCh38
rs748977283 Chr.12: 4627897-4627897 on Build GRCh38

View Full Product Details