Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56741306-56741860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIM1
Entrez Gene ID
5557
Gene Name
primase (DNA) subunit 1
Gene Aliases
p49
UniGene
Hs.534339
Gene Chromosome Location
Chr.12: 56731580-56752362 on Build GRCh38
Transcripts
NM_000946.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043721 c.982+2T>C p.? Chr.12: 56741433-56741433 on Build GRCh38
4645861 c.885G>A p.E295E Chr.12: 56741532-56741532 on Build GRCh38
1263034 c.956_957insAAGAG p.S319fs*34 Chr.12: 56741460-56741461 on Build GRCh38
5933120 c.877G>A p.E293K Chr.12: 56741540-56741540 on Build GRCh38
6755566 c.803G>A p.R268H Chr.12: 56741783-56741783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755595919 Chr.12: 56741463-56741463 on Build GRCh38
rs990234030 Chr.12: 56741763-56741763 on Build GRCh38
rs375009894 Chr.12: 56741709-56741709 on Build GRCh38
rs928901352 Chr.12: 56741799-56741799 on Build GRCh38
rs761936447 Chr.12: 56741414-56741414 on Build GRCh38
rs762003258 Chr.12: 56741572-56741572 on Build GRCh38
rs774680311 Chr.12: 56741782-56741782 on Build GRCh38
rs778674365 Chr.12: 56741520-56741520 on Build GRCh38
rs760694002 Chr.12: 56741553-56741553 on Build GRCh38
rs765555611 Chr.12: 56741426-56741426 on Build GRCh38
rs371616402 Chr.12: 56741541-56741541 on Build GRCh38
rs1009482314 Chr.12: 56741490-56741490 on Build GRCh38
rs376227854 Chr.12: 56741749-56741749 on Build GRCh38
rs964079466 Chr.12: 56741757-56741757 on Build GRCh38
rs914767335 Chr.12: 56741431-56741431 on Build GRCh38
rs764103216 Chr.12: 56741404-56741404 on Build GRCh38
rs755648405 Chr.12: 56741590-56741590 on Build GRCh38
rs767876815 Chr.12: 56741784-56741784 on Build GRCh38
rs991139528 Chr.12: 56741652-56741652 on Build GRCh38
rs746880210 Chr.12: 56741711-56741711 on Build GRCh38
rs540245510 Chr.12: 56741668-56741668 on Build GRCh38
rs934534109 Chr.12: 56741340-56741340 on Build GRCh38
rs780501437 Chr.12: 56741446-56741446 on Build GRCh38
rs375780109 Chr.12: 56741451-56741451 on Build GRCh38
rs766650501 Chr.12: 56741771-56741771 on Build GRCh38
rs756672193 Chr.12: 56741815-56741815 on Build GRCh38
rs758805858 Chr.12: 56741439-56741439 on Build GRCh38
rs1050978705 Chr.12: 56741455-56741455 on Build GRCh38
rs753262093 Chr.12: 56741610-56741610 on Build GRCh38
rs986864943 Chr.12: 56741383-56741383 on Build GRCh38
rs528335225 Chr.12: 56741611-56741611 on Build GRCh38
rs748891516 Chr.12: 56741496-56741496 on Build GRCh38
rs776561378 Chr.12: 56741570-56741570 on Build GRCh38
rs772111021 Chr.12: 56741386-56741386 on Build GRCh38
rs763170414 Chr.12: 56741770-56741770 on Build GRCh38
rs750021325 Chr.12: 56741826-56741826 on Build GRCh38
rs765348875 Chr.12: 56741589-56741589 on Build GRCh38
rs372561007 Chr.12: 56741401-56741402 on Build GRCh38
rs573227901 Chr.12: 56741630-56741630 on Build GRCh38
rs749231835 Chr.12: 56741708-56741709 on Build GRCh38
rs924542609 Chr.12: 56741331-56741331 on Build GRCh38
rs946300470 Chr.12: 56741435-56741435 on Build GRCh38
rs768706079 Chr.12: 56741566-56741566 on Build GRCh38
rs915650033 Chr.12: 56741662-56741662 on Build GRCh38
rs764852841 Chr.12: 56741820-56741820 on Build GRCh38
rs372314613 Chr.12: 56741391-56741391 on Build GRCh38
rs1030665437 Chr.12: 56741637-56741638 on Build GRCh38
rs571312182 Chr.12: 56741494-56741494 on Build GRCh38
rs760797591 Chr.12: 56741395-56741395 on Build GRCh38
rs750677819 Chr.12: 56741590-56741590 on Build GRCh38
rs751993130 Chr.12: 56741600-56741600 on Build GRCh38
rs202130551 Chr.12: 56741408-56741408 on Build GRCh38
rs750765821 Chr.12: 56741428-56741428 on Build GRCh38
rs772023116 Chr.12: 56741528-56741528 on Build GRCh38
rs373008443 Chr.12: 56741728-56741728 on Build GRCh38
rs778666611 Chr.12: 56741613-56741613 on Build GRCh38
rs555773069 Chr.12: 56741708-56741708 on Build GRCh38
rs781750340 Chr.12: 56741608-56741608 on Build GRCh38
rs763259191 Chr.12: 56741599-56741599 on Build GRCh38
rs770474523 Chr.12: 56741506-56741506 on Build GRCh38
rs745537875 Chr.12: 56741618-56741618 on Build GRCh38
rs200687851 Chr.12: 56741795-56741795 on Build GRCh38
rs934460743 Chr.12: 56741813-56741813 on Build GRCh38
rs369525260 Chr.12: 56741714-56741714 on Build GRCh38
rs745625683 Chr.12: 56741521-56741521 on Build GRCh38
rs769941710 Chr.12: 56741735-56741735 on Build GRCh38
rs554450645 Chr.12: 56741643-56741644 on Build GRCh38
rs889172565 Chr.12: 56741472-56741472 on Build GRCh38
rs748140323 Chr.12: 56741734-56741734 on Build GRCh38
rs749240240 Chr.12: 56741454-56741454 on Build GRCh38
rs1054359747 Chr.12: 56741402-56741402 on Build GRCh38
rs938352095 Chr.12: 56741508-56741508 on Build GRCh38
rs777716549 Chr.12: 56741707-56741710 on Build GRCh38
rs759991996 Chr.12: 56741783-56741783 on Build GRCh38
rs7971988 Chr.12: 56741602-56741602 on Build GRCh38

View Full Product Details