Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106378133-106378595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3B
Entrez Gene ID
55703
Gene Name
RNA polymerase III subunit B
Gene Aliases
C128, HLD8, INMAP, RPC2
UniGene
Hs.610795
Gene Chromosome Location
Chr.12: 106357658-106510198 on Build GRCh38
Transcripts
NM_001160708.1 NM_018082.5 XM_017019621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3455484 c.513T>G p.V171V Chr.12: 106378283-106378283 on Build GRCh38
1221512 c.571G>A p.V191M Chr.12: 106378341-106378341 on Build GRCh38
3455485 c.570C>T p.I190I Chr.12: 106378340-106378340 on Build GRCh38
5638015 c.516A>G p.K172K Chr.12: 106378286-106378286 on Build GRCh38
1167836 c.497-1_497insT p.G166fs*7 Chr.12: 106378266-106378267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772944396 Chr.12: 106378340-106378340 on Build GRCh38
rs750555479 Chr.12: 106378407-106378407 on Build GRCh38
rs779184269 Chr.12: 106378405-106378405 on Build GRCh38
rs777933519 Chr.12: 106378242-106378242 on Build GRCh38
rs759507297 Chr.12: 106378358-106378358 on Build GRCh38
rs377689476 Chr.12: 106378430-106378430 on Build GRCh38
rs757480678 Chr.12: 106378246-106378246 on Build GRCh38
rs190625048 Chr.12: 106378549-106378549 on Build GRCh38
rs769452308 Chr.12: 106378331-106378331 on Build GRCh38
rs113468238 Chr.12: 106378307-106378307 on Build GRCh38
rs747540999 Chr.12: 106378421-106378421 on Build GRCh38
rs751544859 Chr.12: 106378356-106378356 on Build GRCh38
rs758798622 Chr.12: 106378411-106378411 on Build GRCh38
rs772513749 Chr.12: 106378263-106378263 on Build GRCh38
rs934925748 Chr.12: 106378162-106378162 on Build GRCh38
rs116652787 Chr.12: 106378200-106378200 on Build GRCh38
rs761499674 Chr.12: 106378316-106378316 on Build GRCh38
rs943578872 Chr.12: 106378427-106378427 on Build GRCh38
rs1044511270 Chr.12: 106378533-106378533 on Build GRCh38
rs768190013 Chr.12: 106378297-106378297 on Build GRCh38
rs201641713 Chr.12: 106378256-106378256 on Build GRCh38
rs951415979 Chr.12: 106378476-106378476 on Build GRCh38
rs759926449 Chr.12: 106378559-106378559 on Build GRCh38
rs558836432 Chr.12: 106378545-106378545 on Build GRCh38
rs561398210 Chr.12: 106378528-106378528 on Build GRCh38
rs747452509 Chr.12: 106378285-106378285 on Build GRCh38
rs993472161 Chr.12: 106378156-106378156 on Build GRCh38
rs1038330332 Chr.12: 106378433-106378433 on Build GRCh38
rs780627697 Chr.12: 106378264-106378264 on Build GRCh38
rs1047730254 Chr.12: 106378514-106378514 on Build GRCh38
rs1000505464 Chr.12: 106378226-106378226 on Build GRCh38
rs988475856 Chr.12: 106378322-106378322 on Build GRCh38
rs1023408191 Chr.12: 106378278-106378278 on Build GRCh38
rs776074515 Chr.12: 106378311-106378311 on Build GRCh38
rs779185308 Chr.12: 106378252-106378252 on Build GRCh38
rs1019919928 Chr.12: 106378521-106378521 on Build GRCh38
rs796481541 Chr.12: 106378346-106378346 on Build GRCh38
rs370156173 Chr.12: 106378413-106378413 on Build GRCh38
rs895907595 Chr.12: 106378163-106378163 on Build GRCh38
rs753938916 Chr.12: 106378366-106378366 on Build GRCh38
rs764291006 Chr.12: 106378364-106378364 on Build GRCh38
rs201182565 Chr.12: 106378313-106378313 on Build GRCh38
rs10861587 Chr.12: 106378157-106378157 on Build GRCh38
rs531206642 Chr.12: 106378455-106378455 on Build GRCh38
rs756274726 Chr.12: 106378230-106378230 on Build GRCh38
rs11112959 Chr.12: 106378510-106378510 on Build GRCh38
rs777338765 Chr.12: 106378199-106378199 on Build GRCh38
rs965366368 Chr.12: 106378557-106378557 on Build GRCh38
rs147582475 Chr.12: 106378381-106378381 on Build GRCh38
rs186465519 Chr.12: 106378169-106378169 on Build GRCh38
rs114687611 Chr.12: 106378443-106378443 on Build GRCh38
rs545766074 Chr.12: 106378341-106378341 on Build GRCh38
rs574693305 Chr.12: 106378190-106378190 on Build GRCh38
rs766028971 Chr.12: 106378355-106378355 on Build GRCh38
rs754144327 Chr.12: 106378243-106378243 on Build GRCh38
rs895856715 Chr.12: 106378524-106378524 on Build GRCh38
rs115779133 Chr.12: 106378560-106378560 on Build GRCh38

View Full Product Details