Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34740938-34741536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA6
Entrez Gene ID
23054
Gene Name
nuclear receptor coactivator 6
Gene Aliases
AIB3, ASC2, NRC, PRIP, RAP250, TRBP
UniGene
Hs.368971
Gene Chromosome Location
Chr.20: 34714774-34825649 on Build GRCh38
Transcripts
NM_001242539.2 NM_001318240.1 NM_014071.4 XM_006723755.3 XM_011528721.2 XM_011528726.2 XM_011528727.2 XM_017027739.1 XM_017027740.1 XM_017027741.1 XM_017027742.1 XM_017027743.1 XM_017027744.1 XM_017027745.1 XM_017027746.1 XM_017027747.1 XM_017027748.1 XM_017027749.1 XM_017027750.1 XM_017027751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1026063 c.5189G>A p.R1730Q Chr.20: 34741067-34741067 on Build GRCh38
1026061 c.5252C>A p.S1751Y Chr.20: 34741004-34741004 on Build GRCh38
283144 c.5267C>A p.S1756Y Chr.20: 34740989-34740989 on Build GRCh38
4581744 c.5109T>A p.P1703P Chr.20: 34741147-34741147 on Build GRCh38
1026067 c.4901C>T p.A1634V Chr.20: 34741355-34741355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772538398 Chr.20: 34741427-34741427 on Build GRCh38
rs779552717 Chr.20: 34741407-34741407 on Build GRCh38
rs763826172 Chr.20: 34741510-34741510 on Build GRCh38
rs773864523 Chr.20: 34741017-34741017 on Build GRCh38
rs754728574 Chr.20: 34741114-34741114 on Build GRCh38
rs987893481 Chr.20: 34741333-34741333 on Build GRCh38
rs780525106 Chr.20: 34741400-34741400 on Build GRCh38
rs150352396 Chr.20: 34741056-34741056 on Build GRCh38
rs750413656 Chr.20: 34741089-34741089 on Build GRCh38
rs749706896 Chr.20: 34741311-34741311 on Build GRCh38
rs774785996 Chr.20: 34741330-34741330 on Build GRCh38
rs763414495 Chr.20: 34741148-34741148 on Build GRCh38
rs373830829 Chr.20: 34741462-34741462 on Build GRCh38
rs568132194 Chr.20: 34741063-34741063 on Build GRCh38
rs188126261 Chr.20: 34741426-34741426 on Build GRCh38
rs932253082 Chr.20: 34741430-34741430 on Build GRCh38
rs964160985 Chr.20: 34741480-34741480 on Build GRCh38
rs200880705 Chr.20: 34741147-34741147 on Build GRCh38
rs758291443 Chr.20: 34741148-34741148 on Build GRCh38
rs777390851 Chr.20: 34741272-34741272 on Build GRCh38
rs751717705 Chr.20: 34740981-34740981 on Build GRCh38
rs1040658058 Chr.20: 34740969-34740969 on Build GRCh38
rs372899926 Chr.20: 34741097-34741097 on Build GRCh38
rs759987385 Chr.20: 34741053-34741053 on Build GRCh38
rs756811793 Chr.20: 34741393-34741393 on Build GRCh38
rs778632318 Chr.20: 34741218-34741218 on Build GRCh38
rs919615252 Chr.20: 34741125-34741125 on Build GRCh38
rs771540987 Chr.20: 34741041-34741041 on Build GRCh38
rs962021855 Chr.20: 34741138-34741138 on Build GRCh38
rs758578211 Chr.20: 34740970-34740970 on Build GRCh38
rs766668035 Chr.20: 34741009-34741009 on Build GRCh38
rs747151159 Chr.20: 34741441-34741441 on Build GRCh38
rs752378178 Chr.20: 34741235-34741235 on Build GRCh38
rs746386264 Chr.20: 34741142-34741142 on Build GRCh38
rs779731696 Chr.20: 34741110-34741110 on Build GRCh38
rs201837457 Chr.20: 34741162-34741162 on Build GRCh38
rs755991725 Chr.20: 34740993-34740993 on Build GRCh38
rs766409894 Chr.20: 34741191-34741191 on Build GRCh38
rs769094280 Chr.20: 34741317-34741317 on Build GRCh38
rs534399488 Chr.20: 34741140-34741140 on Build GRCh38
rs181826821 Chr.20: 34741305-34741305 on Build GRCh38
rs753726700 Chr.20: 34741111-34741111 on Build GRCh38
rs142682209 Chr.20: 34741354-34741354 on Build GRCh38
rs368111032 Chr.20: 34741083-34741083 on Build GRCh38
rs137971603 Chr.20: 34741353-34741353 on Build GRCh38
rs779855865 Chr.20: 34741000-34741000 on Build GRCh38
rs879142252 Chr.20: 34741294-34741294 on Build GRCh38
rs747726440 Chr.20: 34741026-34741026 on Build GRCh38
rs1046194642 Chr.20: 34741225-34741225 on Build GRCh38
rs753690833 Chr.20: 34741198-34741198 on Build GRCh38
rs183716498 Chr.20: 34741419-34741419 on Build GRCh38
rs149244697 Chr.20: 34741404-34741404 on Build GRCh38
rs757406536 Chr.20: 34740982-34740982 on Build GRCh38
rs759936275 Chr.20: 34740960-34740960 on Build GRCh38
rs781094788 Chr.20: 34741506-34741506 on Build GRCh38
rs767705615 Chr.20: 34741168-34741168 on Build GRCh38
rs377670575 Chr.20: 34741167-34741167 on Build GRCh38
rs570655118 Chr.20: 34741115-34741115 on Build GRCh38
rs773565821 Chr.20: 34741341-34741341 on Build GRCh38
rs754852497 Chr.20: 34741210-34741210 on Build GRCh38
rs200324221 Chr.20: 34741339-34741339 on Build GRCh38
rs763214234 Chr.20: 34741057-34741057 on Build GRCh38
rs146019486 Chr.20: 34741355-34741355 on Build GRCh38
rs762030650 Chr.20: 34741070-34741070 on Build GRCh38
rs759631975 Chr.20: 34741032-34741032 on Build GRCh38
rs942936047 Chr.20: 34741107-34741107 on Build GRCh38
rs141876445 Chr.20: 34741377-34741377 on Build GRCh38
rs774734579 Chr.20: 34741166-34741166 on Build GRCh38
rs777225494 Chr.20: 34741141-34741141 on Build GRCh38
rs770872136 Chr.20: 34741460-34741461 on Build GRCh38
rs766347909 Chr.20: 34741344-34741344 on Build GRCh38
rs529099357 Chr.20: 34741504-34741504 on Build GRCh38
rs754115896 Chr.20: 34741010-34741010 on Build GRCh38
rs757980579 Chr.20: 34741252-34741252 on Build GRCh38
rs986985690 Chr.20: 34741087-34741087 on Build GRCh38
rs996354389 Chr.20: 34741044-34741044 on Build GRCh38
rs375266173 Chr.20: 34741067-34741067 on Build GRCh38
rs376737608 Chr.20: 34741137-34741137 on Build GRCh38
rs765223736 Chr.20: 34741496-34741496 on Build GRCh38
rs749801791 Chr.20: 34741403-34741403 on Build GRCh38
rs764095103 Chr.20: 34740977-34740977 on Build GRCh38
rs201602909 Chr.20: 34741313-34741313 on Build GRCh38
rs368564764 Chr.20: 34741086-34741086 on Build GRCh38
rs139829908 Chr.20: 34741365-34741365 on Build GRCh38
rs576965755 Chr.20: 34741307-34741307 on Build GRCh38
rs371013508 Chr.20: 34741301-34741301 on Build GRCh38
rs775373517 Chr.20: 34741499-34741499 on Build GRCh38
rs912810067 Chr.20: 34741095-34741095 on Build GRCh38
rs748592578 Chr.20: 34741422-34741422 on Build GRCh38
rs750475329 Chr.20: 34741180-34741180 on Build GRCh38
rs148432377 Chr.20: 34741452-34741452 on Build GRCh38
rs552060664 Chr.20: 34740968-34740968 on Build GRCh38
rs745776743 Chr.20: 34740988-34740988 on Build GRCh38
rs1014853897 Chr.20: 34741122-34741122 on Build GRCh38
rs980618446 Chr.20: 34741260-34741260 on Build GRCh38
rs1020829004 Chr.20: 34741385-34741385 on Build GRCh38
rs764442540 Chr.20: 34741065-34741065 on Build GRCh38
rs763774739 Chr.20: 34741366-34741366 on Build GRCh38
rs781315597 Chr.20: 34740983-34740983 on Build GRCh38
rs752545586 Chr.20: 34741364-34741364 on Build GRCh38
rs777172496 Chr.20: 34741043-34741043 on Build GRCh38
rs142540483 Chr.20: 34741505-34741505 on Build GRCh38
rs760661602 Chr.20: 34741189-34741189 on Build GRCh38
rs771074082 Chr.20: 34741445-34741445 on Build GRCh38

View Full Product Details