Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37714434-37715026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ6
Entrez Gene ID
3763
Gene Name
potassium voltage-gated channel subfamily J member 6
Gene Aliases
BIR1, GIRK-2, GIRK2, KATP-2, KATP2, KCNJ7, KIR3.2, KPLBS, hiGIRK2
UniGene
Hs.626242
Gene Chromosome Location
Chr.21: 37624223-37916438 on Build GRCh38
Transcripts
NM_002240.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3912087 c.484G>A p.D162N Chr.21: 37714673-37714673 on Build GRCh38
1255353 c.309C>A p.T103T Chr.21: 37714848-37714848 on Build GRCh38
6004755 c.147C>A p.T49T Chr.21: 37715010-37715010 on Build GRCh38
317501 c.491G>T p.C164F Chr.21: 37714666-37714666 on Build GRCh38
6691199 c.168C>T p.Y56Y Chr.21: 37714989-37714989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910998272 Chr.21: 37714896-37714896 on Build GRCh38
rs776790699 Chr.21: 37714619-37714619 on Build GRCh38
rs59497335 Chr.21: 37714686-37714686 on Build GRCh38
rs770961576 Chr.21: 37715006-37715006 on Build GRCh38
rs773044664 Chr.21: 37714812-37714812 on Build GRCh38
rs972394919 Chr.21: 37714805-37714805 on Build GRCh38
rs769894534 Chr.21: 37714809-37714809 on Build GRCh38
rs2070995 Chr.21: 37714662-37714662 on Build GRCh38
rs368206270 Chr.21: 37714663-37714663 on Build GRCh38
rs372624887 Chr.21: 37714635-37714635 on Build GRCh38
rs750928265 Chr.21: 37714923-37714923 on Build GRCh38
rs866066492 Chr.21: 37714975-37714975 on Build GRCh38
rs780695680 Chr.21: 37714684-37714684 on Build GRCh38
rs762020565 Chr.21: 37714801-37714801 on Build GRCh38
rs545127360 Chr.21: 37714596-37714596 on Build GRCh38
rs758878702 Chr.21: 37714944-37714944 on Build GRCh38
rs761967457 Chr.21: 37714629-37714629 on Build GRCh38
rs779332289 Chr.21: 37714746-37714746 on Build GRCh38
rs201523497 Chr.21: 37714628-37714628 on Build GRCh38
rs999857210 Chr.21: 37714976-37714976 on Build GRCh38
rs776985089 Chr.21: 37714797-37714797 on Build GRCh38
rs374502191 Chr.21: 37714539-37714539 on Build GRCh38
rs746288638 Chr.21: 37714548-37714548 on Build GRCh38
rs1050781248 Chr.21: 37714889-37714889 on Build GRCh38
rs971698781 Chr.21: 37714955-37714955 on Build GRCh38
rs769054665 Chr.21: 37714786-37714786 on Build GRCh38
rs201815398 Chr.21: 37714950-37714950 on Build GRCh38
rs755674854 Chr.21: 37714904-37714904 on Build GRCh38
rs746367388 Chr.21: 37714751-37714751 on Build GRCh38
rs750342789 Chr.21: 37714647-37714647 on Build GRCh38
rs200572263 Chr.21: 37714996-37714996 on Build GRCh38
rs984972371 Chr.21: 37714734-37714734 on Build GRCh38
rs781613861 Chr.21: 37714971-37714971 on Build GRCh38
rs772455545 Chr.21: 37714580-37714580 on Build GRCh38
rs866623696 Chr.21: 37714622-37714622 on Build GRCh38
rs771371207 Chr.21: 37714516-37714516 on Build GRCh38
rs909444797 Chr.21: 37714779-37714779 on Build GRCh38
rs772348723 Chr.21: 37714755-37714755 on Build GRCh38
rs372472729 Chr.21: 37714914-37714914 on Build GRCh38
rs767224800 Chr.21: 37714857-37714857 on Build GRCh38
rs962612911 Chr.21: 37714804-37714804 on Build GRCh38
rs368653927 Chr.21: 37714584-37714584 on Build GRCh38
rs769107756 Chr.21: 37714956-37714956 on Build GRCh38
rs780410267 Chr.21: 37714949-37714949 on Build GRCh38
rs933696223 Chr.21: 37714832-37714832 on Build GRCh38
rs759313073 Chr.21: 37714827-37714827 on Build GRCh38
rs200072381 Chr.21: 37714888-37714888 on Build GRCh38
rs780429357 Chr.21: 37714759-37714759 on Build GRCh38
rs369579195 Chr.21: 37714977-37714977 on Build GRCh38
rs960787396 Chr.21: 37714527-37714527 on Build GRCh38
rs749801967 Chr.21: 37714503-37714503 on Build GRCh38
rs755617760 Chr.21: 37714713-37714713 on Build GRCh38
rs377354413 Chr.21: 37714947-37714947 on Build GRCh38
rs762813268 Chr.21: 37715005-37715005 on Build GRCh38
rs148842116 Chr.21: 37714989-37714989 on Build GRCh38
rs572096844 Chr.21: 37714815-37714815 on Build GRCh38
rs747245357 Chr.21: 37714785-37714785 on Build GRCh38
rs986283475 Chr.21: 37714872-37714872 on Build GRCh38
rs754560406 Chr.21: 37714681-37714681 on Build GRCh38
rs751200771 Chr.21: 37714676-37714676 on Build GRCh38
rs752318891 Chr.21: 37714890-37714890 on Build GRCh38
rs762906480 Chr.21: 37714814-37714814 on Build GRCh38
rs764270794 Chr.21: 37714599-37714599 on Build GRCh38
rs143219969 Chr.21: 37714922-37714922 on Build GRCh38
rs786204795 Chr.21: 37714697-37714697 on Build GRCh38
rs751298042 Chr.21: 37714822-37714822 on Build GRCh38
rs786204794 Chr.21: 37714700-37714702 on Build GRCh38
rs200348437 Chr.21: 37715000-37715000 on Build GRCh38

View Full Product Details