Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26452079-26452637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

SNP Detail

SNP ID Chromosome Location
rs750142480 Chr.22: 26452249-26452253 on Build GRCh38
rs755148260 Chr.22: 26452257-26452257 on Build GRCh38
rs999563412 Chr.22: 26452258-26452258 on Build GRCh38
rs890751270 Chr.22: 26452355-26452355 on Build GRCh38
rs529938159 Chr.22: 26452500-26452500 on Build GRCh38
rs143369347 Chr.22: 26452361-26452361 on Build GRCh38
rs971837822 Chr.22: 26452477-26452477 on Build GRCh38
rs886057312 Chr.22: 26452113-26452113 on Build GRCh38
rs752453853 Chr.22: 26452350-26452350 on Build GRCh38
rs927560561 Chr.22: 26452322-26452322 on Build GRCh38
rs752723057 Chr.22: 26452214-26452214 on Build GRCh38
rs561983395 Chr.22: 26452485-26452485 on Build GRCh38
rs952090234 Chr.22: 26452164-26452164 on Build GRCh38
rs200099628 Chr.22: 26452439-26452439 on Build GRCh38
rs909615654 Chr.22: 26452365-26452365 on Build GRCh38
rs548085200 Chr.22: 26452615-26452615 on Build GRCh38
rs887874663 Chr.22: 26452391-26452391 on Build GRCh38
rs886057313 Chr.22: 26452205-26452209 on Build GRCh38
rs937547471 Chr.22: 26452323-26452323 on Build GRCh38
rs779035400 Chr.22: 26452260-26452260 on Build GRCh38
rs778332651 Chr.22: 26452309-26452309 on Build GRCh38
rs922906194 Chr.22: 26452343-26452343 on Build GRCh38
rs977104076 Chr.22: 26452563-26452563 on Build GRCh38
rs749384453 Chr.22: 26452506-26452506 on Build GRCh38
rs967101864 Chr.22: 26452561-26452561 on Build GRCh38
rs779719192 Chr.22: 26452319-26452319 on Build GRCh38
rs758423479 Chr.22: 26452297-26452297 on Build GRCh38
rs945772611 Chr.22: 26452348-26452348 on Build GRCh38
rs187297219 Chr.22: 26452464-26452464 on Build GRCh38
rs200772835 Chr.22: 26452349-26452349 on Build GRCh38
rs915722901 Chr.22: 26452379-26452379 on Build GRCh38
rs746258061 Chr.22: 26452412-26452412 on Build GRCh38
rs1024197400 Chr.22: 26452162-26452162 on Build GRCh38
rs903137577 Chr.22: 26452440-26452440 on Build GRCh38
rs1018412279 Chr.22: 26452301-26452301 on Build GRCh38
rs1048244353 Chr.22: 26452380-26452380 on Build GRCh38
rs762464617 Chr.22: 26452459-26452459 on Build GRCh38
rs770969523 Chr.22: 26452289-26452289 on Build GRCh38
rs575200307 Chr.22: 26452136-26452136 on Build GRCh38
rs747584684 Chr.22: 26452329-26452329 on Build GRCh38
rs181998459 Chr.22: 26452338-26452338 on Build GRCh38
rs867158897 Chr.22: 26452604-26452604 on Build GRCh38
rs62225811 Chr.22: 26452352-26452352 on Build GRCh38
rs777878254 Chr.22: 26452515-26452515 on Build GRCh38
rs999924351 Chr.22: 26452461-26452461 on Build GRCh38
rs1014910744 Chr.22: 26452287-26452287 on Build GRCh38
rs1046049821 Chr.22: 26452431-26452431 on Build GRCh38
rs752755928 Chr.22: 26452264-26452264 on Build GRCh38
rs989570182 Chr.22: 26452294-26452294 on Build GRCh38
rs756466974 Chr.22: 26452473-26452473 on Build GRCh38
rs575920373 Chr.22: 26452392-26452392 on Build GRCh38
rs546286332 Chr.22: 26452433-26452433 on Build GRCh38
rs958180873 Chr.22: 26452292-26452292 on Build GRCh38
rs943975254 Chr.22: 26452360-26452360 on Build GRCh38
rs546959109 Chr.22: 26452483-26452483 on Build GRCh38
rs914716855 Chr.22: 26452532-26452532 on Build GRCh38
rs755051224 Chr.22: 26452507-26452507 on Build GRCh38
rs990261877 Chr.22: 26452324-26452324 on Build GRCh38
rs767814877 Chr.22: 26452371-26452371 on Build GRCh38
rs949903292 Chr.22: 26452410-26452411 on Build GRCh38
rs912304266 Chr.22: 26452593-26452593 on Build GRCh38
rs995770536 Chr.22: 26452373-26452373 on Build GRCh38
rs983479129 Chr.22: 26452278-26452278 on Build GRCh38
rs190837977 Chr.22: 26452482-26452482 on Build GRCh38

View Full Product Details