Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84441061-84441446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCLG1
Entrez Gene ID
8802
Gene Name
succinate-CoA ligase alpha subunit
Gene Aliases
GALPHA, MTDPS9, SUCLA1
UniGene
Hs.270428
Gene Chromosome Location
Chr.2: 84423523-84459462 on Build GRCh38
Transcripts
NM_003849.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424319 c.403G>A p.A135T Chr.2: 84441375-84441375 on Build GRCh38
6792783 c.428T>C p.I143T Chr.2: 84441350-84441350 on Build GRCh38
6792785 c.380C>T p.A127V Chr.2: 84441398-84441398 on Build GRCh38
6509548 c.498G>T p.R166S Chr.2: 84441280-84441280 on Build GRCh38
4841976 c.429C>T p.I143I Chr.2: 84441349-84441349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746067777 Chr.2: 84441379-84441379 on Build GRCh38
rs367999572 Chr.2: 84441135-84441135 on Build GRCh38
rs181953896 Chr.2: 84441426-84441426 on Build GRCh38
rs267607098 Chr.2: 84441330-84441330 on Build GRCh38
rs978216090 Chr.2: 84441217-84441217 on Build GRCh38
rs80272300 Chr.2: 84441196-84441196 on Build GRCh38
rs374072294 Chr.2: 84441252-84441252 on Build GRCh38
rs778953778 Chr.2: 84441113-84441113 on Build GRCh38
rs759773925 Chr.2: 84441137-84441137 on Build GRCh38
rs760848966 Chr.2: 84441203-84441203 on Build GRCh38
rs760113821 Chr.2: 84441414-84441414 on Build GRCh38
rs766931758 Chr.2: 84441406-84441406 on Build GRCh38
rs768770321 Chr.2: 84441239-84441239 on Build GRCh38
rs539338118 Chr.2: 84441168-84441168 on Build GRCh38
rs933451922 Chr.2: 84441317-84441317 on Build GRCh38
rs773231074 Chr.2: 84441405-84441406 on Build GRCh38
rs775892969 Chr.2: 84441272-84441272 on Build GRCh38
rs778891473 Chr.2: 84441422-84441422 on Build GRCh38
rs763324017 Chr.2: 84441276-84441276 on Build GRCh38
rs141331864 Chr.2: 84441297-84441297 on Build GRCh38
rs769785912 Chr.2: 84441231-84441232 on Build GRCh38
rs1038752285 Chr.2: 84441275-84441275 on Build GRCh38
rs749283854 Chr.2: 84441094-84441094 on Build GRCh38
rs755842451 Chr.2: 84441224-84441224 on Build GRCh38
rs200067153 Chr.2: 84441221-84441221 on Build GRCh38
rs779980220 Chr.2: 84441305-84441305 on Build GRCh38
rs899069810 Chr.2: 84441300-84441300 on Build GRCh38
rs747047204 Chr.2: 84441251-84441251 on Build GRCh38
rs770889401 Chr.2: 84441338-84441338 on Build GRCh38
rs959644451 Chr.2: 84441176-84441176 on Build GRCh38
rs1035507077 Chr.2: 84441411-84441411 on Build GRCh38
rs753567873 Chr.2: 84441420-84441420 on Build GRCh38
rs374859194 Chr.2: 84441211-84441211 on Build GRCh38
rs758971676 Chr.2: 84441151-84441151 on Build GRCh38
rs781403752 Chr.2: 84441351-84441351 on Build GRCh38
rs760236068 Chr.2: 84441114-84441116 on Build GRCh38
rs542629113 Chr.2: 84441384-84441384 on Build GRCh38
rs1001604141 Chr.2: 84441115-84441115 on Build GRCh38
rs369591174 Chr.2: 84441232-84441232 on Build GRCh38
rs371630011 Chr.2: 84441419-84441419 on Build GRCh38
rs555605613 Chr.2: 84441164-84441164 on Build GRCh38
rs376171433 Chr.2: 84441383-84441383 on Build GRCh38
rs200978645 Chr.2: 84441139-84441139 on Build GRCh38
rs1033544285 Chr.2: 84441127-84441127 on Build GRCh38
rs766447883 Chr.2: 84441118-84441118 on Build GRCh38
rs201717664 Chr.2: 84441413-84441413 on Build GRCh38
rs797046017 Chr.2: 84441271-84441271 on Build GRCh38
rs754798729 Chr.2: 84441421-84441421 on Build GRCh38
rs34359115 Chr.2: 84441144-84441144 on Build GRCh38
rs577461564 Chr.2: 84441285-84441285 on Build GRCh38
rs776695932 Chr.2: 84441259-84441259 on Build GRCh38
rs778946494 Chr.2: 84441326-84441326 on Build GRCh38
rs140626260 Chr.2: 84441088-84441088 on Build GRCh38
rs146102400 Chr.2: 84441402-84441402 on Build GRCh38
rs753172949 Chr.2: 84441143-84441143 on Build GRCh38
rs753622816 Chr.2: 84441306-84441306 on Build GRCh38
rs772243831 Chr.2: 84441104-84441104 on Build GRCh38
rs774436508 Chr.2: 84441243-84441243 on Build GRCh38
rs776946908 Chr.2: 84441124-84441124 on Build GRCh38
rs943015543 Chr.2: 84441295-84441295 on Build GRCh38
rs575207227 Chr.2: 84441381-84441381 on Build GRCh38
rs149419193 Chr.2: 84441350-84441350 on Build GRCh38
rs775710166 Chr.2: 84441134-84441135 on Build GRCh38
rs759846791 Chr.2: 84441261-84441261 on Build GRCh38
rs373372300 Chr.2: 84441177-84441177 on Build GRCh38
rs779938026 Chr.2: 84441231-84441231 on Build GRCh38
rs369153479 Chr.2: 84441318-84441318 on Build GRCh38
rs773489950 Chr.2: 84441108-84441108 on Build GRCh38
rs769028246 Chr.2: 84441393-84441393 on Build GRCh38
rs200415851 Chr.2: 84441213-84441213 on Build GRCh38
rs11555948 Chr.2: 84441293-84441293 on Build GRCh38
rs1012182134 Chr.2: 84441416-84441416 on Build GRCh38
rs146551543 Chr.2: 84441296-84441296 on Build GRCh38
rs760853032 Chr.2: 84441114-84441114 on Build GRCh38
rs765519303 Chr.2: 84441266-84441266 on Build GRCh38
rs373153604 Chr.2: 84441334-84441334 on Build GRCh38
rs764515402 Chr.2: 84441283-84441283 on Build GRCh38
rs139731592 Chr.2: 84441394-84441394 on Build GRCh38
rs267607099 Chr.2: 84441269-84441269 on Build GRCh38
rs751010719 Chr.2: 84441200-84441200 on Build GRCh38

View Full Product Details